Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 125577953-125578461 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EI24
Entrez Gene ID
9538
Gene Name
EI24, autophagy associated transmembrane protein
Gene Aliases
EPG4, PIG8, TP53I8
UniGene
Hs.643514
Gene Chromosome Location
Chr.11: 125569216-125584689 on Build GRCh38
Transcripts
NM_001290135.1 NM_004879.4 NR_110769.1 NR_110770.1 XM_011543069.1 XM_011543070.1 XM_017018582.1 NR_132372.1

Gene Symbol
STT3A-AS1
Entrez Gene ID
105369550
Gene Name
STT3A antisense RNA 1
Gene Aliases
-
UniGene
Hs.732083
Gene Chromosome Location
Chr.11: 125568108-125592568 on Build GRCh38
Transcripts
NM_001290135.1 NM_004879.4 NR_110769.1 NR_110770.1 XM_011543069.1 XM_011543070.1 XM_017018582.1 NR_132372.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6337104 c.380G>A p.S127N Chr.11: 125578196-125578196 on Build GRCh38
1585478 c.394C>T p.L132F Chr.11: 125578210-125578210 on Build GRCh38
6377557 c.325T>A p.S109T Chr.11: 125578141-125578141 on Build GRCh38
4439522 c.366C>T p.L122L Chr.11: 125578182-125578182 on Build GRCh38
4999914 c.350G>A p.W117* Chr.11: 125578166-125578166 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369795047 Chr.11: 125578104-125578104 on Build GRCh38
rs183284349 Chr.11: 125578350-125578350 on Build GRCh38
rs775383756 Chr.11: 125578302-125578302 on Build GRCh38
rs752294064 Chr.11: 125578017-125578017 on Build GRCh38
rs148165280 Chr.11: 125578085-125578085 on Build GRCh38
rs1026178654 Chr.11: 125578351-125578351 on Build GRCh38
rs769113421 Chr.11: 125578179-125578179 on Build GRCh38
rs1039992789 Chr.11: 125578074-125578074 on Build GRCh38
rs747556759 Chr.11: 125578176-125578176 on Build GRCh38
rs768225838 Chr.11: 125578089-125578089 on Build GRCh38
rs867200337 Chr.11: 125578185-125578185 on Build GRCh38
rs997139117 Chr.11: 125578382-125578382 on Build GRCh38
rs955380254 Chr.11: 125578402-125578402 on Build GRCh38
rs1017162860 Chr.11: 125578011-125578011 on Build GRCh38
rs3831427 Chr.11: 125578092-125578099 on Build GRCh38
rs747713258 Chr.11: 125578230-125578230 on Build GRCh38
rs535678301 Chr.11: 125578081-125578081 on Build GRCh38
rs781717797 Chr.11: 125578184-125578184 on Build GRCh38
rs545799656 Chr.11: 125578058-125578058 on Build GRCh38
rs557210479 Chr.11: 125578371-125578371 on Build GRCh38
rs534625909 Chr.11: 125578163-125578163 on Build GRCh38
rs558889722 Chr.11: 125578319-125578319 on Build GRCh38
rs376488987 Chr.11: 125578103-125578103 on Build GRCh38
rs748896892 Chr.11: 125578262-125578262 on Build GRCh38
rs757125987 Chr.11: 125578312-125578312 on Build GRCh38
rs1002225857 Chr.11: 125578267-125578267 on Build GRCh38
rs74331859 Chr.11: 125578109-125578109 on Build GRCh38
rs975814472 Chr.11: 125578043-125578043 on Build GRCh38
rs977840466 Chr.11: 125578418-125578418 on Build GRCh38
rs924666251 Chr.11: 125578440-125578440 on Build GRCh38
rs187809509 Chr.11: 125578429-125578429 on Build GRCh38
rs773025733 Chr.11: 125578248-125578248 on Build GRCh38
rs191070639 Chr.11: 125578073-125578073 on Build GRCh38
rs758787371 Chr.11: 125578168-125578168 on Build GRCh38
rs984747859 Chr.11: 125578385-125578385 on Build GRCh38
rs376481083 Chr.11: 125578290-125578290 on Build GRCh38
rs1030061518 Chr.11: 125578394-125578394 on Build GRCh38
rs376911502 Chr.11: 125578110-125578110 on Build GRCh38
rs764238263 Chr.11: 125578139-125578139 on Build GRCh38
rs374580824 Chr.11: 125578143-125578143 on Build GRCh38
rs1043918 Chr.11: 125578172-125578172 on Build GRCh38
rs769401768 Chr.11: 125578242-125578242 on Build GRCh38
rs767410233 Chr.11: 125578299-125578299 on Build GRCh38
rs757313787 Chr.11: 125578161-125578161 on Build GRCh38
rs2849219 Chr.11: 125578410-125578410 on Build GRCh38
rs564111061 Chr.11: 125578289-125578289 on Build GRCh38
rs145115670 Chr.11: 125578101-125578101 on Build GRCh38
rs1017151498 Chr.11: 125578372-125578372 on Build GRCh38
rs759330627 Chr.11: 125578126-125578126 on Build GRCh38
rs1043919 Chr.11: 125578201-125578201 on Build GRCh38
rs770771291 Chr.11: 125578281-125578281 on Build GRCh38
rs370510644 Chr.11: 125578117-125578117 on Build GRCh38
rs140296039 Chr.11: 125578164-125578164 on Build GRCh38
rs773948216 Chr.11: 125578285-125578285 on Build GRCh38
rs964279058 Chr.11: 125578379-125578379 on Build GRCh38
rs1049441263 Chr.11: 125578003-125578003 on Build GRCh38
rs550498776 Chr.11: 125578293-125578293 on Build GRCh38
rs761407925 Chr.11: 125578102-125578102 on Build GRCh38

View Full Product Details