Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 9852863-9853422 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLEC2B
Entrez Gene ID
9976
Gene Name
C-type lectin domain family 2 member B
Gene Aliases
AICL, CLECSF2, HP10085, IFNRG1
UniGene
Hs.85201
Gene Chromosome Location
Chr.12: 9852369-9869859 on Build GRCh38
Transcripts
NM_005127.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4037851 c.418A>C p.R140R Chr.12: 9853332-9853332 on Build GRCh38
3454911 c.382G>A p.D128N Chr.12: 9853368-9853368 on Build GRCh38
3687888 c.351G>A p.M117I Chr.12: 9853399-9853399 on Build GRCh38
5684483 c.448T>G p.*150E Chr.12: 9853302-9853302 on Build GRCh38
6355737 c.360T>C p.S120S Chr.12: 9853390-9853390 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs923225134 Chr.12: 9853247-9853247 on Build GRCh38
rs764065996 Chr.12: 9853267-9853267 on Build GRCh38
rs540501947 Chr.12: 9853258-9853258 on Build GRCh38
rs140473877 Chr.12: 9853323-9853323 on Build GRCh38
rs10649717 Chr.12: 9853071-9853072 on Build GRCh38
rs902756467 Chr.12: 9853139-9853139 on Build GRCh38
rs555836324 Chr.12: 9853053-9853053 on Build GRCh38
rs774782334 Chr.12: 9853356-9853356 on Build GRCh38
rs1057091309 Chr.12: 9853348-9853348 on Build GRCh38
rs61914639 Chr.12: 9853089-9853089 on Build GRCh38
rs61914640 Chr.12: 9853093-9853093 on Build GRCh38
rs914310982 Chr.12: 9853311-9853311 on Build GRCh38
rs543200717 Chr.12: 9853178-9853178 on Build GRCh38
rs916674395 Chr.12: 9853058-9853058 on Build GRCh38
rs976368650 Chr.12: 9853235-9853235 on Build GRCh38
rs151287816 Chr.12: 9852952-9852954 on Build GRCh38
rs962489415 Chr.12: 9853115-9853117 on Build GRCh38
rs746875604 Chr.12: 9853075-9853076 on Build GRCh38
rs770427068 Chr.12: 9853361-9853361 on Build GRCh38
rs868826020 Chr.12: 9853111-9853111 on Build GRCh38
rs762316141 Chr.12: 9853358-9853358 on Build GRCh38
rs759086789 Chr.12: 9853369-9853369 on Build GRCh38
rs14999 Chr.12: 9853362-9853362 on Build GRCh38
rs768925278 Chr.12: 9853346-9853346 on Build GRCh38
rs971382458 Chr.12: 9852926-9852926 on Build GRCh38
rs751560856 Chr.12: 9853293-9853293 on Build GRCh38
rs534323668 Chr.12: 9853114-9853114 on Build GRCh38
rs538910460 Chr.12: 9852992-9852992 on Build GRCh38
rs930618573 Chr.12: 9853063-9853063 on Build GRCh38
rs978271662 Chr.12: 9853006-9853006 on Build GRCh38
rs950876558 Chr.12: 9853031-9853031 on Build GRCh38
rs878994865 Chr.12: 9853278-9853278 on Build GRCh38
rs375085537 Chr.12: 9853109-9853109 on Build GRCh38
rs892908511 Chr.12: 9853113-9853113 on Build GRCh38
rs749625504 Chr.12: 9853345-9853345 on Build GRCh38
rs377043184 Chr.12: 9853070-9853071 on Build GRCh38
rs545687364 Chr.12: 9853214-9853214 on Build GRCh38
rs764929468 Chr.12: 9853252-9853252 on Build GRCh38
rs115682932 Chr.12: 9853147-9853147 on Build GRCh38
rs1051467510 Chr.12: 9853160-9853160 on Build GRCh38
rs764977467 Chr.12: 9853383-9853383 on Build GRCh38
rs2594072 Chr.12: 9853179-9853179 on Build GRCh38
rs370105265 Chr.12: 9853087-9853087 on Build GRCh38
rs931778287 Chr.12: 9853291-9853291 on Build GRCh38
rs182560494 Chr.12: 9853044-9853044 on Build GRCh38
rs371186604 Chr.12: 9853079-9853080 on Build GRCh38
rs962071346 Chr.12: 9853230-9853230 on Build GRCh38
rs866878658 Chr.12: 9853057-9853057 on Build GRCh38
rs753321760 Chr.12: 9852955-9852955 on Build GRCh38
rs1011259382 Chr.12: 9852950-9852950 on Build GRCh38
rs778578195 Chr.12: 9853159-9853159 on Build GRCh38
rs776179456 Chr.12: 9853368-9853368 on Build GRCh38
rs984777458 Chr.12: 9853071-9853071 on Build GRCh38
rs148210022 Chr.12: 9853075-9853075 on Build GRCh38
rs947154216 Chr.12: 9853316-9853316 on Build GRCh38
rs565580719 Chr.12: 9853220-9853220 on Build GRCh38
rs892771620 Chr.12: 9852938-9852938 on Build GRCh38
rs58510892 Chr.12: 9853082-9853083 on Build GRCh38
rs996366908 Chr.12: 9852891-9852891 on Build GRCh38
rs757353379 Chr.12: 9853319-9853319 on Build GRCh38
rs61914641 Chr.12: 9853101-9853101 on Build GRCh38
rs550042484 Chr.12: 9853341-9853341 on Build GRCh38
rs764137578 Chr.12: 9852922-9852922 on Build GRCh38
rs746081758 Chr.12: 9853335-9853335 on Build GRCh38
rs961127578 Chr.12: 9853166-9853166 on Build GRCh38
rs201360652 Chr.12: 9853254-9853254 on Build GRCh38
rs61914642 Chr.12: 9853105-9853105 on Build GRCh38
rs142115371 Chr.12: 9853336-9853336 on Build GRCh38
rs1050142143 Chr.12: 9853097-9853097 on Build GRCh38
rs760692325 Chr.12: 9853256-9853258 on Build GRCh38
rs2539933 Chr.12: 9853181-9853181 on Build GRCh38
rs777722637 Chr.12: 9853279-9853279 on Build GRCh38
rs554408221 Chr.12: 9852900-9852900 on Build GRCh38
rs569777311 Chr.12: 9852982-9852982 on Build GRCh38
rs253157 Chr.12: 9853083-9853083 on Build GRCh38
rs909369772 Chr.12: 9853108-9853108 on Build GRCh38
rs111861666 Chr.12: 9853180-9853180 on Build GRCh38
rs77439123 Chr.12: 9853059-9853060 on Build GRCh38
rs199731258 Chr.12: 9853087-9853087 on Build GRCh38
rs66810628 Chr.12: 9853088-9853089 on Build GRCh38
rs756747529 Chr.12: 9852989-9852989 on Build GRCh38
rs879071173 Chr.12: 9853227-9853227 on Build GRCh38
rs759279292 Chr.12: 9853085-9853086 on Build GRCh38
rs866303908 Chr.12: 9853085-9853085 on Build GRCh38
rs967388091 Chr.12: 9852999-9852999 on Build GRCh38
rs975189220 Chr.12: 9853117-9853117 on Build GRCh38
rs752319934 Chr.12: 9853389-9853389 on Build GRCh38
rs554204323 Chr.12: 9853123-9853123 on Build GRCh38
rs201343247 Chr.12: 9853091-9853091 on Build GRCh38
rs780456569 Chr.12: 9853213-9853213 on Build GRCh38
rs530186204 Chr.12: 9853337-9853337 on Build GRCh38
rs762756826 Chr.12: 9853395-9853395 on Build GRCh38
rs763326082 Chr.12: 9853086-9853087 on Build GRCh38
rs866117781 Chr.12: 9853138-9853138 on Build GRCh38
rs386760215 Chr.12: 9853179-9853181 on Build GRCh38

View Full Product Details