Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 64714142-64714739 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNS
Entrez Gene ID
2799
Gene Name
glucosamine (N-acetyl)-6-sulfatase
Gene Aliases
G6S
UniGene
Hs.334534
Gene Chromosome Location
Chr.12: 64713442-64759446 on Build GRCh38
Transcripts
NM_002076.3

SNP Detail

SNP ID Chromosome Location
rs886049762 Chr.12: 64714181-64714181 on Build GRCh38
rs529418480 Chr.12: 64714252-64714252 on Build GRCh38
rs553360517 Chr.12: 64714642-64714642 on Build GRCh38
rs897879142 Chr.12: 64714475-64714475 on Build GRCh38
rs1037319060 Chr.12: 64714334-64714334 on Build GRCh38
rs917772899 Chr.12: 64714576-64714576 on Build GRCh38
rs1028250406 Chr.12: 64714199-64714199 on Build GRCh38
rs978924418 Chr.12: 64714668-64714668 on Build GRCh38
rs150772287 Chr.12: 64714599-64714599 on Build GRCh38
rs967604236 Chr.12: 64714350-64714350 on Build GRCh38
rs980567416 Chr.12: 64714628-64714628 on Build GRCh38
rs995044057 Chr.12: 64714170-64714170 on Build GRCh38
rs967132222 Chr.12: 64714655-64714655 on Build GRCh38
rs891259951 Chr.12: 64714420-64714420 on Build GRCh38
rs549541581 Chr.12: 64714337-64714337 on Build GRCh38
rs1061743 Chr.12: 64714380-64714380 on Build GRCh38
rs566964017 Chr.12: 64714694-64714694 on Build GRCh38
rs551638399 Chr.12: 64714552-64714552 on Build GRCh38
rs1008387023 Chr.12: 64714425-64714425 on Build GRCh38
rs745566534 Chr.12: 64714441-64714441 on Build GRCh38
rs571468415 Chr.12: 64714569-64714569 on Build GRCh38
rs570203117 Chr.12: 64714598-64714598 on Build GRCh38
rs941748050 Chr.12: 64714643-64714643 on Build GRCh38
rs918889489 Chr.12: 64714709-64714709 on Build GRCh38
rs1049191307 Chr.12: 64714518-64714518 on Build GRCh38
rs533516870 Chr.12: 64714605-64714605 on Build GRCh38
rs1047243472 Chr.12: 64714412-64714412 on Build GRCh38
rs778825291 Chr.12: 64714445-64714445 on Build GRCh38
rs976908243 Chr.12: 64714287-64714287 on Build GRCh38
rs1017920778 Chr.12: 64714234-64714234 on Build GRCh38
rs988442450 Chr.12: 64714568-64714568 on Build GRCh38
rs1036841344 Chr.12: 64714463-64714463 on Build GRCh38
rs978591209 Chr.12: 64714356-64714356 on Build GRCh38
rs898753822 Chr.12: 64714345-64714345 on Build GRCh38
rs550181590 Chr.12: 64714409-64714410 on Build GRCh38
rs900823217 Chr.12: 64714595-64714595 on Build GRCh38
rs950078336 Chr.12: 64714459-64714459 on Build GRCh38
rs1007819273 Chr.12: 64714228-64714228 on Build GRCh38
rs141182054 Chr.12: 64714440-64714440 on Build GRCh38
rs369653063 Chr.12: 64714566-64714566 on Build GRCh38
rs1000903432 Chr.12: 64714489-64714489 on Build GRCh38
rs1044766506 Chr.12: 64714230-64714230 on Build GRCh38
rs148217767 Chr.12: 64714203-64714203 on Build GRCh38
rs889348909 Chr.12: 64714217-64714217 on Build GRCh38
rs941737503 Chr.12: 64714319-64714319 on Build GRCh38
rs970566738 Chr.12: 64714617-64714617 on Build GRCh38
rs567930294 Chr.12: 64714294-64714294 on Build GRCh38
rs965076343 Chr.12: 64714244-64714244 on Build GRCh38
rs886049763 Chr.12: 64714709-64714709 on Build GRCh38
rs904941118 Chr.12: 64714293-64714293 on Build GRCh38
rs930740068 Chr.12: 64714492-64714492 on Build GRCh38

View Full Product Details