Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 109930473-109931044 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GIT2
Entrez Gene ID
9815
Gene Name
GIT ArfGAP 2
Gene Aliases
CAT-2, CAT2, PKL
UniGene
Hs.434996
Gene Chromosome Location
Chr.12: 109929792-109996389 on Build GRCh38
Transcripts
NM_001135213.1 NM_001135214.1 NM_014776.3 NM_057169.3 NM_057170.3 XM_005253997.3 XM_005254000.3 XM_006719707.3 XM_006719708.3 XM_006719709.3 XM_006719712.3 XM_017020258.1 XM_017020259.1

SNP Detail

SNP ID Chromosome Location
rs534722290 Chr.12: 109930594-109930594 on Build GRCh38
rs767663169 Chr.12: 109930899-109930899 on Build GRCh38
rs372828602 Chr.12: 109930942-109930942 on Build GRCh38
rs558406727 Chr.12: 109930957-109930957 on Build GRCh38
rs914685770 Chr.12: 109930925-109930925 on Build GRCh38
rs1023413117 Chr.12: 109930706-109930706 on Build GRCh38
rs963418997 Chr.12: 109930675-109930675 on Build GRCh38
rs904252594 Chr.12: 109930538-109930538 on Build GRCh38
rs574863733 Chr.12: 109930633-109930633 on Build GRCh38
rs757231836 Chr.12: 109930672-109930672 on Build GRCh38
rs917127591 Chr.12: 109930778-109930778 on Build GRCh38
rs369547471 Chr.12: 109930810-109930810 on Build GRCh38
rs947485130 Chr.12: 109930944-109930944 on Build GRCh38
rs747523716 Chr.12: 109930900-109930900 on Build GRCh38
rs16940693 Chr.12: 109930592-109930592 on Build GRCh38
rs893218238 Chr.12: 109930556-109930556 on Build GRCh38
rs764441189 Chr.12: 109930551-109930552 on Build GRCh38
rs927148340 Chr.12: 109930740-109930740 on Build GRCh38
rs77766064 Chr.12: 109930999-109930999 on Build GRCh38
rs748930566 Chr.12: 109930733-109930733 on Build GRCh38
rs1053948037 Chr.12: 109930540-109930540 on Build GRCh38
rs1036311733 Chr.12: 109930632-109930632 on Build GRCh38
rs994886746 Chr.12: 109931011-109931011 on Build GRCh38
rs768073095 Chr.12: 109930764-109930764 on Build GRCh38
rs771563091 Chr.12: 109930940-109930940 on Build GRCh38
rs779541252 Chr.12: 109930657-109930657 on Build GRCh38
rs958747814 Chr.12: 109930758-109930758 on Build GRCh38
rs553316643 Chr.12: 109930595-109930595 on Build GRCh38
rs935731906 Chr.12: 109930839-109930839 on Build GRCh38
rs745648475 Chr.12: 109930606-109930606 on Build GRCh38
rs1025333642 Chr.12: 109930790-109930790 on Build GRCh38
rs1039821898 Chr.12: 109930872-109930872 on Build GRCh38
rs1001060442 Chr.12: 109930539-109930539 on Build GRCh38
rs773941968 Chr.12: 109930766-109930766 on Build GRCh38
rs900788154 Chr.12: 109930547-109930547 on Build GRCh38
rs1013081334 Chr.12: 109930666-109930666 on Build GRCh38
rs554380501 Chr.12: 109930801-109930801 on Build GRCh38
rs543164332 Chr.12: 109930941-109930941 on Build GRCh38
rs188536339 Chr.12: 109930913-109930913 on Build GRCh38
rs932112773 Chr.12: 109930903-109930903 on Build GRCh38
rs755839181 Chr.12: 109930622-109930622 on Build GRCh38
rs867329652 Chr.12: 109930834-109930834 on Build GRCh38
rs890685340 Chr.12: 109930963-109930963 on Build GRCh38
rs947411815 Chr.12: 109930577-109930577 on Build GRCh38
rs938647986 Chr.12: 109931012-109931012 on Build GRCh38
rs541914569 Chr.12: 109930674-109930674 on Build GRCh38
rs756379400 Chr.12: 109931009-109931009 on Build GRCh38
rs899296411 Chr.12: 109931010-109931010 on Build GRCh38
rs987100982 Chr.12: 109930511-109930511 on Build GRCh38
rs763373372 Chr.12: 109930521-109930521 on Build GRCh38
rs973244558 Chr.12: 109930705-109930705 on Build GRCh38
rs977070804 Chr.12: 109930809-109930809 on Build GRCh38
rs996444328 Chr.12: 109930560-109930560 on Build GRCh38
rs374095974 Chr.12: 109930696-109930697 on Build GRCh38

View Full Product Details