Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 53215849-53216302 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf123
Entrez Gene ID
54987
Gene Name
chromosome 1 open reading frame 123
Gene Aliases
-
UniGene
Hs.525391
Gene Chromosome Location
Chr.1: 53214099-53220639 on Build GRCh38
Transcripts
NM_001304759.1 NM_001304760.1 NM_017887.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5475814 c.349G>A p.A117T Chr.1: 53216047-53216047 on Build GRCh38
3751295 c.369A>C p.S123S Chr.1: 53216027-53216027 on Build GRCh38
2193300 c.397C>A p.Q133K Chr.1: 53215999-53215999 on Build GRCh38
910776 c.371G>T p.G124V Chr.1: 53216025-53216025 on Build GRCh38
4878198 c.370G>A p.G124R Chr.1: 53216026-53216026 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756774484 Chr.1: 53216045-53216045 on Build GRCh38
rs375042095 Chr.1: 53216098-53216098 on Build GRCh38
rs148032803 Chr.1: 53216062-53216062 on Build GRCh38
rs755312565 Chr.1: 53215994-53215995 on Build GRCh38
rs74880381 Chr.1: 53216120-53216120 on Build GRCh38
rs113278028 Chr.1: 53216053-53216053 on Build GRCh38
rs764701489 Chr.1: 53215899-53215899 on Build GRCh38
rs769681882 Chr.1: 53216051-53216051 on Build GRCh38
rs897849613 Chr.1: 53216137-53216137 on Build GRCh38
rs766833705 Chr.1: 53216096-53216096 on Build GRCh38
rs899860784 Chr.1: 53215923-53215923 on Build GRCh38
rs965699932 Chr.1: 53215992-53215992 on Build GRCh38
rs772274279 Chr.1: 53216074-53216074 on Build GRCh38
rs1051051311 Chr.1: 53216161-53216161 on Build GRCh38
rs768812658 Chr.1: 53215948-53215948 on Build GRCh38
rs775218797 Chr.1: 53215944-53215944 on Build GRCh38
rs372043684 Chr.1: 53216124-53216124 on Build GRCh38
rs763225804 Chr.1: 53215946-53215946 on Build GRCh38
rs777435864 Chr.1: 53216013-53216013 on Build GRCh38
rs926196897 Chr.1: 53216268-53216268 on Build GRCh38
rs1046935326 Chr.1: 53215889-53215889 on Build GRCh38
rs368410313 Chr.1: 53216071-53216071 on Build GRCh38
rs750433701 Chr.1: 53215965-53215965 on Build GRCh38
rs1012175137 Chr.1: 53216206-53216206 on Build GRCh38
rs374003750 Chr.1: 53216063-53216063 on Build GRCh38
rs558756386 Chr.1: 53216146-53216146 on Build GRCh38
rs762262731 Chr.1: 53215918-53215918 on Build GRCh38
rs939144980 Chr.1: 53215879-53215879 on Build GRCh38
rs185450345 Chr.1: 53216178-53216178 on Build GRCh38
rs774701797 Chr.1: 53215912-53215912 on Build GRCh38
rs1038757870 Chr.1: 53215921-53215921 on Build GRCh38
rs958827742 Chr.1: 53216281-53216281 on Build GRCh38
rs180739194 Chr.1: 53216114-53216114 on Build GRCh38
rs374880744 Chr.1: 53216178-53216178 on Build GRCh38
rs115884122 Chr.1: 53216225-53216225 on Build GRCh38
rs957591641 Chr.1: 53216195-53216195 on Build GRCh38
rs542668521 Chr.1: 53215905-53215905 on Build GRCh38
rs2297664 Chr.1: 53216184-53216184 on Build GRCh38
rs36101657 Chr.1: 53216272-53216273 on Build GRCh38
rs189937356 Chr.1: 53215959-53215959 on Build GRCh38
rs950197012 Chr.1: 53216115-53216115 on Build GRCh38
rs370689910 Chr.1: 53216084-53216084 on Build GRCh38
rs201506667 Chr.1: 53215942-53215942 on Build GRCh38
rs927649983 Chr.1: 53216231-53216231 on Build GRCh38
rs113619772 Chr.1: 53216147-53216147 on Build GRCh38
rs1031779971 Chr.1: 53216186-53216186 on Build GRCh38
rs998933416 Chr.1: 53216177-53216177 on Build GRCh38
rs376884421 Chr.1: 53216060-53216060 on Build GRCh38
rs189390940 Chr.1: 53216217-53216217 on Build GRCh38
rs758369506 Chr.1: 53216152-53216152 on Build GRCh38
rs116341139 Chr.1: 53216170-53216170 on Build GRCh38
rs754201552 Chr.1: 53215974-53215974 on Build GRCh38
rs747237230 Chr.1: 53215962-53215963 on Build GRCh38
rs17108139 Chr.1: 53216110-53216110 on Build GRCh38
rs914880655 Chr.1: 53216059-53216059 on Build GRCh38
rs774530368 Chr.1: 53215955-53215955 on Build GRCh38
rs762292303 Chr.1: 53216102-53216102 on Build GRCh38
rs768075807 Chr.1: 53215958-53215958 on Build GRCh38
rs372620461 Chr.1: 53215966-53215966 on Build GRCh38
rs372135240 Chr.1: 53216103-53216103 on Build GRCh38
rs906296222 Chr.1: 53215873-53215873 on Build GRCh38
rs374927409 Chr.1: 53216095-53216095 on Build GRCh38
rs758039598 Chr.1: 53215998-53215998 on Build GRCh38
rs960443540 Chr.1: 53216248-53216248 on Build GRCh38
rs377670747 Chr.1: 53216218-53216218 on Build GRCh38
rs966177453 Chr.1: 53216185-53216185 on Build GRCh38
rs762004097 Chr.1: 53215956-53215956 on Build GRCh38
rs559185932 Chr.1: 53215917-53215917 on Build GRCh38
rs781045218 Chr.1: 53216046-53216046 on Build GRCh38
rs1134688 Chr.1: 53216027-53216027 on Build GRCh38

View Full Product Details