Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 74197694-74198224 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FPGT
Entrez Gene ID
8790
Gene Name
fucose-1-phosphate guanylyltransferase
Gene Aliases
GFPP
UniGene
Hs.480085
Gene Chromosome Location
Chr.1: 74198212-74208702 on Build GRCh38
Transcripts
NM_001199328.2 NM_001199329.2 NM_003838.4 NM_001112808.2 NM_001199327.1 NM_001105659.1 NM_001322315.1 XM_011540658.1 XM_011540663.2 XM_017000286.1 XM_017000287.1 XM_017000288.1

Gene Symbol
FPGT-TNNI3K
Entrez Gene ID
100526835
Gene Name
FPGT-TNNI3K readthrough
Gene Aliases
-
UniGene
Hs.480085
Gene Chromosome Location
Chr.1: 74198212-74544432 on Build GRCh38
Transcripts
NM_001199328.2 NM_001199329.2 NM_003838.4 NM_001112808.2 NM_001199327.1 NM_001105659.1 NM_001322315.1 XM_011540658.1 XM_011540663.2 XM_017000286.1 XM_017000287.1 XM_017000288.1

Gene Symbol
LRRIQ3
Entrez Gene ID
127255
Gene Name
leucine rich repeats and IQ motif containing 3
Gene Aliases
LRRC44
UniGene
Hs.644625
Gene Chromosome Location
Chr.1: 74026015-74198201 on Build GRCh38
Transcripts
NM_001199328.2 NM_001199329.2 NM_003838.4 NM_001112808.2 NM_001199327.1 NM_001105659.1 NM_001322315.1 XM_011540658.1 XM_011540663.2 XM_017000286.1 XM_017000287.1 XM_017000288.1

SNP Detail

SNP ID Chromosome Location
rs947473135 Chr.1: 74197790-74197790 on Build GRCh38
rs569881223 Chr.1: 74198186-74198186 on Build GRCh38
rs913257330 Chr.1: 74198095-74198095 on Build GRCh38
rs749897104 Chr.1: 74197823-74197823 on Build GRCh38
rs776193087 Chr.1: 74198204-74198204 on Build GRCh38
rs778748702 Chr.1: 74198109-74198109 on Build GRCh38
rs886151973 Chr.1: 74198170-74198170 on Build GRCh38
rs574310481 Chr.1: 74198009-74198009 on Build GRCh38
rs770166566 Chr.1: 74198203-74198203 on Build GRCh38
rs1029576422 Chr.1: 74197736-74197736 on Build GRCh38
rs139721001 Chr.1: 74198117-74198117 on Build GRCh38
rs34913402 Chr.1: 74197982-74197983 on Build GRCh38
rs927412510 Chr.1: 74197795-74197795 on Build GRCh38
rs184792890 Chr.1: 74197939-74197939 on Build GRCh38
rs1042320294 Chr.1: 74197841-74197841 on Build GRCh38
rs1054930309 Chr.1: 74197898-74197898 on Build GRCh38
rs551677653 Chr.1: 74198179-74198179 on Build GRCh38
rs974844002 Chr.1: 74197744-74197744 on Build GRCh38
rs746461735 Chr.1: 74198194-74198194 on Build GRCh38
rs750510096 Chr.1: 74197938-74197938 on Build GRCh38
rs1005249757 Chr.1: 74197853-74197853 on Build GRCh38
rs1020258214 Chr.1: 74197975-74197975 on Build GRCh38
rs77030123 Chr.1: 74198167-74198167 on Build GRCh38
rs978830597 Chr.1: 74197793-74197793 on Build GRCh38
rs1029915702 Chr.1: 74197934-74197935 on Build GRCh38
rs763303473 Chr.1: 74197808-74197808 on Build GRCh38
rs565972704 Chr.1: 74198151-74198151 on Build GRCh38
rs570661850 Chr.1: 74197953-74197953 on Build GRCh38
rs886100533 Chr.1: 74197962-74197962 on Build GRCh38
rs1040949209 Chr.1: 74198062-74198069 on Build GRCh38
rs776299899 Chr.1: 74198197-74198197 on Build GRCh38
rs764385265 Chr.1: 74197908-74197908 on Build GRCh38
rs939167921 Chr.1: 74197882-74197882 on Build GRCh38
rs966181907 Chr.1: 74197731-74197731 on Build GRCh38
rs757393061 Chr.1: 74197946-74197946 on Build GRCh38
rs917408382 Chr.1: 74198053-74198053 on Build GRCh38
rs936072591 Chr.1: 74197801-74197801 on Build GRCh38
rs906401986 Chr.1: 74197842-74197842 on Build GRCh38
rs185634617 Chr.1: 74198147-74198147 on Build GRCh38
rs1027764041 Chr.1: 74197952-74197952 on Build GRCh38
rs1039988060 Chr.1: 74198184-74198184 on Build GRCh38
rs990575840 Chr.1: 74197803-74197803 on Build GRCh38
rs189356677 Chr.1: 74198017-74198017 on Build GRCh38
rs11806946 Chr.1: 74198025-74198025 on Build GRCh38
rs967402346 Chr.1: 74198106-74198106 on Build GRCh38
rs913212103 Chr.1: 74197807-74197807 on Build GRCh38
rs937461265 Chr.1: 74198050-74198050 on Build GRCh38
rs552365796 Chr.1: 74197864-74197864 on Build GRCh38
rs913229425 Chr.1: 74197777-74197777 on Build GRCh38
rs770505202 Chr.1: 74198196-74198196 on Build GRCh38
rs368869827 Chr.1: 74198159-74198159 on Build GRCh38
rs978757110 Chr.1: 74198019-74198023 on Build GRCh38
rs1004513676 Chr.1: 74198182-74198182 on Build GRCh38
rs998009242 Chr.1: 74197923-74197923 on Build GRCh38
rs34030954 Chr.1: 74197804-74197804 on Build GRCh38
rs541806703 Chr.1: 74198015-74198015 on Build GRCh38
rs550261166 Chr.1: 74198085-74198085 on Build GRCh38
rs191636757 Chr.1: 74197812-74197812 on Build GRCh38
rs546653717 Chr.1: 74198122-74198122 on Build GRCh38
rs990236558 Chr.1: 74198088-74198088 on Build GRCh38
rs922027745 Chr.1: 74197758-74197758 on Build GRCh38
rs180933483 Chr.1: 74198052-74198052 on Build GRCh38
rs1057271728 Chr.1: 74197815-74197815 on Build GRCh38
rs930424424 Chr.1: 74198160-74198160 on Build GRCh38
rs1050926512 Chr.1: 74198126-74198126 on Build GRCh38
rs888123043 Chr.1: 74197851-74197851 on Build GRCh38
rs1412825 Chr.1: 74198130-74198130 on Build GRCh38
rs531554403 Chr.1: 74198074-74198074 on Build GRCh38
rs565506746 Chr.1: 74197829-74197829 on Build GRCh38
rs768045749 Chr.1: 74197768-74197768 on Build GRCh38
rs896062499 Chr.1: 74197838-74197838 on Build GRCh38
rs576256259 Chr.1: 74197847-74197847 on Build GRCh38
rs996315283 Chr.1: 74198034-74198034 on Build GRCh38
rs1047748710 Chr.1: 74198079-74198079 on Build GRCh38
rs546147721 Chr.1: 74198047-74198047 on Build GRCh38
rs555856424 Chr.1: 74198008-74198008 on Build GRCh38

View Full Product Details