Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 111198355-111198953 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DENND2D
Entrez Gene ID
79961
Gene Name
DENN domain containing 2D
Gene Aliases
-
UniGene
Hs.557850
Gene Chromosome Location
Chr.1: 111185969-111204791 on Build GRCh38
Transcripts
NM_001271833.1 NM_024901.4 XM_006710921.2 XM_006710922.1 XM_006710923.1 XM_006710924.1 XM_011542187.1 XM_011542189.1 XM_011542190.2 XM_017002386.1 XM_017002387.1 XM_017002388.1 XM_017002389.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3740534 c.290T>C p.L97P Chr.1: 111198696-111198696 on Build GRCh38
5603360 c.275A>T p.E92V Chr.1: 111198711-111198711 on Build GRCh38
4396678 c.322G>A p.G108R Chr.1: 111198664-111198664 on Build GRCh38
4217762 c.244C>T p.R82W Chr.1: 111198742-111198742 on Build GRCh38
3688874 c.286C>T p.L96L Chr.1: 111198700-111198700 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1042349869 Chr.1: 111198461-111198461 on Build GRCh38
rs932214764 Chr.1: 111198921-111198921 on Build GRCh38
rs980904494 Chr.1: 111198899-111198899 on Build GRCh38
rs200450897 Chr.1: 111198745-111198745 on Build GRCh38
rs776127765 Chr.1: 111198649-111198649 on Build GRCh38
rs767828445 Chr.1: 111198668-111198668 on Build GRCh38
rs77930554 Chr.1: 111198513-111198513 on Build GRCh38
rs1009143802 Chr.1: 111198497-111198497 on Build GRCh38
rs143149220 Chr.1: 111198871-111198871 on Build GRCh38
rs776185934 Chr.1: 111198876-111198876 on Build GRCh38
rs996713496 Chr.1: 111198518-111198518 on Build GRCh38
rs35184750 Chr.1: 111198700-111198700 on Build GRCh38
rs865867875 Chr.1: 111198915-111198915 on Build GRCh38
rs746755770 Chr.1: 111198704-111198704 on Build GRCh38
rs202107158 Chr.1: 111198589-111198589 on Build GRCh38
rs769477901 Chr.1: 111198917-111198917 on Build GRCh38
rs368169530 Chr.1: 111198582-111198582 on Build GRCh38
rs765483359 Chr.1: 111198787-111198787 on Build GRCh38
rs139774926 Chr.1: 111198577-111198577 on Build GRCh38
rs893861535 Chr.1: 111198413-111198413 on Build GRCh38
rs990052177 Chr.1: 111198819-111198819 on Build GRCh38
rs369860619 Chr.1: 111198763-111198763 on Build GRCh38
rs540223648 Chr.1: 111198379-111198379 on Build GRCh38
rs914427686 Chr.1: 111198650-111198650 on Build GRCh38
rs755456471 Chr.1: 111198492-111198492 on Build GRCh38
rs976724509 Chr.1: 111198384-111198384 on Build GRCh38
rs904351088 Chr.1: 111198802-111198802 on Build GRCh38
rs141276517 Chr.1: 111198727-111198727 on Build GRCh38
rs573384912 Chr.1: 111198736-111198736 on Build GRCh38
rs1004634527 Chr.1: 111198715-111198715 on Build GRCh38
rs983437469 Chr.1: 111198881-111198881 on Build GRCh38
rs922647193 Chr.1: 111198651-111198651 on Build GRCh38
rs774968116 Chr.1: 111198730-111198730 on Build GRCh38
rs753820798 Chr.1: 111198769-111198770 on Build GRCh38
rs771815611 Chr.1: 111198614-111198614 on Build GRCh38
rs1014591160 Chr.1: 111198556-111198556 on Build GRCh38
rs769022033 Chr.1: 111198593-111198593 on Build GRCh38
rs764899669 Chr.1: 111198653-111198653 on Build GRCh38
rs3748713 Chr.1: 111198847-111198847 on Build GRCh38
rs756629194 Chr.1: 111198580-111198580 on Build GRCh38
rs191564309 Chr.1: 111198873-111198873 on Build GRCh38
rs762569338 Chr.1: 111198666-111198666 on Build GRCh38
rs560544637 Chr.1: 111198385-111198385 on Build GRCh38
rs760593790 Chr.1: 111198639-111198639 on Build GRCh38
rs368846232 Chr.1: 111198686-111198686 on Build GRCh38
rs777784507 Chr.1: 111198591-111198591 on Build GRCh38
rs375758587 Chr.1: 111198722-111198722 on Build GRCh38
rs779238633 Chr.1: 111198594-111198594 on Build GRCh38
rs922811862 Chr.1: 111198403-111198403 on Build GRCh38
rs375867823 Chr.1: 111198652-111198652 on Build GRCh38
rs774200256 Chr.1: 111198897-111198898 on Build GRCh38
rs780575756 Chr.1: 111198538-111198538 on Build GRCh38
rs1047653759 Chr.1: 111198434-111198434 on Build GRCh38
rs1034036521 Chr.1: 111198801-111198801 on Build GRCh38
rs772337248 Chr.1: 111198702-111198702 on Build GRCh38
rs748122243 Chr.1: 111198643-111198643 on Build GRCh38
rs35510030 Chr.1: 111198668-111198669 on Build GRCh38
rs773049072 Chr.1: 111198622-111198622 on Build GRCh38
rs999952937 Chr.1: 111198826-111198826 on Build GRCh38
rs983086971 Chr.1: 111198658-111198658 on Build GRCh38
rs964106545 Chr.1: 111198615-111198615 on Build GRCh38
rs371592275 Chr.1: 111198585-111198585 on Build GRCh38
rs111385340 Chr.1: 111198906-111198906 on Build GRCh38
rs548238851 Chr.1: 111198641-111198641 on Build GRCh38
rs1018076424 Chr.1: 111198842-111198842 on Build GRCh38
rs981135696 Chr.1: 111198570-111198570 on Build GRCh38
rs756594745 Chr.1: 111198685-111198685 on Build GRCh38
rs534943659 Chr.1: 111198864-111198864 on Build GRCh38
rs755153104 Chr.1: 111198695-111198695 on Build GRCh38
rs1030066454 Chr.1: 111198857-111198857 on Build GRCh38
rs748353437 Chr.1: 111198596-111198596 on Build GRCh38
rs910077098 Chr.1: 111198437-111198437 on Build GRCh38
rs761062864 Chr.1: 111198750-111198750 on Build GRCh38
rs752818121 Chr.1: 111198524-111198524 on Build GRCh38
rs944820780 Chr.1: 111198887-111198887 on Build GRCh38
rs41312686 Chr.1: 111198566-111198566 on Build GRCh38
rs369436297 Chr.1: 111198793-111198793 on Build GRCh38
rs998251193 Chr.1: 111198848-111198848 on Build GRCh38
rs951757894 Chr.1: 111198537-111198537 on Build GRCh38
rs562996187 Chr.1: 111198525-111198525 on Build GRCh38
rs967223368 Chr.1: 111198854-111198854 on Build GRCh38
rs555155245 Chr.1: 111198824-111198824 on Build GRCh38
rs902519104 Chr.1: 111198674-111198674 on Build GRCh38
rs745712904 Chr.1: 111198603-111198603 on Build GRCh38
rs1019653357 Chr.1: 111198502-111198502 on Build GRCh38
rs752386343 Chr.1: 111198664-111198664 on Build GRCh38
rs755530521 Chr.1: 111198765-111198765 on Build GRCh38
rs946819073 Chr.1: 111198453-111198453 on Build GRCh38
rs916891816 Chr.1: 111198924-111198924 on Build GRCh38
rs937131601 Chr.1: 111198377-111198377 on Build GRCh38
rs549444383 Chr.1: 111198703-111198703 on Build GRCh38
rs750775156 Chr.1: 111198748-111198748 on Build GRCh38
rs958852003 Chr.1: 111198803-111198803 on Build GRCh38
rs577241346 Chr.1: 111198411-111198411 on Build GRCh38
rs759437352 Chr.1: 111198726-111198726 on Build GRCh38
rs750733830 Chr.1: 111198683-111198683 on Build GRCh38
rs762577834 Chr.1: 111198742-111198742 on Build GRCh38
rs923986024 Chr.1: 111198880-111198880 on Build GRCh38
rs373984735 Chr.1: 111198692-111198692 on Build GRCh38
rs77286992 Chr.1: 111198913-111198913 on Build GRCh38
rs770793520 Chr.1: 111198710-111198710 on Build GRCh38
rs1010171692 Chr.1: 111198553-111198553 on Build GRCh38
rs201350699 Chr.1: 111198701-111198701 on Build GRCh38
rs535262267 Chr.1: 111198791-111198791 on Build GRCh38

View Full Product Details