Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 111349802-111350338 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PIFO
Entrez Gene ID
128344
Gene Name
primary cilia formation
Gene Aliases
C1orf88, pitchfork
UniGene
Hs.172510
Gene Chromosome Location
Chr.1: 111324663-111353017 on Build GRCh38
Transcripts
NM_001300831.1 NM_181643.5 NR_125338.1 XM_005270472.1 XM_017000322.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6120574 c.472G>C p.V158L Chr.1: 111350188-111350188 on Build GRCh38
212185 c.422C>T p.P141L Chr.1: 111350138-111350138 on Build GRCh38
674352 c.473T>A p.V158D Chr.1: 111350189-111350189 on Build GRCh38
5843057 c.451G>T p.G151* Chr.1: 111350167-111350167 on Build GRCh38
7161668 c.505G>A p.E169K Chr.1: 111350221-111350221 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs543781679 Chr.1: 111350018-111350018 on Build GRCh38
rs762666026 Chr.1: 111350169-111350169 on Build GRCh38
rs1034545298 Chr.1: 111350236-111350236 on Build GRCh38
rs575564423 Chr.1: 111350000-111350000 on Build GRCh38
rs914310543 Chr.1: 111350295-111350295 on Build GRCh38
rs773468106 Chr.1: 111350195-111350195 on Build GRCh38
rs532303863 Chr.1: 111350150-111350150 on Build GRCh38
rs765276862 Chr.1: 111350212-111350212 on Build GRCh38
rs368637416 Chr.1: 111350105-111350105 on Build GRCh38
rs947366626 Chr.1: 111350177-111350177 on Build GRCh38
rs202227757 Chr.1: 111350067-111350067 on Build GRCh38
rs764990191 Chr.1: 111350087-111350087 on Build GRCh38
rs752954545 Chr.1: 111350234-111350234 on Build GRCh38
rs771902595 Chr.1: 111350136-111350136 on Build GRCh38
rs553199572 Chr.1: 111349911-111349911 on Build GRCh38
rs374843160 Chr.1: 111350056-111350056 on Build GRCh38
rs376791033 Chr.1: 111350134-111350134 on Build GRCh38
rs747941434 Chr.1: 111350055-111350055 on Build GRCh38
rs979073067 Chr.1: 111350191-111350191 on Build GRCh38
rs981642848 Chr.1: 111349997-111349997 on Build GRCh38
rs551023453 Chr.1: 111349838-111349838 on Build GRCh38
rs761038570 Chr.1: 111350200-111350200 on Build GRCh38
rs538883143 Chr.1: 111349975-111349975 on Build GRCh38
rs937132131 Chr.1: 111350019-111350019 on Build GRCh38
rs371024578 Chr.1: 111350138-111350138 on Build GRCh38
rs562750946 Chr.1: 111350201-111350201 on Build GRCh38
rs760505900 Chr.1: 111350074-111350074 on Build GRCh38
rs539461537 Chr.1: 111349932-111349932 on Build GRCh38
rs763759949 Chr.1: 111350190-111350190 on Build GRCh38
rs913155558 Chr.1: 111349902-111349902 on Build GRCh38
rs567760629 Chr.1: 111349866-111349866 on Build GRCh38
rs758436214 Chr.1: 111349827-111349827 on Build GRCh38
rs372205144 Chr.1: 111349988-111349988 on Build GRCh38
rs752397072 Chr.1: 111350100-111350100 on Build GRCh38
rs1008022998 Chr.1: 111349861-111349861 on Build GRCh38
rs1015185550 Chr.1: 111349987-111349987 on Build GRCh38
rs983446685 Chr.1: 111349980-111349980 on Build GRCh38
rs548343385 Chr.1: 111350139-111350139 on Build GRCh38
rs372640374 Chr.1: 111350125-111350125 on Build GRCh38
rs149188444 Chr.1: 111350209-111350209 on Build GRCh38
rs942545875 Chr.1: 111350214-111350214 on Build GRCh38
rs780558079 Chr.1: 111350128-111350128 on Build GRCh38
rs891075326 Chr.1: 111350078-111350078 on Build GRCh38
rs757356684 Chr.1: 111350232-111350232 on Build GRCh38
rs868827826 Chr.1: 111350135-111350135 on Build GRCh38
rs201565676 Chr.1: 111350153-111350153 on Build GRCh38
rs943955103 Chr.1: 111350127-111350127 on Build GRCh38
rs567007166 Chr.1: 111349925-111349925 on Build GRCh38
rs756607202 Chr.1: 111350113-111350113 on Build GRCh38
rs765392772 Chr.1: 111350203-111350203 on Build GRCh38
rs901241454 Chr.1: 111350025-111350025 on Build GRCh38
rs56255516 Chr.1: 111350287-111350287 on Build GRCh38
rs777995298 Chr.1: 111350235-111350235 on Build GRCh38
rs192787128 Chr.1: 111350033-111350033 on Build GRCh38
rs900634698 Chr.1: 111349919-111349919 on Build GRCh38
rs373100710 Chr.1: 111350261-111350261 on Build GRCh38
rs759058970 Chr.1: 111350083-111350083 on Build GRCh38
rs750976573 Chr.1: 111350106-111350106 on Build GRCh38
rs927444939 Chr.1: 111350006-111350006 on Build GRCh38
rs776028575 Chr.1: 111350081-111350081 on Build GRCh38
rs942826577 Chr.1: 111349992-111349992 on Build GRCh38
rs145133389 Chr.1: 111350303-111350303 on Build GRCh38
rs184877714 Chr.1: 111350034-111350034 on Build GRCh38
rs771849518 Chr.1: 111350059-111350059 on Build GRCh38
rs770863484 Chr.1: 111350076-111350076 on Build GRCh38
rs1018054849 Chr.1: 111349894-111349894 on Build GRCh38
rs868212483 Chr.1: 111349956-111349956 on Build GRCh38
rs1041538344 Chr.1: 111350001-111350001 on Build GRCh38
rs759187687 Chr.1: 111350156-111350156 on Build GRCh38
rs372973539 Chr.1: 111350315-111350315 on Build GRCh38
rs775334326 Chr.1: 111350168-111350168 on Build GRCh38
rs754339072 Chr.1: 111350207-111350207 on Build GRCh38
rs988454543 Chr.1: 111349881-111349881 on Build GRCh38
rs3820443 Chr.1: 111350283-111350283 on Build GRCh38
rs982275057 Chr.1: 111350111-111350111 on Build GRCh38
rs762192157 Chr.1: 111350126-111350128 on Build GRCh38
rs753873152 Chr.1: 111350130-111350130 on Build GRCh38
rs769418476 Chr.1: 111350158-111350158 on Build GRCh38
rs952152137 Chr.1: 111349941-111349941 on Build GRCh38
rs1014927044 Chr.1: 111350187-111350187 on Build GRCh38
rs77570804 Chr.1: 111350247-111350247 on Build GRCh38
rs530043333 Chr.1: 111350279-111350279 on Build GRCh38
rs1027681850 Chr.1: 111349926-111349926 on Build GRCh38
rs757074329 Chr.1: 111350260-111350260 on Build GRCh38
rs116979896 Chr.1: 111349883-111349883 on Build GRCh38

View Full Product Details