Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 111160917-111161508 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEPT1
Entrez Gene ID
10390
Gene Name
choline/ethanolamine phosphotransferase 1
Gene Aliases
-
UniGene
Hs.363572
Gene Chromosome Location
Chr.1: 111139436-111185102 on Build GRCh38
Transcripts
NM_001007794.1 NM_006090.3 XM_006710285.3 XM_011540496.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1332653 c.546_547insT p.C185fs*28 Chr.1: 111161213-111161214 on Build GRCh38
6296710 c.557G>T p.C186F Chr.1: 111161224-111161224 on Build GRCh38
893757 c.587C>T p.A196V Chr.1: 111161254-111161254 on Build GRCh38
1667803 c.594G>A p.W198* Chr.1: 111161261-111161261 on Build GRCh38
2119079 c.503G>T p.G168V Chr.1: 111161170-111161170 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1064008 Chr.1: 111161224-111161224 on Build GRCh38
rs978414756 Chr.1: 111161457-111161457 on Build GRCh38
rs559005805 Chr.1: 111161426-111161426 on Build GRCh38
rs960516830 Chr.1: 111161418-111161418 on Build GRCh38
rs762302823 Chr.1: 111161313-111161313 on Build GRCh38
rs76144595 Chr.1: 111161229-111161229 on Build GRCh38
rs571497696 Chr.1: 111160954-111160954 on Build GRCh38
rs539676295 Chr.1: 111161385-111161385 on Build GRCh38
rs975260995 Chr.1: 111161442-111161442 on Build GRCh38
rs376073985 Chr.1: 111161049-111161049 on Build GRCh38
rs148174312 Chr.1: 111161266-111161266 on Build GRCh38
rs768037139 Chr.1: 111161317-111161317 on Build GRCh38
rs769589248 Chr.1: 111161255-111161255 on Build GRCh38
rs771804613 Chr.1: 111161250-111161250 on Build GRCh38
rs898692117 Chr.1: 111161056-111161056 on Build GRCh38
rs554005812 Chr.1: 111161311-111161311 on Build GRCh38
rs753868920 Chr.1: 111161337-111161337 on Build GRCh38
rs565968002 Chr.1: 111161465-111161466 on Build GRCh38
rs567601468 Chr.1: 111161207-111161207 on Build GRCh38
rs1035845782 Chr.1: 111161383-111161383 on Build GRCh38
rs770875885 Chr.1: 111161267-111161267 on Build GRCh38
rs780048500 Chr.1: 111161150-111161150 on Build GRCh38
rs202065837 Chr.1: 111161137-111161137 on Build GRCh38
rs978424457 Chr.1: 111161050-111161050 on Build GRCh38
rs779040820 Chr.1: 111161237-111161237 on Build GRCh38
rs779927872 Chr.1: 111161450-111161451 on Build GRCh38
rs564779174 Chr.1: 111160969-111160970 on Build GRCh38
rs968462081 Chr.1: 111161449-111161449 on Build GRCh38
rs957162561 Chr.1: 111161432-111161432 on Build GRCh38
rs959816249 Chr.1: 111160967-111160967 on Build GRCh38
rs759078198 Chr.1: 111161287-111161287 on Build GRCh38
rs370896689 Chr.1: 111161109-111161109 on Build GRCh38
rs533901597 Chr.1: 111161120-111161120 on Build GRCh38
rs754992953 Chr.1: 111161340-111161340 on Build GRCh38
rs922450419 Chr.1: 111161136-111161136 on Build GRCh38
rs755260487 Chr.1: 111161200-111161200 on Build GRCh38
rs747949785 Chr.1: 111161239-111161239 on Build GRCh38
rs761145344 Chr.1: 111161322-111161322 on Build GRCh38
rs764965634 Chr.1: 111161107-111161107 on Build GRCh38
rs371036014 Chr.1: 111161103-111161103 on Build GRCh38
rs531230636 Chr.1: 111161038-111161038 on Build GRCh38
rs1006138836 Chr.1: 111161270-111161270 on Build GRCh38
rs775237829 Chr.1: 111161312-111161312 on Build GRCh38
rs753941514 Chr.1: 111161195-111161195 on Build GRCh38
rs548232900 Chr.1: 111161054-111161054 on Build GRCh38
rs181815696 Chr.1: 111160969-111160969 on Build GRCh38
rs568145466 Chr.1: 111161084-111161084 on Build GRCh38
rs397808502 Chr.1: 111160970-111160971 on Build GRCh38
rs934077047 Chr.1: 111161463-111161463 on Build GRCh38
rs924310674 Chr.1: 111161467-111161467 on Build GRCh38
rs568789463 Chr.1: 111161037-111161037 on Build GRCh38
rs924486980 Chr.1: 111161086-111161086 on Build GRCh38
rs143092939 Chr.1: 111161392-111161393 on Build GRCh38
rs1022680304 Chr.1: 111161434-111161434 on Build GRCh38
rs967315381 Chr.1: 111161332-111161332 on Build GRCh38
rs915570156 Chr.1: 111160970-111160970 on Build GRCh38
rs766906686 Chr.1: 111161329-111161329 on Build GRCh38
rs776059319 Chr.1: 111161279-111161279 on Build GRCh38
rs186592058 Chr.1: 111161320-111161320 on Build GRCh38
rs756614833 Chr.1: 111161142-111161142 on Build GRCh38
rs762214682 Chr.1: 111161114-111161114 on Build GRCh38
rs71096395 Chr.1: 111160954-111160955 on Build GRCh38
rs377593173 Chr.1: 111161151-111161151 on Build GRCh38

View Full Product Details