Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 1369607-1370200 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FKBP1A
Entrez Gene ID
2280
Gene Name
FK506 binding protein 1A
Gene Aliases
FKBP-12, FKBP-1A, FKBP1, FKBP12, PKC12, PKCI2, PPIASE
UniGene
Hs.471933
Gene Chromosome Location
Chr.20: 1368977-1393172 on Build GRCh38
Transcripts
NM_000801.4 NM_001199786.1 NR_037661.1 NR_040047.1 NR_040048.1 NR_040049.1

Gene Symbol
FKBP1A-SDCBP2
Entrez Gene ID
100528031
Gene Name
FKBP1A-SDCBP2 readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.471933
Gene Chromosome Location
Chr.20: 1309909-1393172 on Build GRCh38
Transcripts
NM_000801.4 NM_001199786.1 NR_037661.1 NR_040047.1 NR_040048.1 NR_040049.1

Gene Symbol
SDCBP2-AS1
Entrez Gene ID
100507495
Gene Name
SDCBP2 antisense RNA 1
Gene Aliases
-
UniGene
Hs.655785
Gene Chromosome Location
Chr.20: 1325343-1378735 on Build GRCh38
Transcripts
NM_000801.4 NM_001199786.1 NR_037661.1 NR_040047.1 NR_040048.1 NR_040049.1

SNP Detail

SNP ID Chromosome Location
rs1052927128 Chr.20: 1370010-1370010 on Build GRCh38
rs935838301 Chr.20: 1370008-1370008 on Build GRCh38
rs1040097127 Chr.20: 1369895-1369895 on Build GRCh38
rs566192347 Chr.20: 1369731-1369731 on Build GRCh38
rs529713198 Chr.20: 1370142-1370142 on Build GRCh38
rs983838887 Chr.20: 1369733-1369733 on Build GRCh38
rs145404495 Chr.20: 1369930-1369930 on Build GRCh38
rs376131801 Chr.20: 1369701-1369701 on Build GRCh38
rs6041749 Chr.20: 1370028-1370028 on Build GRCh38
rs537120656 Chr.20: 1369883-1369883 on Build GRCh38
rs1008569706 Chr.20: 1369725-1369725 on Build GRCh38
rs190714238 Chr.20: 1369642-1369642 on Build GRCh38
rs773496036 Chr.20: 1369769-1369769 on Build GRCh38
rs530800255 Chr.20: 1369931-1369932 on Build GRCh38
rs758498752 Chr.20: 1369724-1369724 on Build GRCh38
rs1027846356 Chr.20: 1369920-1369920 on Build GRCh38
rs564894185 Chr.20: 1370086-1370086 on Build GRCh38
rs1000263220 Chr.20: 1369702-1369707 on Build GRCh38
rs529651885 Chr.20: 1369728-1369728 on Build GRCh38
rs559491604 Chr.20: 1369730-1369730 on Build GRCh38
rs900231969 Chr.20: 1370071-1370071 on Build GRCh38
rs45473597 Chr.20: 1369688-1369688 on Build GRCh38
rs773117898 Chr.20: 1370014-1370014 on Build GRCh38
rs911607525 Chr.20: 1370132-1370132 on Build GRCh38
rs1038693829 Chr.20: 1370141-1370141 on Build GRCh38
rs112687529 Chr.20: 1370115-1370118 on Build GRCh38
rs916670187 Chr.20: 1369789-1369789 on Build GRCh38
rs573002082 Chr.20: 1369797-1369797 on Build GRCh38
rs34401063 Chr.20: 1369723-1369723 on Build GRCh38
rs960658819 Chr.20: 1369932-1369932 on Build GRCh38
rs8392 Chr.20: 1370065-1370065 on Build GRCh38
rs944368233 Chr.20: 1370046-1370046 on Build GRCh38
rs550608954 Chr.20: 1369649-1369649 on Build GRCh38
rs529373359 Chr.20: 1369821-1369822 on Build GRCh38
rs756346750 Chr.20: 1369723-1369724 on Build GRCh38
rs769654835 Chr.20: 1370055-1370055 on Build GRCh38
rs755260902 Chr.20: 1369978-1369979 on Build GRCh38
rs756496309 Chr.20: 1369921-1369921 on Build GRCh38
rs575859500 Chr.20: 1369701-1369705 on Build GRCh38
rs1056580937 Chr.20: 1369675-1369678 on Build GRCh38
rs375283376 Chr.20: 1370006-1370006 on Build GRCh38
rs776938605 Chr.20: 1369910-1369910 on Build GRCh38
rs11712 Chr.20: 1369864-1369864 on Build GRCh38
rs558030239 Chr.20: 1369808-1369808 on Build GRCh38
rs11550413 Chr.20: 1369837-1369837 on Build GRCh38
rs530480273 Chr.20: 1369648-1369648 on Build GRCh38
rs931726108 Chr.20: 1370128-1370128 on Build GRCh38
rs980327524 Chr.20: 1369997-1369997 on Build GRCh38
rs879138498 Chr.20: 1369722-1369723 on Build GRCh38
rs77001193 Chr.20: 1369720-1369720 on Build GRCh38
rs752454371 Chr.20: 1369904-1369904 on Build GRCh38
rs146670225 Chr.20: 1369698-1369698 on Build GRCh38
rs544299004 Chr.20: 1370121-1370121 on Build GRCh38
rs45553532 Chr.20: 1369656-1369656 on Build GRCh38
rs900283691 Chr.20: 1369905-1369905 on Build GRCh38
rs530641430 Chr.20: 1370117-1370117 on Build GRCh38
rs887918095 Chr.20: 1370152-1370152 on Build GRCh38
rs935958291 Chr.20: 1369815-1369815 on Build GRCh38
rs891478235 Chr.20: 1369722-1369722 on Build GRCh38
rs755407950 Chr.20: 1370031-1370032 on Build GRCh38
rs149201864 Chr.20: 1370137-1370137 on Build GRCh38
rs939441093 Chr.20: 1369671-1369671 on Build GRCh38
rs948589309 Chr.20: 1369689-1369689 on Build GRCh38
rs397795937 Chr.20: 1369732-1369732 on Build GRCh38
rs369976663 Chr.20: 1369631-1369631 on Build GRCh38
rs73581254 Chr.20: 1369919-1369919 on Build GRCh38
rs187106305 Chr.20: 1369638-1369638 on Build GRCh38
rs183005812 Chr.20: 1369756-1369756 on Build GRCh38
rs762713394 Chr.20: 1370111-1370111 on Build GRCh38
rs1052750099 Chr.20: 1369720-1369720 on Build GRCh38
rs1018655649 Chr.20: 1369726-1369726 on Build GRCh38
rs550532523 Chr.20: 1369988-1369988 on Build GRCh38
rs960709715 Chr.20: 1369761-1369761 on Build GRCh38
rs907945653 Chr.20: 1369650-1369650 on Build GRCh38
rs992201376 Chr.20: 1369939-1369939 on Build GRCh38
rs553228856 Chr.20: 1369990-1369990 on Build GRCh38
rs1052976810 Chr.20: 1369840-1369840 on Build GRCh38
rs943145785 Chr.20: 1370133-1370133 on Build GRCh38
rs987321301 Chr.20: 1370127-1370127 on Build GRCh38
rs760809823 Chr.20: 1369799-1369799 on Build GRCh38
rs564998081 Chr.20: 1369888-1369888 on Build GRCh38
rs931568629 Chr.20: 1370173-1370173 on Build GRCh38
rs772224870 Chr.20: 1370005-1370005 on Build GRCh38
rs770618418 Chr.20: 1369733-1369733 on Build GRCh38
rs895623307 Chr.20: 1369685-1369685 on Build GRCh38
rs916646224 Chr.20: 1369971-1369971 on Build GRCh38

View Full Product Details