Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 36184892-36185417 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPB41L1
Entrez Gene ID
2036
Gene Name
erythrocyte membrane protein band 4.1 like 1
Gene Aliases
4.1N, MRD11
UniGene
Hs.437422
Gene Chromosome Location
Chr.20: 36064891-36232799 on Build GRCh38
Transcripts
NM_001258329.1 NM_001258330.1 NM_001258331.1 NM_012156.2 NM_177996.2 XM_011528667.1 XM_011528671.1 XM_011528673.1 XM_011528677.1 XM_011528681.1 XM_011528682.1 XM_011528683.1 XM_011528685.1 XM_017027711.1 XM_017027712.1 XM_017027713.1 XM_017027714.1 XM_017027715.1 XM_017027716.1 XM_017027717.1 XM_017027718.1 XM_017027719.1 XM_017027720.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6375971 c.662A>T p.Q221L Chr.20: 36185212-36185212 on Build GRCh38
6415898 c.764T>A p.M255K Chr.20: 36185314-36185314 on Build GRCh38
7184107 c.633C>T p.F211F Chr.20: 36185276-36185276 on Build GRCh38
6654597 c.539C>T p.T180M Chr.20: 36185182-36185182 on Build GRCh38
4909635 c.741C>T p.T247T Chr.20: 36185291-36185291 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs950828460 Chr.20: 36185037-36185037 on Build GRCh38
rs749076422 Chr.20: 36185091-36185091 on Build GRCh38
rs770063780 Chr.20: 36185258-36185258 on Build GRCh38
rs773677983 Chr.20: 36185385-36185385 on Build GRCh38
rs528130768 Chr.20: 36185095-36185095 on Build GRCh38
rs368264497 Chr.20: 36185370-36185370 on Build GRCh38
rs145441241 Chr.20: 36185216-36185216 on Build GRCh38
rs151312225 Chr.20: 36185293-36185293 on Build GRCh38
rs747080627 Chr.20: 36185325-36185325 on Build GRCh38
rs757020463 Chr.20: 36185292-36185292 on Build GRCh38
rs532730569 Chr.20: 36185011-36185011 on Build GRCh38
rs899507158 Chr.20: 36185068-36185068 on Build GRCh38
rs778844004 Chr.20: 36185212-36185212 on Build GRCh38
rs559930402 Chr.20: 36184953-36184953 on Build GRCh38
rs767155612 Chr.20: 36185168-36185168 on Build GRCh38
rs144222676 Chr.20: 36185204-36185204 on Build GRCh38
rs748336862 Chr.20: 36185265-36185265 on Build GRCh38
rs765764837 Chr.20: 36185156-36185156 on Build GRCh38
rs770152364 Chr.20: 36185376-36185376 on Build GRCh38
rs924028893 Chr.20: 36185189-36185189 on Build GRCh38
rs376321014 Chr.20: 36185290-36185290 on Build GRCh38
rs148438227 Chr.20: 36185255-36185255 on Build GRCh38
rs909444779 Chr.20: 36185054-36185054 on Build GRCh38
rs943638131 Chr.20: 36185008-36185008 on Build GRCh38
rs374238583 Chr.20: 36185362-36185362 on Build GRCh38
rs6142528 Chr.20: 36185205-36185205 on Build GRCh38
rs552462148 Chr.20: 36185047-36185047 on Build GRCh38
rs199985301 Chr.20: 36185277-36185277 on Build GRCh38
rs1007197211 Chr.20: 36184939-36184939 on Build GRCh38
rs987302350 Chr.20: 36185019-36185019 on Build GRCh38
rs866698199 Chr.20: 36185317-36185317 on Build GRCh38
rs772092946 Chr.20: 36185217-36185217 on Build GRCh38
rs757202579 Chr.20: 36185113-36185113 on Build GRCh38
rs17093425 Chr.20: 36184991-36184991 on Build GRCh38
rs377709134 Chr.20: 36185155-36185155 on Build GRCh38
rs747135476 Chr.20: 36185229-36185229 on Build GRCh38
rs760911252 Chr.20: 36185107-36185107 on Build GRCh38
rs780018641 Chr.20: 36185321-36185321 on Build GRCh38
rs753653225 Chr.20: 36185188-36185188 on Build GRCh38
rs1042416629 Chr.20: 36185180-36185180 on Build GRCh38
rs893060314 Chr.20: 36184931-36184931 on Build GRCh38
rs541540717 Chr.20: 36184918-36184918 on Build GRCh38
rs777338272 Chr.20: 36185187-36185187 on Build GRCh38
rs745861970 Chr.20: 36185072-36185072 on Build GRCh38
rs66823537 Chr.20: 36185132-36185132 on Build GRCh38
rs751041730 Chr.20: 36185160-36185160 on Build GRCh38
rs768904051 Chr.20: 36185246-36185246 on Build GRCh38
rs745942930 Chr.20: 36185102-36185102 on Build GRCh38
rs755922186 Chr.20: 36185080-36185080 on Build GRCh38
rs753437652 Chr.20: 36185291-36185291 on Build GRCh38
rs755085093 Chr.20: 36185333-36185333 on Build GRCh38
rs140658821 Chr.20: 36185313-36185313 on Build GRCh38
rs772144203 Chr.20: 36185104-36185104 on Build GRCh38
rs781412221 Chr.20: 36185358-36185358 on Build GRCh38
rs778773659 Chr.20: 36185097-36185097 on Build GRCh38
rs201058000 Chr.20: 36185271-36185271 on Build GRCh38
rs780851676 Chr.20: 36185377-36185378 on Build GRCh38
rs780140662 Chr.20: 36185218-36185218 on Build GRCh38
rs752354193 Chr.20: 36185183-36185183 on Build GRCh38
rs773802695 Chr.20: 36185264-36185264 on Build GRCh38
rs1033634508 Chr.20: 36185215-36185215 on Build GRCh38
rs945508190 Chr.20: 36184992-36184992 on Build GRCh38
rs556946313 Chr.20: 36185300-36185300 on Build GRCh38
rs114208832 Chr.20: 36185318-36185318 on Build GRCh38
rs570723490 Chr.20: 36185083-36185083 on Build GRCh38
rs917937075 Chr.20: 36185153-36185153 on Build GRCh38
rs1026030614 Chr.20: 36185342-36185342 on Build GRCh38
rs140844629 Chr.20: 36184985-36184985 on Build GRCh38
rs997307090 Chr.20: 36184996-36184996 on Build GRCh38
rs530103909 Chr.20: 36184954-36184954 on Build GRCh38
rs768863926 Chr.20: 36185115-36185115 on Build GRCh38
rs954766450 Chr.20: 36185018-36185018 on Build GRCh38
rs965805186 Chr.20: 36184993-36184993 on Build GRCh38
rs767030871 Chr.20: 36185268-36185268 on Build GRCh38
rs759054810 Chr.20: 36185166-36185166 on Build GRCh38
rs886208909 Chr.20: 36185379-36185379 on Build GRCh38
rs142551778 Chr.20: 36185276-36185276 on Build GRCh38

View Full Product Details