Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 33370403-33370796 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDK5RAP1
Entrez Gene ID
51654
Gene Name
CDK5 regulatory subunit associated protein 1
Gene Aliases
C20orf34, C42, CGI-05, HSPC167
UniGene
Hs.435952
Gene Chromosome Location
Chr.20: 33358839-33402768 on Build GRCh38
Transcripts
NM_001278167.1 NM_001278168.1 NM_001278169.1 NM_016082.4 NM_016408.3 XM_011528856.2 XM_011528857.2 XM_011528858.1 XM_011528859.2 XM_011528860.1 XM_017027876.1 XM_017027877.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5760155 c.1345G>T p.E449* Chr.20: 33370546-33370546 on Build GRCh38
478028 c.1295T>A p.F432Y Chr.20: 33370596-33370596 on Build GRCh38
1632241 c.1264G>T p.V422L Chr.20: 33370627-33370627 on Build GRCh38
7103931 c.1386G>A p.M462I Chr.20: 33370505-33370505 on Build GRCh38
1172712 c.1342C>T p.R448W Chr.20: 33370549-33370549 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756854389 Chr.20: 33370482-33370482 on Build GRCh38
rs267605887 Chr.20: 33370607-33370607 on Build GRCh38
rs146896589 Chr.20: 33370540-33370540 on Build GRCh38
rs554756261 Chr.20: 33370741-33370741 on Build GRCh38
rs759669447 Chr.20: 33370451-33370451 on Build GRCh38
rs774284099 Chr.20: 33370670-33370670 on Build GRCh38
rs139039517 Chr.20: 33370571-33370571 on Build GRCh38
rs933109054 Chr.20: 33370579-33370579 on Build GRCh38
rs376440387 Chr.20: 33370666-33370666 on Build GRCh38
rs746138461 Chr.20: 33370649-33370649 on Build GRCh38
rs759662758 Chr.20: 33370496-33370497 on Build GRCh38
rs763905669 Chr.20: 33370676-33370676 on Build GRCh38
rs1037309139 Chr.20: 33370718-33370718 on Build GRCh38
rs910293110 Chr.20: 33370629-33370629 on Build GRCh38
rs775495983 Chr.20: 33370656-33370656 on Build GRCh38
rs752735399 Chr.20: 33370458-33370458 on Build GRCh38
rs149014886 Chr.20: 33370663-33370663 on Build GRCh38
rs773663535 Chr.20: 33370507-33370507 on Build GRCh38
rs142957144 Chr.20: 33370577-33370577 on Build GRCh38
rs753422405 Chr.20: 33370612-33370612 on Build GRCh38
rs755661320 Chr.20: 33370487-33370487 on Build GRCh38
rs904819328 Chr.20: 33370591-33370591 on Build GRCh38
rs769735883 Chr.20: 33370651-33370651 on Build GRCh38
rs770995894 Chr.20: 33370518-33370518 on Build GRCh38
rs142017117 Chr.20: 33370500-33370500 on Build GRCh38
rs751315997 Chr.20: 33370567-33370567 on Build GRCh38
rs765029786 Chr.20: 33370455-33370455 on Build GRCh38
rs775371763 Chr.20: 33370530-33370530 on Build GRCh38
rs763913713 Chr.20: 33370470-33370470 on Build GRCh38
rs751285624 Chr.20: 33370696-33370696 on Build GRCh38
rs747413318 Chr.20: 33370515-33370515 on Build GRCh38
rs765412202 Chr.20: 33370553-33370554 on Build GRCh38
rs968605198 Chr.20: 33370527-33370527 on Build GRCh38
rs745307895 Chr.20: 33370485-33370485 on Build GRCh38
rs929465491 Chr.20: 33370557-33370557 on Build GRCh38
rs747323110 Chr.20: 33370627-33370627 on Build GRCh38
rs758182787 Chr.20: 33370459-33370459 on Build GRCh38
rs779285423 Chr.20: 33370494-33370494 on Build GRCh38
rs771386890 Chr.20: 33370636-33370636 on Build GRCh38
rs929585178 Chr.20: 33370768-33370768 on Build GRCh38
rs1021234722 Chr.20: 33370695-33370695 on Build GRCh38
rs750146436 Chr.20: 33370583-33370583 on Build GRCh38
rs751187334 Chr.20: 33370472-33370472 on Build GRCh38
rs769980619 Chr.20: 33370529-33370529 on Build GRCh38
rs759524666 Chr.20: 33370528-33370528 on Build GRCh38
rs202040830 Chr.20: 33370559-33370559 on Build GRCh38
rs371213961 Chr.20: 33370616-33370616 on Build GRCh38
rs201086142 Chr.20: 33370450-33370450 on Build GRCh38
rs762924338 Chr.20: 33370548-33370548 on Build GRCh38
rs757152226 Chr.20: 33370750-33370750 on Build GRCh38
rs374785718 Chr.20: 33370640-33370640 on Build GRCh38
rs199766779 Chr.20: 33370584-33370584 on Build GRCh38
rs375505184 Chr.20: 33370502-33370502 on Build GRCh38
rs758938214 Chr.20: 33370613-33370613 on Build GRCh38
rs1032232101 Chr.20: 33370621-33370621 on Build GRCh38
rs201072879 Chr.20: 33370441-33370441 on Build GRCh38
rs776928550 Chr.20: 33370519-33370519 on Build GRCh38

View Full Product Details