Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 23260722-23261314 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCR
Entrez Gene ID
613
Gene Name
BCR, RhoGEF and GTPase activating protein
Gene Aliases
ALL, BCR1, CML, D22S11, D22S662, PHL
UniGene
Hs.517461
Gene Chromosome Location
Chr.22: 23180365-23318037 on Build GRCh38
Transcripts
NM_004327.3 NM_021574.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5681304 c.1528G>T p.E510* Chr.22: 23261016-23261016 on Build GRCh38
6611110 c.1540C>G p.L514V Chr.22: 23261028-23261028 on Build GRCh38
3309465 c.1549C>T p.L517L Chr.22: 23261037-23261037 on Build GRCh38
3309463 c.1539C>G p.Y513* Chr.22: 23261027-23261027 on Build GRCh38
95780 c.1542G>T p.L514L Chr.22: 23261030-23261030 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs528865767 Chr.22: 23260771-23260771 on Build GRCh38
rs975732213 Chr.22: 23260846-23260846 on Build GRCh38
rs146490261 Chr.22: 23261033-23261033 on Build GRCh38
rs1029687544 Chr.22: 23261163-23261163 on Build GRCh38
rs547286402 Chr.22: 23260793-23260793 on Build GRCh38
rs189544210 Chr.22: 23261278-23261278 on Build GRCh38
rs749888340 Chr.22: 23260828-23260828 on Build GRCh38
rs779526967 Chr.22: 23260834-23260834 on Build GRCh38
rs75267590 Chr.22: 23260863-23260863 on Build GRCh38
rs548152037 Chr.22: 23261275-23261275 on Build GRCh38
rs116891220 Chr.22: 23261085-23261085 on Build GRCh38
rs543283013 Chr.22: 23260742-23260742 on Build GRCh38
rs778821199 Chr.22: 23260851-23260851 on Build GRCh38
rs751227235 Chr.22: 23260836-23260836 on Build GRCh38
rs975785987 Chr.22: 23261132-23261132 on Build GRCh38
rs367931519 Chr.22: 23260960-23260960 on Build GRCh38
rs1014036127 Chr.22: 23260967-23260967 on Build GRCh38
rs9624070 Chr.22: 23261282-23261282 on Build GRCh38
rs1036834599 Chr.22: 23260907-23260907 on Build GRCh38
rs549088500 Chr.22: 23260836-23260836 on Build GRCh38
rs752427251 Chr.22: 23260943-23260943 on Build GRCh38
rs775542219 Chr.22: 23260866-23260866 on Build GRCh38
rs746986541 Chr.22: 23260991-23260991 on Build GRCh38
rs755700880 Chr.22: 23260830-23260830 on Build GRCh38
rs764892702 Chr.22: 23261090-23261090 on Build GRCh38
rs538787836 Chr.22: 23260999-23260999 on Build GRCh38
rs971278814 Chr.22: 23260776-23260776 on Build GRCh38
rs759394069 Chr.22: 23261001-23261001 on Build GRCh38
rs752375839 Chr.22: 23261096-23261096 on Build GRCh38
rs754721475 Chr.22: 23260937-23260937 on Build GRCh38
rs765797777 Chr.22: 23261083-23261083 on Build GRCh38
rs778432175 Chr.22: 23260942-23260942 on Build GRCh38
rs763924204 Chr.22: 23261105-23261105 on Build GRCh38
rs748476079 Chr.22: 23261016-23261016 on Build GRCh38
rs955382655 Chr.22: 23261187-23261187 on Build GRCh38
rs1017467874 Chr.22: 23260780-23260780 on Build GRCh38
rs967471461 Chr.22: 23260822-23260822 on Build GRCh38
rs879757242 Chr.22: 23260752-23260752 on Build GRCh38
rs775739799 Chr.22: 23260860-23260862 on Build GRCh38
rs780890083 Chr.22: 23261057-23261059 on Build GRCh38
rs984208921 Chr.22: 23260772-23260772 on Build GRCh38
rs373782745 Chr.22: 23260933-23260933 on Build GRCh38
rs967146806 Chr.22: 23261130-23261130 on Build GRCh38
rs34900178 Chr.22: 23261287-23261288 on Build GRCh38
rs773827153 Chr.22: 23261054-23261054 on Build GRCh38
rs768888530 Chr.22: 23260876-23260876 on Build GRCh38
rs373724162 Chr.22: 23260842-23260842 on Build GRCh38
rs535817727 Chr.22: 23260753-23260753 on Build GRCh38
rs924451548 Chr.22: 23260853-23260853 on Build GRCh38
rs922893918 Chr.22: 23261165-23261165 on Build GRCh38
rs770497328 Chr.22: 23260887-23260895 on Build GRCh38
rs765957243 Chr.22: 23260922-23260922 on Build GRCh38
rs771925460 Chr.22: 23260856-23260856 on Build GRCh38
rs1034158900 Chr.22: 23261070-23261070 on Build GRCh38
rs932219661 Chr.22: 23260854-23260854 on Build GRCh38
rs9624069 Chr.22: 23260891-23260891 on Build GRCh38
rs750991222 Chr.22: 23260914-23260914 on Build GRCh38
rs996821947 Chr.22: 23261156-23261156 on Build GRCh38
rs951652674 Chr.22: 23260757-23260757 on Build GRCh38
rs776136533 Chr.22: 23261084-23261084 on Build GRCh38
rs769891719 Chr.22: 23260865-23260865 on Build GRCh38
rs990394273 Chr.22: 23261279-23261279 on Build GRCh38
rs768857111 Chr.22: 23261000-23261000 on Build GRCh38
rs953619787 Chr.22: 23261186-23261186 on Build GRCh38
rs550173854 Chr.22: 23261115-23261115 on Build GRCh38
rs1034104852 Chr.22: 23260792-23260792 on Build GRCh38
rs774730471 Chr.22: 23261015-23261015 on Build GRCh38
rs758156437 Chr.22: 23261102-23261102 on Build GRCh38
rs570068657 Chr.22: 23261250-23261250 on Build GRCh38
rs777646028 Chr.22: 23260971-23260971 on Build GRCh38
rs754702152 Chr.22: 23260844-23260844 on Build GRCh38
rs990277047 Chr.22: 23260799-23260799 on Build GRCh38
rs765159430 Chr.22: 23261049-23261049 on Build GRCh38
rs994118605 Chr.22: 23260744-23260744 on Build GRCh38
rs1003730835 Chr.22: 23261002-23261002 on Build GRCh38
rs1020481971 Chr.22: 23261124-23261124 on Build GRCh38
rs144134677 Chr.22: 23260997-23260997 on Build GRCh38
rs115539548 Chr.22: 23260873-23260873 on Build GRCh38
rs760225629 Chr.22: 23260920-23260920 on Build GRCh38
rs933909520 Chr.22: 23260841-23260841 on Build GRCh38
rs761317547 Chr.22: 23261065-23261065 on Build GRCh38
rs964037015 Chr.22: 23260800-23260800 on Build GRCh38
rs752454484 Chr.22: 23261265-23261265 on Build GRCh38
rs1011390997 Chr.22: 23261107-23261107 on Build GRCh38
rs774846395 Chr.22: 23260887-23260887 on Build GRCh38
rs115439368 Chr.22: 23260852-23260852 on Build GRCh38
rs575527925 Chr.22: 23260855-23260855 on Build GRCh38
rs897794258 Chr.22: 23260741-23260742 on Build GRCh38
rs865987665 Chr.22: 23260819-23260819 on Build GRCh38
rs975413358 Chr.22: 23260811-23260811 on Build GRCh38

View Full Product Details