Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 26432970-26433526 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASPHD2
Entrez Gene ID
57168
Gene Name
aspartate beta-hydroxylase domain containing 2
Gene Aliases
-
UniGene
Hs.567547
Gene Chromosome Location
Chr.22: 26429314-26445012 on Build GRCh38
Transcripts
NM_020437.4

SNP Detail

SNP ID Chromosome Location
rs867461915 Chr.22: 26433179-26433179 on Build GRCh38
rs1056472323 Chr.22: 26433403-26433403 on Build GRCh38
rs763428397 Chr.22: 26433081-26433081 on Build GRCh38
rs543618476 Chr.22: 26433392-26433392 on Build GRCh38
rs200432654 Chr.22: 26433277-26433277 on Build GRCh38
rs919285636 Chr.22: 26433317-26433317 on Build GRCh38
rs531492458 Chr.22: 26433386-26433386 on Build GRCh38
rs1027970350 Chr.22: 26433122-26433122 on Build GRCh38
rs1057390478 Chr.22: 26433374-26433374 on Build GRCh38
rs5844702 Chr.22: 26433372-26433373 on Build GRCh38
rs765914950 Chr.22: 26433027-26433027 on Build GRCh38
rs764499714 Chr.22: 26433199-26433199 on Build GRCh38
rs1009699949 Chr.22: 26433038-26433038 on Build GRCh38
rs955253596 Chr.22: 26433146-26433146 on Build GRCh38
rs139634413 Chr.22: 26433430-26433430 on Build GRCh38
rs751834516 Chr.22: 26433218-26433218 on Build GRCh38
rs869120228 Chr.22: 26433373-26433374 on Build GRCh38
rs892521951 Chr.22: 26433423-26433423 on Build GRCh38
rs922481732 Chr.22: 26433166-26433166 on Build GRCh38
rs545108313 Chr.22: 26433230-26433230 on Build GRCh38
rs866473356 Chr.22: 26433470-26433470 on Build GRCh38
rs377498486 Chr.22: 26433055-26433055 on Build GRCh38
rs397726718 Chr.22: 26433382-26433383 on Build GRCh38
rs767755023 Chr.22: 26433431-26433431 on Build GRCh38
rs377458767 Chr.22: 26433111-26433111 on Build GRCh38
rs955886054 Chr.22: 26433092-26433092 on Build GRCh38
rs942125061 Chr.22: 26433266-26433266 on Build GRCh38
rs1031333056 Chr.22: 26433089-26433089 on Build GRCh38
rs939315904 Chr.22: 26433389-26433389 on Build GRCh38
rs986558011 Chr.22: 26433157-26433157 on Build GRCh38
rs534639735 Chr.22: 26433316-26433316 on Build GRCh38
rs757457899 Chr.22: 26433429-26433429 on Build GRCh38
rs753779226 Chr.22: 26433357-26433357 on Build GRCh38
rs191790128 Chr.22: 26433024-26433024 on Build GRCh38
rs926638230 Chr.22: 26433379-26433380 on Build GRCh38
rs955746213 Chr.22: 26433476-26433476 on Build GRCh38
rs201373264 Chr.22: 26433383-26433383 on Build GRCh38
rs1013471622 Chr.22: 26433002-26433002 on Build GRCh38
rs929276819 Chr.22: 26433348-26433348 on Build GRCh38
rs1029025902 Chr.22: 26433489-26433489 on Build GRCh38
rs904045146 Chr.22: 26433458-26433458 on Build GRCh38
rs948624242 Chr.22: 26433350-26433350 on Build GRCh38
rs79294516 Chr.22: 26433057-26433057 on Build GRCh38
rs763700908 Chr.22: 26433209-26433209 on Build GRCh38
rs77151567 Chr.22: 26433427-26433427 on Build GRCh38

View Full Product Details