Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 38982160-38982729 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APOBEC3B
Entrez Gene ID
9582
Gene Name
apolipoprotein B mRNA editing enzyme catalytic subunit 3B
Gene Aliases
A3B, APOBEC1L, ARCD3, ARP4, DJ742C19.2, PHRBNL, bK150C2.2
UniGene
Hs.226307
Gene Chromosome Location
Chr.22: 38982399-38992779 on Build GRCh38
Transcripts
NM_001270411.1 NM_004900.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1308165 c.3G>C p.M1I Chr.22: 38982456-38982456 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs181767284 Chr.22: 38982295-38982295 on Build GRCh38
rs374220745 Chr.22: 38982556-38982556 on Build GRCh38
rs1011738660 Chr.22: 38982481-38982481 on Build GRCh38
rs760880636 Chr.22: 38982519-38982519 on Build GRCh38
rs537212122 Chr.22: 38982441-38982442 on Build GRCh38
rs373482687 Chr.22: 38982465-38982465 on Build GRCh38
rs1021837508 Chr.22: 38982508-38982508 on Build GRCh38
rs376795731 Chr.22: 38982425-38982425 on Build GRCh38
rs373327958 Chr.22: 38982433-38982433 on Build GRCh38
rs973652995 Chr.22: 38982642-38982642 on Build GRCh38
rs921844191 Chr.22: 38982643-38982643 on Build GRCh38
rs747770155 Chr.22: 38982251-38982251 on Build GRCh38
rs78871147 Chr.22: 38982318-38982318 on Build GRCh38
rs549724171 Chr.22: 38982568-38982568 on Build GRCh38
rs113318548 Chr.22: 38982625-38982625 on Build GRCh38
rs763819604 Chr.22: 38982449-38982449 on Build GRCh38
rs568066803 Chr.22: 38982619-38982619 on Build GRCh38
rs778968630 Chr.22: 38982470-38982470 on Build GRCh38
rs576342247 Chr.22: 38982424-38982424 on Build GRCh38
rs768196828 Chr.22: 38982219-38982219 on Build GRCh38
rs6001353 Chr.22: 38982647-38982647 on Build GRCh38
rs201854400 Chr.22: 38982454-38982454 on Build GRCh38
rs778792296 Chr.22: 38982421-38982421 on Build GRCh38
rs1051711745 Chr.22: 38982670-38982670 on Build GRCh38
rs777144656 Chr.22: 38982435-38982435 on Build GRCh38
rs369907102 Chr.22: 38982428-38982428 on Build GRCh38
rs113903377 Chr.22: 38982247-38982247 on Build GRCh38
rs113491060 Chr.22: 38982234-38982235 on Build GRCh38
rs553902178 Chr.22: 38982260-38982260 on Build GRCh38
rs911536508 Chr.22: 38982694-38982694 on Build GRCh38
rs368812226 Chr.22: 38982444-38982444 on Build GRCh38
rs941871415 Chr.22: 38982393-38982393 on Build GRCh38
rs976672098 Chr.22: 38982268-38982276 on Build GRCh38
rs572414678 Chr.22: 38982279-38982279 on Build GRCh38
rs748601228 Chr.22: 38982509-38982509 on Build GRCh38
rs960051289 Chr.22: 38982580-38982580 on Build GRCh38
rs928700158 Chr.22: 38982355-38982355 on Build GRCh38
rs766157877 Chr.22: 38982696-38982696 on Build GRCh38
rs191137305 Chr.22: 38982676-38982676 on Build GRCh38
rs760342782 Chr.22: 38982446-38982446 on Build GRCh38
rs536822161 Chr.22: 38982246-38982246 on Build GRCh38
rs564167762 Chr.22: 38982388-38982388 on Build GRCh38
rs376485822 Chr.22: 38982475-38982475 on Build GRCh38
rs763458923 Chr.22: 38982570-38982570 on Build GRCh38
rs757233336 Chr.22: 38982420-38982420 on Build GRCh38
rs199854700 Chr.22: 38982436-38982436 on Build GRCh38
rs963169355 Chr.22: 38982637-38982637 on Build GRCh38
rs570622859 Chr.22: 38982661-38982661 on Build GRCh38
rs907690067 Chr.22: 38982378-38982378 on Build GRCh38
rs769594836 Chr.22: 38982585-38982585 on Build GRCh38
rs371321488 Chr.22: 38982520-38982520 on Build GRCh38
rs375334718 Chr.22: 38982505-38982505 on Build GRCh38
rs181909898 Chr.22: 38982697-38982697 on Build GRCh38
rs956233265 Chr.22: 38982230-38982230 on Build GRCh38
rs938761058 Chr.22: 38982377-38982377 on Build GRCh38
rs752306308 Chr.22: 38982445-38982445 on Build GRCh38
rs781402799 Chr.22: 38982503-38982503 on Build GRCh38
rs570497857 Chr.22: 38982555-38982555 on Build GRCh38
rs200275128 Chr.22: 38982238-38982239 on Build GRCh38
rs552480033 Chr.22: 38982545-38982545 on Build GRCh38
rs543764189 Chr.22: 38982429-38982429 on Build GRCh38
rs983521471 Chr.22: 38982604-38982604 on Build GRCh38
rs990350599 Chr.22: 38982239-38982244 on Build GRCh38
rs527629935 Chr.22: 38982476-38982476 on Build GRCh38
rs753668578 Chr.22: 38982411-38982411 on Build GRCh38
rs768999602 Chr.22: 38982498-38982498 on Build GRCh38
rs747184139 Chr.22: 38982486-38982486 on Build GRCh38
rs1037621430 Chr.22: 38982397-38982397 on Build GRCh38

View Full Product Details