Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 19187356-19187934 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLTCL1
Entrez Gene ID
8218
Gene Name
clathrin heavy chain like 1
Gene Aliases
CHC22, CLH22, CLTCL, CLTD
UniGene
Hs.368266
Gene Chromosome Location
Chr.22: 19179473-19291716 on Build GRCh38
Transcripts
NM_001835.3 NM_007098.3 XM_011530401.2 XM_017028953.1 XM_017028954.1 XM_017028955.1 XM_017028956.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
260150 c.4599C>A p.L1533L Chr.22: 19187564-19187564 on Build GRCh38
6630832 c.4533G>A p.A1511A Chr.22: 19187630-19187630 on Build GRCh38
5513665 c.4531G>A p.A1511T Chr.22: 19187632-19187632 on Build GRCh38
4646048 c.4487C>T p.A1496V Chr.22: 19187676-19187676 on Build GRCh38
5513666 c.4531G>A p.A1511T Chr.22: 19187632-19187632 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782250309 Chr.22: 19187713-19187713 on Build GRCh38
rs117414027 Chr.22: 19187719-19187719 on Build GRCh38
rs571605817 Chr.22: 19187585-19187585 on Build GRCh38
rs374313086 Chr.22: 19187639-19187639 on Build GRCh38
rs782482513 Chr.22: 19187561-19187561 on Build GRCh38
rs896822693 Chr.22: 19187786-19187786 on Build GRCh38
rs967079201 Chr.22: 19187691-19187691 on Build GRCh38
rs201078077 Chr.22: 19187753-19187753 on Build GRCh38
rs150055374 Chr.22: 19187435-19187435 on Build GRCh38
rs781945094 Chr.22: 19187625-19187625 on Build GRCh38
rs542871808 Chr.22: 19187848-19187848 on Build GRCh38
rs782029058 Chr.22: 19187720-19187720 on Build GRCh38
rs781870191 Chr.22: 19187742-19187742 on Build GRCh38
rs782506779 Chr.22: 19187772-19187772 on Build GRCh38
rs782379522 Chr.22: 19187611-19187611 on Build GRCh38
rs889520125 Chr.22: 19187816-19187816 on Build GRCh38
rs782618837 Chr.22: 19187711-19187711 on Build GRCh38
rs952233770 Chr.22: 19187600-19187600 on Build GRCh38
rs782562132 Chr.22: 19187578-19187578 on Build GRCh38
rs529186334 Chr.22: 19187836-19187836 on Build GRCh38
rs949278260 Chr.22: 19187736-19187736 on Build GRCh38
rs563563356 Chr.22: 19187820-19187820 on Build GRCh38
rs781914945 Chr.22: 19187429-19187429 on Build GRCh38
rs782067048 Chr.22: 19187637-19187637 on Build GRCh38
rs147730809 Chr.22: 19187498-19187498 on Build GRCh38
rs782236800 Chr.22: 19187591-19187591 on Build GRCh38
rs781824888 Chr.22: 19187557-19187557 on Build GRCh38
rs567198819 Chr.22: 19187769-19187769 on Build GRCh38
rs782043581 Chr.22: 19187536-19187536 on Build GRCh38
rs35119282 Chr.22: 19187508-19187509 on Build GRCh38
rs375337155 Chr.22: 19187780-19187780 on Build GRCh38
rs782202321 Chr.22: 19187703-19187703 on Build GRCh38
rs925146645 Chr.22: 19187470-19187470 on Build GRCh38
rs782084602 Chr.22: 19187627-19187627 on Build GRCh38
rs782370649 Chr.22: 19187732-19187732 on Build GRCh38
rs937787133 Chr.22: 19187488-19187488 on Build GRCh38
rs782694366 Chr.22: 19187726-19187726 on Build GRCh38
rs543372299 Chr.22: 19187795-19187795 on Build GRCh38
rs782022990 Chr.22: 19187615-19187615 on Build GRCh38
rs540713106 Chr.22: 19187727-19187727 on Build GRCh38
rs782146059 Chr.22: 19187748-19187748 on Build GRCh38
rs367845108 Chr.22: 19187700-19187700 on Build GRCh38
rs752255359 Chr.22: 19187857-19187857 on Build GRCh38
rs1027111957 Chr.22: 19187599-19187599 on Build GRCh38
rs73877048 Chr.22: 19187496-19187496 on Build GRCh38
rs782781279 Chr.22: 19187562-19187562 on Build GRCh38
rs781883437 Chr.22: 19187563-19187563 on Build GRCh38
rs782798990 Chr.22: 19187635-19187635 on Build GRCh38
rs781794442 Chr.22: 19187754-19187754 on Build GRCh38
rs781838510 Chr.22: 19187765-19187765 on Build GRCh38
rs782501177 Chr.22: 19187722-19187722 on Build GRCh38
rs1043985803 Chr.22: 19187464-19187464 on Build GRCh38
rs199562273 Chr.22: 19187649-19187649 on Build GRCh38
rs1048158304 Chr.22: 19187798-19187798 on Build GRCh38
rs1007705831 Chr.22: 19187831-19187831 on Build GRCh38
rs577516955 Chr.22: 19187791-19187791 on Build GRCh38
rs377501080 Chr.22: 19187630-19187630 on Build GRCh38
rs913997888 Chr.22: 19187433-19187433 on Build GRCh38
rs547779543 Chr.22: 19187623-19187623 on Build GRCh38
rs782060051 Chr.22: 19187540-19187540 on Build GRCh38
rs782454983 Chr.22: 19187755-19187755 on Build GRCh38
rs201496262 Chr.22: 19187590-19187590 on Build GRCh38
rs767038413 Chr.22: 19187819-19187819 on Build GRCh38
rs782617572 Chr.22: 19187759-19187759 on Build GRCh38
rs782352716 Chr.22: 19187517-19187517 on Build GRCh38
rs529068579 Chr.22: 19187434-19187434 on Build GRCh38
rs781811148 Chr.22: 19187763-19187763 on Build GRCh38
rs781835475 Chr.22: 19187647-19187647 on Build GRCh38
rs540973262 Chr.22: 19187757-19187757 on Build GRCh38
rs553137718 Chr.22: 19187710-19187710 on Build GRCh38
rs782757294 Chr.22: 19187741-19187741 on Build GRCh38
rs781787199 Chr.22: 19187636-19187636 on Build GRCh38
rs182434700 Chr.22: 19187583-19187583 on Build GRCh38
rs528638076 Chr.22: 19187886-19187886 on Build GRCh38
rs782646824 Chr.22: 19187666-19187666 on Build GRCh38
rs782133716 Chr.22: 19187522-19187522 on Build GRCh38
rs55790642 Chr.22: 19187701-19187701 on Build GRCh38
rs782324502 Chr.22: 19187725-19187725 on Build GRCh38
rs782200797 Chr.22: 19187516-19187516 on Build GRCh38
rs1013283284 Chr.22: 19187545-19187545 on Build GRCh38
rs781979391 Chr.22: 19187521-19187521 on Build GRCh38
rs782567059 Chr.22: 19187580-19187580 on Build GRCh38
rs1036699830 Chr.22: 19187550-19187550 on Build GRCh38
rs782801230 Chr.22: 19187525-19187525 on Build GRCh38
rs782714719 Chr.22: 19187542-19187542 on Build GRCh38

View Full Product Details