Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 32416944-32417509 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BPIFC
Entrez Gene ID
254240
Gene Name
BPI fold containing family C
Gene Aliases
BPIL2
UniGene
Hs.372939
Gene Chromosome Location
Chr.22: 32413845-32464465 on Build GRCh38
Transcripts
NM_174932.2 XM_011530088.2 XM_011530089.1 XM_011530090.1 XM_011530091.2 XM_017028740.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
726529 c.1304G>C p.G435A Chr.22: 32417105-32417105 on Build GRCh38
1033626 c.1282C>A p.L428I Chr.22: 32417127-32417127 on Build GRCh38
7100904 c.1280T>G p.I427S Chr.22: 32417129-32417129 on Build GRCh38
5930086 c.1286C>T p.S429L Chr.22: 32417123-32417123 on Build GRCh38
2939114 c.1287G>A p.S429S Chr.22: 32417122-32417122 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs552237642 Chr.22: 32417334-32417334 on Build GRCh38
rs1050295311 Chr.22: 32417121-32417121 on Build GRCh38
rs564322847 Chr.22: 32417206-32417206 on Build GRCh38
rs912524507 Chr.22: 32417101-32417101 on Build GRCh38
rs746412189 Chr.22: 32417048-32417048 on Build GRCh38
rs926078252 Chr.22: 32417486-32417486 on Build GRCh38
rs534866056 Chr.22: 32417353-32417353 on Build GRCh38
rs778475123 Chr.22: 32417122-32417122 on Build GRCh38
rs774877190 Chr.22: 32417189-32417190 on Build GRCh38
rs1022176260 Chr.22: 32417402-32417402 on Build GRCh38
rs146172628 Chr.22: 32417361-32417361 on Build GRCh38
rs929114230 Chr.22: 32417231-32417231 on Build GRCh38
rs780526134 Chr.22: 32417177-32417177 on Build GRCh38
rs183645369 Chr.22: 32417396-32417396 on Build GRCh38
rs750142966 Chr.22: 32417087-32417087 on Build GRCh38
rs936075447 Chr.22: 32417487-32417487 on Build GRCh38
rs772057416 Chr.22: 32417086-32417086 on Build GRCh38
rs374354376 Chr.22: 32417167-32417167 on Build GRCh38
rs766358369 Chr.22: 32417096-32417096 on Build GRCh38
rs3215462 Chr.22: 32417163-32417163 on Build GRCh38
rs954167941 Chr.22: 32417422-32417422 on Build GRCh38
rs887544783 Chr.22: 32417244-32417244 on Build GRCh38
rs549825562 Chr.22: 32417221-32417221 on Build GRCh38
rs892002656 Chr.22: 32417384-32417384 on Build GRCh38
rs201131557 Chr.22: 32417033-32417033 on Build GRCh38
rs1009919103 Chr.22: 32417239-32417239 on Build GRCh38
rs1049915705 Chr.22: 32417480-32417480 on Build GRCh38
rs774600969 Chr.22: 32417065-32417065 on Build GRCh38
rs370915663 Chr.22: 32417143-32417143 on Build GRCh38
rs948448503 Chr.22: 32417250-32417250 on Build GRCh38
rs757942681 Chr.22: 32417132-32417132 on Build GRCh38
rs781602942 Chr.22: 32417047-32417047 on Build GRCh38
rs377322949 Chr.22: 32417485-32417485 on Build GRCh38
rs779888925 Chr.22: 32417190-32417192 on Build GRCh38
rs749436477 Chr.22: 32417183-32417183 on Build GRCh38
rs552103582 Chr.22: 32417226-32417255 on Build GRCh38
rs774246595 Chr.22: 32417185-32417185 on Build GRCh38
rs908235040 Chr.22: 32417370-32417370 on Build GRCh38
rs919119891 Chr.22: 32417190-32417190 on Build GRCh38
rs906861191 Chr.22: 32417264-32417264 on Build GRCh38
rs769388143 Chr.22: 32417190-32417193 on Build GRCh38
rs200041707 Chr.22: 32417123-32417123 on Build GRCh38
rs5844996 Chr.22: 32417190-32417190 on Build GRCh38
rs952470856 Chr.22: 32417353-32417353 on Build GRCh38
rs569596712 Chr.22: 32417242-32417242 on Build GRCh38
rs983794432 Chr.22: 32417365-32417365 on Build GRCh38
rs1000232617 Chr.22: 32417169-32417169 on Build GRCh38
rs754526871 Chr.22: 32417118-32417118 on Build GRCh38
rs796820256 Chr.22: 32417452-32417453 on Build GRCh38
rs780934946 Chr.22: 32417190-32417191 on Build GRCh38
rs908182982 Chr.22: 32417024-32417024 on Build GRCh38
rs970989233 Chr.22: 32417288-32417288 on Build GRCh38
rs759278256 Chr.22: 32417191-32417192 on Build GRCh38
rs878930534 Chr.22: 32417191-32417191 on Build GRCh38
rs538261846 Chr.22: 32417261-32417261 on Build GRCh38
rs748431624 Chr.22: 32417186-32417186 on Build GRCh38
rs78986210 Chr.22: 32417235-32417235 on Build GRCh38
rs537708802 Chr.22: 32417466-32417466 on Build GRCh38
rs925888646 Chr.22: 32417212-32417212 on Build GRCh38
rs532410782 Chr.22: 32416972-32416972 on Build GRCh38
rs368778265 Chr.22: 32417061-32417061 on Build GRCh38
rs1019649680 Chr.22: 32417439-32417439 on Build GRCh38
rs713760 Chr.22: 32417352-32417352 on Build GRCh38
rs1049320769 Chr.22: 32417243-32417243 on Build GRCh38
rs770249117 Chr.22: 32417056-32417056 on Build GRCh38
rs529974562 Chr.22: 32417168-32417168 on Build GRCh38
rs201536043 Chr.22: 32417346-32417346 on Build GRCh38
rs761560439 Chr.22: 32417079-32417079 on Build GRCh38
rs546232238 Chr.22: 32417018-32417018 on Build GRCh38
rs752262567 Chr.22: 32417192-32417192 on Build GRCh38
rs760691059 Chr.22: 32417095-32417095 on Build GRCh38
rs983743530 Chr.22: 32417002-32417002 on Build GRCh38
rs547648214 Chr.22: 32417100-32417100 on Build GRCh38
rs985386925 Chr.22: 32417207-32417207 on Build GRCh38
rs1007361174 Chr.22: 32417438-32417438 on Build GRCh38
rs752375648 Chr.22: 32417130-32417130 on Build GRCh38
rs770677745 Chr.22: 32417413-32417413 on Build GRCh38
rs980626486 Chr.22: 32417351-32417351 on Build GRCh38
rs868797467 Chr.22: 32416996-32416996 on Build GRCh38

View Full Product Details