Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 24175344-24175940 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM228A
Entrez Gene ID
653140
Gene Name
family with sequence similarity 228 member A
Gene Aliases
C2orf84
UniGene
Hs.741736
Gene Chromosome Location
Chr.2: 24175043-24191698 on Build GRCh38
Transcripts
NM_001040710.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5136613 c.93+10C>T p.? Chr.2: 24175583-24175583 on Build GRCh38
1407008 c.87A>G p.L29L Chr.2: 24175567-24175567 on Build GRCh38
7358523 c.41G>T p.R14M Chr.2: 24175521-24175521 on Build GRCh38
5150501 c.5C>T p.A2V Chr.2: 24175485-24175485 on Build GRCh38
3735188 c.71C>T p.P24L Chr.2: 24175551-24175551 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372054514 Chr.2: 24175458-24175458 on Build GRCh38
rs746000882 Chr.2: 24175515-24175515 on Build GRCh38
rs149710261 Chr.2: 24175509-24175509 on Build GRCh38
rs1048753789 Chr.2: 24175835-24175835 on Build GRCh38
rs747128968 Chr.2: 24175563-24175563 on Build GRCh38
rs375770165 Chr.2: 24175529-24175529 on Build GRCh38
rs187938077 Chr.2: 24175693-24175693 on Build GRCh38
rs1022516682 Chr.2: 24175489-24175489 on Build GRCh38
rs774749261 Chr.2: 24175529-24175533 on Build GRCh38
rs761169252 Chr.2: 24175441-24175441 on Build GRCh38
rs994864575 Chr.2: 24175398-24175398 on Build GRCh38
rs116450336 Chr.2: 24175585-24175585 on Build GRCh38
rs1009595837 Chr.2: 24175762-24175763 on Build GRCh38
rs1022718273 Chr.2: 24175763-24175763 on Build GRCh38
rs770475618 Chr.2: 24175559-24175559 on Build GRCh38
rs755275962 Chr.2: 24175621-24175621 on Build GRCh38
rs956933452 Chr.2: 24175467-24175467 on Build GRCh38
rs919036773 Chr.2: 24175813-24175813 on Build GRCh38
rs991938837 Chr.2: 24175870-24175870 on Build GRCh38
rs758405362 Chr.2: 24175524-24175524 on Build GRCh38
rs967985627 Chr.2: 24175767-24175767 on Build GRCh38
rs372838900 Chr.2: 24175523-24175523 on Build GRCh38
rs753706949 Chr.2: 24175610-24175610 on Build GRCh38
rs1029451022 Chr.2: 24175407-24175407 on Build GRCh38
rs759614584 Chr.2: 24175610-24175610 on Build GRCh38
rs960720455 Chr.2: 24175849-24175849 on Build GRCh38
rs752422837 Chr.2: 24175503-24175503 on Build GRCh38
rs543111385 Chr.2: 24175557-24175557 on Build GRCh38
rs766092933 Chr.2: 24175474-24175474 on Build GRCh38
rs968337785 Chr.2: 24175801-24175801 on Build GRCh38
rs754324936 Chr.2: 24175726-24175726 on Build GRCh38
rs908885768 Chr.2: 24175857-24175857 on Build GRCh38
rs1005746698 Chr.2: 24175391-24175391 on Build GRCh38
rs762344950 Chr.2: 24175472-24175472 on Build GRCh38
rs910443656 Chr.2: 24175795-24175795 on Build GRCh38
rs897540574 Chr.2: 24175609-24175610 on Build GRCh38
rs764160794 Chr.2: 24175601-24175601 on Build GRCh38
rs765532697 Chr.2: 24175809-24175809 on Build GRCh38
rs747210211 Chr.2: 24175550-24175550 on Build GRCh38
rs1045360980 Chr.2: 24175880-24175880 on Build GRCh38
rs183755517 Chr.2: 24175636-24175636 on Build GRCh38
rs755873796 Chr.2: 24175589-24175589 on Build GRCh38
rs577851822 Chr.2: 24175408-24175408 on Build GRCh38
rs368132241 Chr.2: 24175451-24175451 on Build GRCh38
rs772387377 Chr.2: 24175435-24175435 on Build GRCh38
rs1033392340 Chr.2: 24175843-24175843 on Build GRCh38
rs774035546 Chr.2: 24175774-24175774 on Build GRCh38
rs2042467 Chr.2: 24175412-24175412 on Build GRCh38
rs188029883 Chr.2: 24175375-24175375 on Build GRCh38
rs368326733 Chr.2: 24175487-24175487 on Build GRCh38
rs2042468 Chr.2: 24175577-24175577 on Build GRCh38
rs759921877 Chr.2: 24175778-24175778 on Build GRCh38
rs368941941 Chr.2: 24175568-24175568 on Build GRCh38
rs973908279 Chr.2: 24175909-24175909 on Build GRCh38
rs527821712 Chr.2: 24175553-24175553 on Build GRCh38
rs759201569 Chr.2: 24175829-24175829 on Build GRCh38
rs780005959 Chr.2: 24175615-24175615 on Build GRCh38
rs765789805 Chr.2: 24175569-24175569 on Build GRCh38
rs767312501 Chr.2: 24175488-24175488 on Build GRCh38
rs1019057020 Chr.2: 24175739-24175739 on Build GRCh38
rs780382145 Chr.2: 24175546-24175546 on Build GRCh38
rs193234740 Chr.2: 24175625-24175625 on Build GRCh38
rs888743836 Chr.2: 24175650-24175650 on Build GRCh38
rs892606355 Chr.2: 24175764-24175764 on Build GRCh38
rs567154124 Chr.2: 24175890-24175890 on Build GRCh38
rs75121759 Chr.2: 24175586-24175586 on Build GRCh38
rs376569182 Chr.2: 24175461-24175461 on Build GRCh38
rs1003691046 Chr.2: 24175728-24175728 on Build GRCh38
rs757162005 Chr.2: 24175611-24175611 on Build GRCh38
rs980838248 Chr.2: 24175842-24175842 on Build GRCh38
rs898412309 Chr.2: 24175630-24175630 on Build GRCh38
rs930606262 Chr.2: 24175729-24175740 on Build GRCh38
rs772804635 Chr.2: 24175468-24175468 on Build GRCh38
rs986011191 Chr.2: 24175773-24175773 on Build GRCh38
rs145570150 Chr.2: 24175898-24175898 on Build GRCh38
rs775546054 Chr.2: 24175438-24175438 on Build GRCh38
rs976306466 Chr.2: 24175807-24175807 on Build GRCh38
rs759193008 Chr.2: 24175583-24175583 on Build GRCh38
rs908400850 Chr.2: 24175609-24175609 on Build GRCh38
rs769129979 Chr.2: 24175443-24175443 on Build GRCh38
rs948330620 Chr.2: 24175879-24175879 on Build GRCh38
rs763965654 Chr.2: 24175615-24175616 on Build GRCh38
rs1038267405 Chr.2: 24175622-24175622 on Build GRCh38
rs929044915 Chr.2: 24175817-24175817 on Build GRCh38
rs771086625 Chr.2: 24175435-24175435 on Build GRCh38
rs751163749 Chr.2: 24175482-24175482 on Build GRCh38
rs553024075 Chr.2: 24175704-24175704 on Build GRCh38
rs556376130 Chr.2: 24175394-24175394 on Build GRCh38
rs1009709374 Chr.2: 24175777-24175777 on Build GRCh38
rs765801249 Chr.2: 24175464-24175464 on Build GRCh38
rs375306135 Chr.2: 24175430-24175430 on Build GRCh38
rs533656546 Chr.2: 24175796-24175796 on Build GRCh38
rs1003745336 Chr.2: 24175904-24175904 on Build GRCh38
rs555411830 Chr.2: 24175799-24175799 on Build GRCh38
rs756074297 Chr.2: 24175507-24175507 on Build GRCh38

View Full Product Details