Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 17772304-17772837 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GEN1
Entrez Gene ID
348654
Gene Name
GEN1, Holliday junction 5' flap endonuclease
Gene Aliases
Gen
UniGene
Hs.467793
Gene Chromosome Location
Chr.2: 17753687-17785920 on Build GRCh38
Transcripts
NM_001130009.2 NM_182625.4 XM_005262613.4 XM_006712005.3 XM_011532820.2 XM_011532821.2 XM_011532822.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6089459 c.827A>G p.N276S Chr.2: 17772658-17772658 on Build GRCh38
4940869 c.953A>G p.K318R Chr.2: 17772784-17772784 on Build GRCh38
1012676 c.829G>T p.G277* Chr.2: 17772660-17772660 on Build GRCh38
717942 c.850G>A p.D284N Chr.2: 17772681-17772681 on Build GRCh38
6214360 c.823C>A p.R275S Chr.2: 17772654-17772654 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752560981 Chr.2: 17772718-17772718 on Build GRCh38
rs1038650657 Chr.2: 17772794-17772794 on Build GRCh38
rs546530701 Chr.2: 17772369-17772369 on Build GRCh38
rs770385553 Chr.2: 17772767-17772767 on Build GRCh38
rs773872904 Chr.2: 17772787-17772787 on Build GRCh38
rs113050456 Chr.2: 17772552-17772552 on Build GRCh38
rs996483723 Chr.2: 17772541-17772541 on Build GRCh38
rs553720738 Chr.2: 17772616-17772616 on Build GRCh38
rs780972803 Chr.2: 17772614-17772614 on Build GRCh38
rs962193584 Chr.2: 17772642-17772642 on Build GRCh38
rs991939369 Chr.2: 17772365-17772365 on Build GRCh38
rs201202527 Chr.2: 17772689-17772689 on Build GRCh38
rs148607792 Chr.2: 17772736-17772736 on Build GRCh38
rs202192107 Chr.2: 17772735-17772735 on Build GRCh38
rs770440644 Chr.2: 17772659-17772659 on Build GRCh38
rs777688680 Chr.2: 17772721-17772721 on Build GRCh38
rs561031093 Chr.2: 17772352-17772352 on Build GRCh38
rs750088839 Chr.2: 17772590-17772590 on Build GRCh38
rs1042842270 Chr.2: 17772387-17772387 on Build GRCh38
rs369259669 Chr.2: 17772611-17772611 on Build GRCh38
rs1037916103 Chr.2: 17772425-17772425 on Build GRCh38
rs762442199 Chr.2: 17772658-17772658 on Build GRCh38
rs368769308 Chr.2: 17772467-17772467 on Build GRCh38
rs759522575 Chr.2: 17772443-17772443 on Build GRCh38
rs770459600 Chr.2: 17772413-17772413 on Build GRCh38
rs1039267244 Chr.2: 17772562-17772562 on Build GRCh38
rs894020819 Chr.2: 17772576-17772576 on Build GRCh38
rs139945353 Chr.2: 17772634-17772634 on Build GRCh38
rs780226254 Chr.2: 17772729-17772729 on Build GRCh38
rs762637631 Chr.2: 17772592-17772592 on Build GRCh38
rs748788774 Chr.2: 17772784-17772784 on Build GRCh38
rs774216696 Chr.2: 17772419-17772419 on Build GRCh38
rs927940872 Chr.2: 17772380-17772380 on Build GRCh38
rs775286168 Chr.2: 17772805-17772805 on Build GRCh38
rs773878519 Chr.2: 17772780-17772780 on Build GRCh38
rs202239270 Chr.2: 17772655-17772655 on Build GRCh38
rs959369092 Chr.2: 17772652-17772652 on Build GRCh38
rs570008382 Chr.2: 17772638-17772638 on Build GRCh38
rs529668990 Chr.2: 17772356-17772356 on Build GRCh38
rs963262901 Chr.2: 17772665-17772665 on Build GRCh38
rs767213865 Chr.2: 17772524-17772524 on Build GRCh38
rs915791830 Chr.2: 17772377-17772378 on Build GRCh38
rs371772153 Chr.2: 17772725-17772725 on Build GRCh38
rs756019387 Chr.2: 17772720-17772720 on Build GRCh38
rs868330956 Chr.2: 17772636-17772646 on Build GRCh38
rs928736223 Chr.2: 17772336-17772336 on Build GRCh38
rs2052745 Chr.2: 17772381-17772381 on Build GRCh38
rs762584233 Chr.2: 17772667-17772667 on Build GRCh38
rs533161635 Chr.2: 17772487-17772487 on Build GRCh38
rs764642571 Chr.2: 17772763-17772780 on Build GRCh38
rs146276503 Chr.2: 17772654-17772654 on Build GRCh38
rs973291253 Chr.2: 17772700-17772700 on Build GRCh38
rs760414260 Chr.2: 17772807-17772807 on Build GRCh38
rs369081159 Chr.2: 17772439-17772439 on Build GRCh38
rs1007797322 Chr.2: 17772545-17772545 on Build GRCh38
rs759224196 Chr.2: 17772704-17772704 on Build GRCh38
rs919029058 Chr.2: 17772464-17772464 on Build GRCh38
rs775885155 Chr.2: 17772700-17772705 on Build GRCh38
rs779020468 Chr.2: 17772628-17772628 on Build GRCh38
rs116651705 Chr.2: 17772583-17772583 on Build GRCh38
rs373132364 Chr.2: 17772630-17772630 on Build GRCh38
rs300175 Chr.2: 17772760-17772760 on Build GRCh38
rs757283045 Chr.2: 17772624-17772624 on Build GRCh38
rs772347003 Chr.2: 17772612-17772613 on Build GRCh38
rs869312794 Chr.2: 17772788-17772788 on Build GRCh38
rs960062604 Chr.2: 17772342-17772342 on Build GRCh38
rs370143023 Chr.2: 17772785-17772785 on Build GRCh38
rs911606054 Chr.2: 17772750-17772750 on Build GRCh38
rs766134831 Chr.2: 17772595-17772595 on Build GRCh38
rs894073402 Chr.2: 17772802-17772802 on Build GRCh38
rs915797845 Chr.2: 17772402-17772402 on Build GRCh38
rs900955239 Chr.2: 17772493-17772493 on Build GRCh38
rs915911691 Chr.2: 17772764-17772764 on Build GRCh38
rs767087056 Chr.2: 17772796-17772796 on Build GRCh38
rs552485962 Chr.2: 17772480-17772480 on Build GRCh38
rs556414723 Chr.2: 17772672-17772672 on Build GRCh38
rs761094417 Chr.2: 17772742-17772753 on Build GRCh38
rs149884551 Chr.2: 17772706-17772706 on Build GRCh38
rs759293604 Chr.2: 17772793-17772793 on Build GRCh38
rs374326909 Chr.2: 17772724-17772724 on Build GRCh38
rs751360597 Chr.2: 17772610-17772610 on Build GRCh38
rs772781859 Chr.2: 17772663-17772663 on Build GRCh38
rs747287864 Chr.2: 17772648-17772648 on Build GRCh38
rs991398286 Chr.2: 17772392-17772392 on Build GRCh38
rs548433334 Chr.2: 17772519-17772519 on Build GRCh38
rs781563407 Chr.2: 17772749-17772749 on Build GRCh38
rs531036922 Chr.2: 17772497-17772497 on Build GRCh38

View Full Product Details