Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 53292276-53292863 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCP1A
Entrez Gene ID
55802
Gene Name
decapping mRNA 1A
Gene Aliases
HSA275986, Nbla00360, SMAD4IP1, SMIF
UniGene
Hs.476353
Gene Chromosome Location
Chr.3: 53283423-53347627 on Build GRCh38
Transcripts
NM_001290204.1 NM_001290205.1 NM_001290206.1 NM_001290207.1 NM_018403.6

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
460752 c.808C>G p.Q270E Chr.3: 53292644-53292644 on Build GRCh38
1203148 c.836C>T p.A279V Chr.3: 53292616-53292616 on Build GRCh38
6314815 c.804C>A p.A268A Chr.3: 53292648-53292648 on Build GRCh38
4119285 c.735G>A p.L245L Chr.3: 53292717-53292717 on Build GRCh38
4676571 c.1140C>T p.N380N Chr.3: 53292312-53292312 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781827654 Chr.3: 53292464-53292464 on Build GRCh38
rs782478458 Chr.3: 53292505-53292505 on Build GRCh38
rs782594632 Chr.3: 53292506-53292506 on Build GRCh38
rs782088982 Chr.3: 53292530-53292530 on Build GRCh38
rs782784741 Chr.3: 53292835-53292835 on Build GRCh38
rs913725499 Chr.3: 53292557-53292557 on Build GRCh38
rs782393066 Chr.3: 53292795-53292795 on Build GRCh38
rs782692637 Chr.3: 53292428-53292428 on Build GRCh38
rs781966543 Chr.3: 53292688-53292688 on Build GRCh38
rs782157688 Chr.3: 53292491-53292491 on Build GRCh38
rs782544282 Chr.3: 53292497-53292497 on Build GRCh38
rs546312435 Chr.3: 53292409-53292409 on Build GRCh38
rs571745219 Chr.3: 53292558-53292558 on Build GRCh38
rs367934700 Chr.3: 53292614-53292614 on Build GRCh38
rs35988197 Chr.3: 53292562-53292562 on Build GRCh38
rs545563039 Chr.3: 53292758-53292758 on Build GRCh38
rs781937557 Chr.3: 53292311-53292311 on Build GRCh38
rs781832945 Chr.3: 53292366-53292366 on Build GRCh38
rs781818197 Chr.3: 53292603-53292603 on Build GRCh38
rs199632820 Chr.3: 53292437-53292437 on Build GRCh38
rs375268167 Chr.3: 53292708-53292708 on Build GRCh38
rs532893620 Chr.3: 53292401-53292401 on Build GRCh38
rs551318997 Chr.3: 53292514-53292514 on Build GRCh38
rs960146118 Chr.3: 53292695-53292695 on Build GRCh38
rs782226733 Chr.3: 53292813-53292813 on Build GRCh38
rs782176345 Chr.3: 53292642-53292642 on Build GRCh38
rs782414422 Chr.3: 53292340-53292340 on Build GRCh38
rs782134348 Chr.3: 53292353-53292353 on Build GRCh38
rs782119551 Chr.3: 53292459-53292459 on Build GRCh38
rs782700313 Chr.3: 53292699-53292699 on Build GRCh38
rs782233128 Chr.3: 53292309-53292309 on Build GRCh38
rs527911664 Chr.3: 53292490-53292490 on Build GRCh38
rs988906652 Chr.3: 53292799-53292799 on Build GRCh38
rs781859914 Chr.3: 53292712-53292712 on Build GRCh38
rs782675604 Chr.3: 53292759-53292759 on Build GRCh38
rs782550767 Chr.3: 53292390-53292390 on Build GRCh38
rs782284886 Chr.3: 53292301-53292301 on Build GRCh38
rs782724134 Chr.3: 53292382-53292382 on Build GRCh38
rs34229192 Chr.3: 53292458-53292458 on Build GRCh38
rs201480018 Chr.3: 53292389-53292389 on Build GRCh38
rs374845014 Chr.3: 53292659-53292659 on Build GRCh38
rs373006464 Chr.3: 53292798-53292798 on Build GRCh38
rs375827844 Chr.3: 53292453-53292453 on Build GRCh38
rs368073286 Chr.3: 53292306-53292306 on Build GRCh38
rs191188886 Chr.3: 53292310-53292310 on Build GRCh38
rs890845801 Chr.3: 53292612-53292612 on Build GRCh38
rs782161271 Chr.3: 53292709-53292709 on Build GRCh38
rs782532915 Chr.3: 53292613-53292613 on Build GRCh38
rs375815016 Chr.3: 53292544-53292544 on Build GRCh38
rs892336556 Chr.3: 53292379-53292379 on Build GRCh38
rs782789189 Chr.3: 53292710-53292710 on Build GRCh38
rs376692040 Chr.3: 53292312-53292312 on Build GRCh38
rs781923882 Chr.3: 53292532-53292532 on Build GRCh38
rs186062414 Chr.3: 53292650-53292650 on Build GRCh38
rs369794168 Chr.3: 53292412-53292412 on Build GRCh38
rs782717150 Chr.3: 53292531-53292531 on Build GRCh38
rs371848238 Chr.3: 53292639-53292639 on Build GRCh38
rs34366563 Chr.3: 53292504-53292504 on Build GRCh38
rs782342251 Chr.3: 53292823-53292823 on Build GRCh38
rs782704435 Chr.3: 53292470-53292470 on Build GRCh38
rs781937534 Chr.3: 53292465-53292465 on Build GRCh38
rs782017473 Chr.3: 53292346-53292346 on Build GRCh38
rs373193756 Chr.3: 53292423-53292423 on Build GRCh38
rs372198236 Chr.3: 53292680-53292680 on Build GRCh38
rs782080770 Chr.3: 53292696-53292696 on Build GRCh38
rs569103562 Chr.3: 53292498-53292498 on Build GRCh38
rs781900316 Chr.3: 53292595-53292595 on Build GRCh38
rs781819550 Chr.3: 53292583-53292583 on Build GRCh38
rs782760651 Chr.3: 53292364-53292364 on Build GRCh38
rs371261804 Chr.3: 53292402-53292402 on Build GRCh38
rs782494181 Chr.3: 53292716-53292716 on Build GRCh38
rs782038259 Chr.3: 53292837-53292837 on Build GRCh38
rs966542734 Chr.3: 53292836-53292836 on Build GRCh38
rs782344114 Chr.3: 53292632-53292632 on Build GRCh38
rs782460066 Chr.3: 53292585-53292585 on Build GRCh38
rs536298652 Chr.3: 53292643-53292643 on Build GRCh38
rs782271554 Chr.3: 53292429-53292429 on Build GRCh38
rs782617481 Chr.3: 53292717-53292717 on Build GRCh38
rs782278076 Chr.3: 53292769-53292769 on Build GRCh38
rs1037645346 Chr.3: 53292619-53292619 on Build GRCh38
rs369538805 Chr.3: 53292736-53292736 on Build GRCh38
rs913746501 Chr.3: 53292805-53292805 on Build GRCh38
rs782753531 Chr.3: 53292460-53292460 on Build GRCh38
rs267599901 Chr.3: 53292820-53292820 on Build GRCh38
rs781910025 Chr.3: 53292487-53292487 on Build GRCh38
rs200543667 Chr.3: 53292307-53292307 on Build GRCh38
rs1049509358 Chr.3: 53292602-53292602 on Build GRCh38
rs200307108 Chr.3: 53292537-53292537 on Build GRCh38
rs182281123 Chr.3: 53292454-53292454 on Build GRCh38
rs782234532 Chr.3: 53292616-53292616 on Build GRCh38
rs782076499 Chr.3: 53292372-53292372 on Build GRCh38
rs1035743671 Chr.3: 53292693-53292693 on Build GRCh38
rs781975316 Chr.3: 53292527-53292527 on Build GRCh38
rs377530427 Chr.3: 53292513-53292513 on Build GRCh38
rs924957141 Chr.3: 53292580-53292580 on Build GRCh38
rs375123165 Chr.3: 53292435-53292435 on Build GRCh38
rs782045307 Chr.3: 53292445-53292445 on Build GRCh38
rs376753632 Chr.3: 53292408-53292408 on Build GRCh38
rs782563087 Chr.3: 53292593-53292593 on Build GRCh38
rs782390302 Chr.3: 53292431-53292431 on Build GRCh38
rs368916174 Chr.3: 53292678-53292678 on Build GRCh38
rs35983535 Chr.3: 53292579-53292579 on Build GRCh38
rs200464638 Chr.3: 53292578-53292578 on Build GRCh38

View Full Product Details