Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 27890092-27890632 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRE
Entrez Gene ID
9577
Gene Name
brain and reproductive organ-expressed (TNFRSF1A modulator)
Gene Aliases
BRCC4, BRCC45
UniGene
Hs.258314
Gene Chromosome Location
Chr.2: 27890615-28338901 on Build GRCh38
Transcripts
NM_001261840.1 NM_004899.4 NM_199191.2 NM_199192.2 NM_199193.2 NM_199194.2 NR_028308.1 NM_001287580.1 NM_022128.2

Gene Symbol
BRE-AS1
Entrez Gene ID
100302650
Gene Name
BRE antisense RNA 1
Gene Aliases
-
UniGene
Hs.729719
Gene Chromosome Location
Chr.2: 27889456-27891114 on Build GRCh38
Transcripts
NM_001261840.1 NM_004899.4 NM_199191.2 NM_199192.2 NM_199193.2 NM_199194.2 NR_028308.1 NM_001287580.1 NM_022128.2

Gene Symbol
RBKS
Entrez Gene ID
64080
Gene Name
ribokinase
Gene Aliases
RBSK, RK
UniGene
Hs.11916
Gene Chromosome Location
Chr.2: 27781364-27890396 on Build GRCh38
Transcripts
NM_001261840.1 NM_004899.4 NM_199191.2 NM_199192.2 NM_199193.2 NM_199194.2 NR_028308.1 NM_001287580.1 NM_022128.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5396831 c.81C>T p.D27D Chr.2: 27890265-27890265 on Build GRCh38
5622844 c.38A>G p.E13G Chr.2: 27890308-27890308 on Build GRCh38
5042415 c.14G>A p.G5E Chr.2: 27890332-27890332 on Build GRCh38
7132989 c.82C>T p.L28L Chr.2: 27890264-27890264 on Build GRCh38
3580750 c.60G>A p.V20V Chr.2: 27890286-27890286 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs148593620 Chr.2: 27890328-27890328 on Build GRCh38
rs559551018 Chr.2: 27890459-27890459 on Build GRCh38
rs975126113 Chr.2: 27890435-27890435 on Build GRCh38
rs746461331 Chr.2: 27890230-27890230 on Build GRCh38
rs747173924 Chr.2: 27890495-27890495 on Build GRCh38
rs762050106 Chr.2: 27890271-27890271 on Build GRCh38
rs75602512 Chr.2: 27890395-27890395 on Build GRCh38
rs567504415 Chr.2: 27890268-27890268 on Build GRCh38
rs1009188924 Chr.2: 27890417-27890417 on Build GRCh38
rs972111302 Chr.2: 27890452-27890452 on Build GRCh38
rs756166439 Chr.2: 27890234-27890237 on Build GRCh38
rs140995009 Chr.2: 27890289-27890290 on Build GRCh38
rs957832741 Chr.2: 27890440-27890440 on Build GRCh38
rs781359981 Chr.2: 27890506-27890507 on Build GRCh38
rs764161169 Chr.2: 27890207-27890207 on Build GRCh38
rs777387047 Chr.2: 27890384-27890384 on Build GRCh38
rs927904283 Chr.2: 27890553-27890553 on Build GRCh38
rs985210953 Chr.2: 27890496-27890496 on Build GRCh38
rs138851838 Chr.2: 27890158-27890158 on Build GRCh38
rs959210944 Chr.2: 27890178-27890178 on Build GRCh38
rs764299524 Chr.2: 27890262-27890262 on Build GRCh38
rs114146461 Chr.2: 27890466-27890466 on Build GRCh38
rs201634958 Chr.2: 27890251-27890251 on Build GRCh38
rs773544042 Chr.2: 27890379-27890379 on Build GRCh38
rs538064099 Chr.2: 27890567-27890567 on Build GRCh38
rs543611264 Chr.2: 27890373-27890373 on Build GRCh38
rs973615092 Chr.2: 27890189-27890189 on Build GRCh38
rs992413569 Chr.2: 27890184-27890184 on Build GRCh38
rs976213732 Chr.2: 27890536-27890536 on Build GRCh38
rs549921090 Chr.2: 27890592-27890592 on Build GRCh38
rs540639332 Chr.2: 27890394-27890394 on Build GRCh38
rs770496078 Chr.2: 27890360-27890360 on Build GRCh38
rs775968520 Chr.2: 27890332-27890332 on Build GRCh38
rs369002158 Chr.2: 27890227-27890227 on Build GRCh38
rs760727351 Chr.2: 27890257-27890257 on Build GRCh38
rs571058235 Chr.2: 27890593-27890593 on Build GRCh38
rs780350201 Chr.2: 27890397-27890397 on Build GRCh38
rs1013403664 Chr.2: 27890396-27890396 on Build GRCh38
rs771177489 Chr.2: 27890243-27890243 on Build GRCh38
rs1047836648 Chr.2: 27890578-27890578 on Build GRCh38
rs758539982 Chr.2: 27890206-27890206 on Build GRCh38
rs886490538 Chr.2: 27890581-27890581 on Build GRCh38
rs551447527 Chr.2: 27890164-27890164 on Build GRCh38
rs761807527 Chr.2: 27890356-27890356 on Build GRCh38
rs778821130 Chr.2: 27890223-27890223 on Build GRCh38
rs1024851311 Chr.2: 27890412-27890412 on Build GRCh38
rs994972531 Chr.2: 27890571-27890571 on Build GRCh38
rs780706666 Chr.2: 27890242-27890242 on Build GRCh38
rs1019217645 Chr.2: 27890423-27890423 on Build GRCh38
rs184123655 Chr.2: 27890197-27890197 on Build GRCh38
rs1046174635 Chr.2: 27890538-27890538 on Build GRCh38
rs191606675 Chr.2: 27890167-27890167 on Build GRCh38
rs776816973 Chr.2: 27890570-27890570 on Build GRCh38
rs945161917 Chr.2: 27890335-27890335 on Build GRCh38
rs747552945 Chr.2: 27890334-27890334 on Build GRCh38
rs190142506 Chr.2: 27890293-27890293 on Build GRCh38
rs901583294 Chr.2: 27890333-27890333 on Build GRCh38
rs780801527 Chr.2: 27890301-27890301 on Build GRCh38
rs910793515 Chr.2: 27890497-27890497 on Build GRCh38
rs374249982 Chr.2: 27890347-27890347 on Build GRCh38
rs137874195 Chr.2: 27890280-27890280 on Build GRCh38
rs765442643 Chr.2: 27890357-27890357 on Build GRCh38
rs776735951 Chr.2: 27890352-27890352 on Build GRCh38
rs572347197 Chr.2: 27890339-27890339 on Build GRCh38
rs113651030 Chr.2: 27890290-27890290 on Build GRCh38
rs561164535 Chr.2: 27890371-27890371 on Build GRCh38
rs1020616896 Chr.2: 27890137-27890137 on Build GRCh38
rs758375653 Chr.2: 27890228-27890228 on Build GRCh38
rs774727775 Chr.2: 27890245-27890245 on Build GRCh38
rs967930469 Chr.2: 27890146-27890146 on Build GRCh38
rs748872380 Chr.2: 27890260-27890260 on Build GRCh38
rs920913783 Chr.2: 27890439-27890439 on Build GRCh38
rs577283069 Chr.2: 27890286-27890286 on Build GRCh38
rs560275212 Chr.2: 27890419-27890419 on Build GRCh38
rs780127805 Chr.2: 27890288-27890288 on Build GRCh38
rs777835683 Chr.2: 27890238-27890241 on Build GRCh38
rs775986725 Chr.2: 27890256-27890256 on Build GRCh38
rs774159732 Chr.2: 27890394-27890394 on Build GRCh38
rs746169402 Chr.2: 27890327-27890327 on Build GRCh38
rs759925783 Chr.2: 27890246-27890246 on Build GRCh38
rs377174524 Chr.2: 27890233-27890233 on Build GRCh38
rs149354783 Chr.2: 27890372-27890372 on Build GRCh38
rs950860284 Chr.2: 27890413-27890413 on Build GRCh38
rs754143786 Chr.2: 27890210-27890210 on Build GRCh38
rs376141839 Chr.2: 27890362-27890362 on Build GRCh38
rs1042096542 Chr.2: 27890606-27890606 on Build GRCh38
rs528020619 Chr.2: 27890449-27890449 on Build GRCh38
rs747633256 Chr.2: 27890315-27890315 on Build GRCh38
rs574663262 Chr.2: 27890604-27890604 on Build GRCh38
rs996811507 Chr.2: 27890597-27890597 on Build GRCh38
rs756995099 Chr.2: 27890221-27890221 on Build GRCh38
rs372982880 Chr.2: 27890231-27890231 on Build GRCh38
rs780144481 Chr.2: 27890229-27890229 on Build GRCh38
rs771142173 Chr.2: 27890502-27890502 on Build GRCh38
rs562982045 Chr.2: 27890381-27890381 on Build GRCh38
rs537574219 Chr.2: 27890276-27890276 on Build GRCh38
rs997077066 Chr.2: 27890554-27890567 on Build GRCh38
rs370831016 Chr.2: 27890318-27890318 on Build GRCh38
rs138553386 Chr.2: 27890296-27890296 on Build GRCh38
rs777295403 Chr.2: 27890319-27890319 on Build GRCh38
rs144772878 Chr.2: 27890309-27890309 on Build GRCh38

View Full Product Details