Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 101269841-101270293 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNOT11
Entrez Gene ID
55571
Gene Name
CCR4-NOT transcription complex subunit 11
Gene Aliases
C2orf29, C40
UniGene
Hs.744080
Gene Chromosome Location
Chr.2: 101252883-101270316 on Build GRCh38
Transcripts
NM_017546.4

SNP Detail

SNP ID Chromosome Location
rs917865697 Chr.2: 101270155-101270155 on Build GRCh38
rs567671192 Chr.2: 101270196-101270196 on Build GRCh38
rs1018256119 Chr.2: 101270266-101270266 on Build GRCh38
rs560973625 Chr.2: 101269908-101269908 on Build GRCh38
rs536204792 Chr.2: 101270245-101270245 on Build GRCh38
rs540345956 Chr.2: 101269990-101269990 on Build GRCh38
rs930190649 Chr.2: 101270244-101270244 on Build GRCh38
rs968086957 Chr.2: 101270073-101270073 on Build GRCh38
rs994806535 Chr.2: 101269974-101269974 on Build GRCh38
rs368378746 Chr.2: 101270162-101270162 on Build GRCh38
rs1029939855 Chr.2: 101269992-101269992 on Build GRCh38
rs914886210 Chr.2: 101269887-101269887 on Build GRCh38
rs922307685 Chr.2: 101270254-101270254 on Build GRCh38
rs574547609 Chr.2: 101270108-101270108 on Build GRCh38
rs1036134529 Chr.2: 101270219-101270219 on Build GRCh38
rs899136791 Chr.2: 101270122-101270122 on Build GRCh38
rs1022468696 Chr.2: 101270230-101270230 on Build GRCh38
rs11545033 Chr.2: 101270214-101270214 on Build GRCh38
rs1012263045 Chr.2: 101270123-101270123 on Build GRCh38
rs1010624248 Chr.2: 101270201-101270201 on Build GRCh38
rs1018369313 Chr.2: 101270071-101270071 on Build GRCh38
rs553057963 Chr.2: 101270270-101270270 on Build GRCh38
rs906775408 Chr.2: 101270146-101270149 on Build GRCh38
rs955346744 Chr.2: 101270022-101270022 on Build GRCh38
rs1038908189 Chr.2: 101269961-101269961 on Build GRCh38
rs796491819 Chr.2: 101270076-101270077 on Build GRCh38
rs897333251 Chr.2: 101269964-101269964 on Build GRCh38
rs564362703 Chr.2: 101270077-101270079 on Build GRCh38
rs796455649 Chr.2: 101270135-101270135 on Build GRCh38
rs887565655 Chr.2: 101269954-101269954 on Build GRCh38
rs149754769 Chr.2: 101270161-101270161 on Build GRCh38
rs553202663 Chr.2: 101270065-101270065 on Build GRCh38
rs529624022 Chr.2: 101269953-101269953 on Build GRCh38
rs922523664 Chr.2: 101270006-101270006 on Build GRCh38
rs891423744 Chr.2: 101270250-101270250 on Build GRCh38
rs755212873 Chr.2: 101270231-101270231 on Build GRCh38
rs748998357 Chr.2: 101269975-101269976 on Build GRCh38
rs1005928201 Chr.2: 101269955-101269955 on Build GRCh38
rs184924911 Chr.2: 101270193-101270193 on Build GRCh38
rs1044783647 Chr.2: 101269926-101269926 on Build GRCh38
rs191629014 Chr.2: 101270074-101270074 on Build GRCh38
rs933950107 Chr.2: 101270047-101270047 on Build GRCh38
rs1038209491 Chr.2: 101270088-101270088 on Build GRCh38
rs1007150863 Chr.2: 101270258-101270258 on Build GRCh38
rs1031223992 Chr.2: 101270101-101270101 on Build GRCh38
rs963640715 Chr.2: 101270242-101270242 on Build GRCh38
rs766193557 Chr.2: 101269933-101269933 on Build GRCh38
rs12114 Chr.2: 101270141-101270141 on Build GRCh38

View Full Product Details