Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 196646085-196646598 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC150
Entrez Gene ID
284992
Gene Name
coiled-coil domain containing 150
Gene Aliases
-
UniGene
Hs.132519
Gene Chromosome Location
Chr.2: 196639632-196733668 on Build GRCh38
Transcripts
NM_001080539.1 XM_006712438.3 XM_011510985.2 XM_011510988.2 XM_017003861.1 XM_017003862.1 XM_017003863.1 XM_017003864.1 XM_017003865.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1738681 c.101A>G p.Q34R Chr.2: 196646429-196646429 on Build GRCh38
259962 c.73G>A p.A25T Chr.2: 196646401-196646401 on Build GRCh38
7209136 c.97C>G p.Q33E Chr.2: 196646425-196646425 on Build GRCh38
476700 c.66C>A p.N22K Chr.2: 196646394-196646394 on Build GRCh38
3838207 c.79G>A p.E27K Chr.2: 196646407-196646407 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753355962 Chr.2: 196646486-196646486 on Build GRCh38
rs897624916 Chr.2: 196646123-196646123 on Build GRCh38
rs748898120 Chr.2: 196646308-196646308 on Build GRCh38
rs747959200 Chr.2: 196646323-196646323 on Build GRCh38
rs537880984 Chr.2: 196646397-196646397 on Build GRCh38
rs531591279 Chr.2: 196646274-196646274 on Build GRCh38
rs907171991 Chr.2: 196646419-196646419 on Build GRCh38
rs745959864 Chr.2: 196646391-196646391 on Build GRCh38
rs1002930666 Chr.2: 196646434-196646434 on Build GRCh38
rs929153673 Chr.2: 196646370-196646370 on Build GRCh38
rs986079054 Chr.2: 196646567-196646567 on Build GRCh38
rs755650637 Chr.2: 196646359-196646359 on Build GRCh38
rs549312035 Chr.2: 196646353-196646353 on Build GRCh38
rs747131196 Chr.2: 196646333-196646333 on Build GRCh38
rs376522181 Chr.2: 196646534-196646534 on Build GRCh38
rs982148064 Chr.2: 196646239-196646239 on Build GRCh38
rs542606155 Chr.2: 196646181-196646181 on Build GRCh38
rs914462143 Chr.2: 196646280-196646280 on Build GRCh38
rs754805245 Chr.2: 196646311-196646311 on Build GRCh38
rs557798336 Chr.2: 196646436-196646436 on Build GRCh38
rs972048411 Chr.2: 196646207-196646207 on Build GRCh38
rs758135093 Chr.2: 196646504-196646504 on Build GRCh38
rs1041380863 Chr.2: 196646300-196646300 on Build GRCh38
rs774841862 Chr.2: 196646447-196646447 on Build GRCh38
rs142273672 Chr.2: 196646210-196646210 on Build GRCh38
rs759051580 Chr.2: 196646487-196646487 on Build GRCh38
rs779705618 Chr.2: 196646304-196646304 on Build GRCh38
rs902895968 Chr.2: 196646147-196646147 on Build GRCh38
rs1008251973 Chr.2: 196646170-196646170 on Build GRCh38
rs781044967 Chr.2: 196646182-196646182 on Build GRCh38
rs767998042 Chr.2: 196646459-196646459 on Build GRCh38
rs1016515882 Chr.2: 196646179-196646179 on Build GRCh38
rs1055697284 Chr.2: 196646130-196646130 on Build GRCh38
rs776913567 Chr.2: 196646385-196646385 on Build GRCh38
rs373391754 Chr.2: 196646392-196646392 on Build GRCh38
rs183194600 Chr.2: 196646159-196646159 on Build GRCh38
rs1027545729 Chr.2: 196646149-196646149 on Build GRCh38
rs759623150 Chr.2: 196646339-196646339 on Build GRCh38
rs551479554 Chr.2: 196646249-196646249 on Build GRCh38
rs201237992 Chr.2: 196646381-196646381 on Build GRCh38
rs749975484 Chr.2: 196646293-196646293 on Build GRCh38
rs369159032 Chr.2: 196646488-196646488 on Build GRCh38
rs777655736 Chr.2: 196646508-196646508 on Build GRCh38
rs568886595 Chr.2: 196646394-196646394 on Build GRCh38
rs780172294 Chr.2: 196646546-196646546 on Build GRCh38
rs758188490 Chr.2: 196646377-196646377 on Build GRCh38
rs902948254 Chr.2: 196646310-196646310 on Build GRCh38
rs564235121 Chr.2: 196646119-196646119 on Build GRCh38
rs373298600 Chr.2: 196646512-196646512 on Build GRCh38
rs369386644 Chr.2: 196646373-196646373 on Build GRCh38
rs775760234 Chr.2: 196646395-196646395 on Build GRCh38
rs1022973901 Chr.2: 196646416-196646416 on Build GRCh38
rs746702389 Chr.2: 196646309-196646310 on Build GRCh38
rs991299838 Chr.2: 196646559-196646559 on Build GRCh38
rs1023385754 Chr.2: 196646164-196646164 on Build GRCh38
rs887679591 Chr.2: 196646150-196646150 on Build GRCh38
rs765806725 Chr.2: 196646360-196646360 on Build GRCh38
rs947045673 Chr.2: 196646289-196646289 on Build GRCh38
rs771966537 Chr.2: 196646325-196646325 on Build GRCh38
rs771150375 Chr.2: 196646384-196646384 on Build GRCh38
rs61743572 Chr.2: 196646345-196646345 on Build GRCh38
rs550099389 Chr.2: 196646511-196646511 on Build GRCh38
rs746879767 Chr.2: 196646383-196646383 on Build GRCh38
rs1031686582 Chr.2: 196646462-196646462 on Build GRCh38
rs374839443 Chr.2: 196646451-196646451 on Build GRCh38
rs755601334 Chr.2: 196646303-196646303 on Build GRCh38
rs781244582 Chr.2: 196646533-196646533 on Build GRCh38
rs771097204 Chr.2: 196646334-196646334 on Build GRCh38
rs369939924 Chr.2: 196646132-196646132 on Build GRCh38
rs753502575 Chr.2: 196646369-196646369 on Build GRCh38
rs187124338 Chr.2: 196646255-196646255 on Build GRCh38
rs567112696 Chr.2: 196646343-196646343 on Build GRCh38
rs778514199 Chr.2: 196646316-196646316 on Build GRCh38
rs956109548 Chr.2: 196646485-196646485 on Build GRCh38
rs773250406 Chr.2: 196646329-196646329 on Build GRCh38
rs752342601 Chr.2: 196646497-196646497 on Build GRCh38
rs377254708 Chr.2: 196646338-196646338 on Build GRCh38
rs114388971 Chr.2: 196646261-196646261 on Build GRCh38
rs34248242 Chr.2: 196646498-196646498 on Build GRCh38
rs769870324 Chr.2: 196646538-196646538 on Build GRCh38
rs748139600 Chr.2: 196646250-196646250 on Build GRCh38
rs764635996 Chr.2: 196646346-196646346 on Build GRCh38
rs531382089 Chr.2: 196646228-196646228 on Build GRCh38
rs754675524 Chr.2: 196646542-196646544 on Build GRCh38
rs200183130 Chr.2: 196646372-196646372 on Build GRCh38
rs772078951 Chr.2: 196646168-196646168 on Build GRCh38
rs372552459 Chr.2: 196646371-196646371 on Build GRCh38
rs764843571 Chr.2: 196646495-196646495 on Build GRCh38
rs1011232569 Chr.2: 196646141-196646141 on Build GRCh38

View Full Product Details