Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 77426966-77427524 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VIPAS39
Entrez Gene ID
63894
Gene Name
VPS33B interacting protein, apical-basolateral polarity regulator, spe-39 homolog
Gene Aliases
C14orf133, SPE-39, SPE39, VIPAR, VPS16B, hSPE-39
UniGene
Hs.16157
Gene Chromosome Location
Chr.14: 77426675-77457640 on Build GRCh38
Transcripts
NM_001193314.1 NM_001193315.1 NM_001193316.1 NM_001193317.1 NM_022067.3 XM_011537066.1 XM_017021579.1

SNP Detail

SNP ID Chromosome Location
rs921854316 Chr.14: 77427025-77427025 on Build GRCh38
rs948264623 Chr.14: 77427388-77427388 on Build GRCh38
rs921860094 Chr.14: 77427349-77427349 on Build GRCh38
rs1048859591 Chr.14: 77427066-77427066 on Build GRCh38
rs184406103 Chr.14: 77427078-77427078 on Build GRCh38
rs561082655 Chr.14: 77427479-77427479 on Build GRCh38
rs970311915 Chr.14: 77427107-77427107 on Build GRCh38
rs1023660917 Chr.14: 77427094-77427094 on Build GRCh38
rs1044706011 Chr.14: 77427499-77427499 on Build GRCh38
rs956320036 Chr.14: 77427196-77427196 on Build GRCh38
rs8009177 Chr.14: 77427088-77427088 on Build GRCh38
rs1011906770 Chr.14: 77427091-77427091 on Build GRCh38
rs985974744 Chr.14: 77427019-77427019 on Build GRCh38
rs907741425 Chr.14: 77427428-77427428 on Build GRCh38
rs78980929 Chr.14: 77427085-77427085 on Build GRCh38
rs1033146188 Chr.14: 77427152-77427152 on Build GRCh38
rs995454022 Chr.14: 77427455-77427455 on Build GRCh38
rs564726501 Chr.14: 77427379-77427379 on Build GRCh38
rs762173985 Chr.14: 77427338-77427338 on Build GRCh38
rs181857719 Chr.14: 77427461-77427461 on Build GRCh38
rs755634666 Chr.14: 77427202-77427202 on Build GRCh38
rs545031919 Chr.14: 77427306-77427306 on Build GRCh38
rs974693586 Chr.14: 77427339-77427339 on Build GRCh38
rs552959243 Chr.14: 77427429-77427429 on Build GRCh38
rs896818672 Chr.14: 77427442-77427442 on Build GRCh38
rs189471916 Chr.14: 77427139-77427139 on Build GRCh38
rs572517135 Chr.14: 77427218-77427218 on Build GRCh38
rs1000782689 Chr.14: 77427138-77427138 on Build GRCh38
rs112684413 Chr.14: 77427255-77427255 on Build GRCh38
rs1036739370 Chr.14: 77427437-77427437 on Build GRCh38
rs144661415 Chr.14: 77427061-77427061 on Build GRCh38
rs923938956 Chr.14: 77427166-77427166 on Build GRCh38
rs1011937869 Chr.14: 77427486-77427486 on Build GRCh38
rs1046971577 Chr.14: 77427413-77427413 on Build GRCh38
rs930492941 Chr.14: 77427048-77427048 on Build GRCh38
rs1004595321 Chr.14: 77427079-77427079 on Build GRCh38
rs1049371869 Chr.14: 77427415-77427415 on Build GRCh38
rs966043251 Chr.14: 77427226-77427226 on Build GRCh38

View Full Product Details