Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 44966101-44966460 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TERB2
Entrez Gene ID
145645
Gene Name
telomere repeat binding bouquet formation protein 2
Gene Aliases
C15orf43
UniGene
Hs.567701
Gene Chromosome Location
Chr.15: 44956674-44979223 on Build GRCh38
Transcripts
NM_152448.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3501607 c.428C>T p.S143F Chr.15: 44966237-44966237 on Build GRCh38
222292 c.388G>A p.E130K Chr.15: 44966197-44966197 on Build GRCh38
1608413 c.405delA p.T136fs*16 Chr.15: 44966214-44966214 on Build GRCh38
5007817 c.379A>T p.T127S Chr.15: 44966188-44966188 on Build GRCh38
1708098 c.362C>T p.T121I Chr.15: 44966171-44966171 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs202094227 Chr.15: 44966243-44966243 on Build GRCh38
rs770697777 Chr.15: 44966230-44966230 on Build GRCh38
rs773938133 Chr.15: 44966232-44966232 on Build GRCh38
rs769886013 Chr.15: 44966213-44966213 on Build GRCh38
rs35986068 Chr.15: 44966374-44966375 on Build GRCh38
rs899165935 Chr.15: 44966388-44966389 on Build GRCh38
rs201486222 Chr.15: 44966196-44966196 on Build GRCh38
rs540973128 Chr.15: 44966130-44966130 on Build GRCh38
rs966510971 Chr.15: 44966137-44966137 on Build GRCh38
rs779015160 Chr.15: 44966154-44966154 on Build GRCh38
rs750660742 Chr.15: 44966272-44966272 on Build GRCh38
rs774552757 Chr.15: 44966319-44966319 on Build GRCh38
rs748138616 Chr.15: 44966211-44966211 on Build GRCh38
rs753257058 Chr.15: 44966141-44966141 on Build GRCh38
rs563991948 Chr.15: 44966143-44966143 on Build GRCh38
rs768801269 Chr.15: 44966294-44966294 on Build GRCh38
rs780260629 Chr.15: 44966175-44966175 on Build GRCh38
rs77861669 Chr.15: 44966160-44966160 on Build GRCh38
rs549093018 Chr.15: 44966401-44966401 on Build GRCh38
rs866428355 Chr.15: 44966212-44966212 on Build GRCh38
rs757566716 Chr.15: 44966267-44966267 on Build GRCh38
rs907686465 Chr.15: 44966371-44966371 on Build GRCh38
rs569669628 Chr.15: 44966307-44966307 on Build GRCh38
rs199669613 Chr.15: 44966358-44966359 on Build GRCh38
rs768411130 Chr.15: 44966224-44966224 on Build GRCh38
rs758616832 Chr.15: 44966280-44966280 on Build GRCh38
rs746567003 Chr.15: 44966143-44966143 on Build GRCh38
rs372123246 Chr.15: 44966281-44966281 on Build GRCh38
rs754284200 Chr.15: 44966263-44966263 on Build GRCh38
rs764605417 Chr.15: 44966258-44966258 on Build GRCh38
rs149924364 Chr.15: 44966239-44966239 on Build GRCh38
rs534212444 Chr.15: 44966373-44966374 on Build GRCh38
rs398039517 Chr.15: 44966381-44966382 on Build GRCh38
rs778147931 Chr.15: 44966148-44966148 on Build GRCh38
rs367780363 Chr.15: 44966167-44966167 on Build GRCh38
rs763081707 Chr.15: 44966275-44966275 on Build GRCh38
rs747110702 Chr.15: 44966283-44966283 on Build GRCh38
rs940818037 Chr.15: 44966374-44966374 on Build GRCh38
rs111436722 Chr.15: 44966162-44966162 on Build GRCh38
rs144596851 Chr.15: 44966441-44966441 on Build GRCh38
rs368720229 Chr.15: 44966247-44966247 on Build GRCh38
rs529009551 Chr.15: 44966344-44966344 on Build GRCh38
rs1038186613 Chr.15: 44966375-44966375 on Build GRCh38
rs73419356 Chr.15: 44966352-44966352 on Build GRCh38
rs749683352 Chr.15: 44966151-44966151 on Build GRCh38
rs779323051 Chr.15: 44966271-44966271 on Build GRCh38
rs773238138 Chr.15: 44966218-44966218 on Build GRCh38

View Full Product Details