Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 72287146-72287498 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CELF6
Entrez Gene ID
60677
Gene Name
CUGBP, Elav-like family member 6
Gene Aliases
BRUNOL6
UniGene
Hs.348342
Gene Chromosome Location
Chr.15: 72284727-72320184 on Build GRCh38
Transcripts
NM_001172684.1 NM_001172685.1 NM_052840.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4813651 c.1089T>C p.D363D Chr.15: 72287283-72287283 on Build GRCh38
3503704 c.1010C>T p.A337V Chr.15: 72287362-72287362 on Build GRCh38
3944227 c.1403A>T p.K468M Chr.15: 72287308-72287308 on Build GRCh38
6626191 c.1377C>T p.G459G Chr.15: 72287334-72287334 on Build GRCh38
4671427 c.1396A>T p.R466W Chr.15: 72287315-72287315 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs946705323 Chr.15: 72287199-72287199 on Build GRCh38
rs746434889 Chr.15: 72287273-72287273 on Build GRCh38
rs368557479 Chr.15: 72287364-72287364 on Build GRCh38
rs780007802 Chr.15: 72287346-72287346 on Build GRCh38
rs147656826 Chr.15: 72287304-72287304 on Build GRCh38
rs765370247 Chr.15: 72287341-72287341 on Build GRCh38
rs746332755 Chr.15: 72287400-72287400 on Build GRCh38
rs915214324 Chr.15: 72287188-72287188 on Build GRCh38
rs145685721 Chr.15: 72287276-72287276 on Build GRCh38
rs746140894 Chr.15: 72287249-72287249 on Build GRCh38
rs771675030 Chr.15: 72287424-72287424 on Build GRCh38
rs767169894 Chr.15: 72287226-72287226 on Build GRCh38
rs780658521 Chr.15: 72287423-72287423 on Build GRCh38
rs899527099 Chr.15: 72287205-72287205 on Build GRCh38
rs760379153 Chr.15: 72287294-72287294 on Build GRCh38
rs17852852 Chr.15: 72287305-72287305 on Build GRCh38
rs192243214 Chr.15: 72287467-72287467 on Build GRCh38
rs865826368 Chr.15: 72287422-72287422 on Build GRCh38
rs988541549 Chr.15: 72287174-72287174 on Build GRCh38
rs781551701 Chr.15: 72287387-72287387 on Build GRCh38
rs758582557 Chr.15: 72287248-72287248 on Build GRCh38
rs773764325 Chr.15: 72287277-72287277 on Build GRCh38
rs1050474881 Chr.15: 72287213-72287213 on Build GRCh38
rs770306505 Chr.15: 72287417-72287417 on Build GRCh38
rs556722577 Chr.15: 72287220-72287220 on Build GRCh38
rs772954799 Chr.15: 72287293-72287293 on Build GRCh38
rs766332023 Chr.15: 72287298-72287298 on Build GRCh38
rs199625679 Chr.15: 72287344-72287344 on Build GRCh38
rs757606553 Chr.15: 72287369-72287369 on Build GRCh38
rs771601100 Chr.15: 72287289-72287289 on Build GRCh38
rs752900807 Chr.15: 72287243-72287243 on Build GRCh38
rs770780248 Chr.15: 72287444-72287444 on Build GRCh38
rs760413690 Chr.15: 72287441-72287441 on Build GRCh38
rs531424466 Chr.15: 72287184-72287184 on Build GRCh38
rs771326396 Chr.15: 72287251-72287251 on Build GRCh38
rs777828179 Chr.15: 72287357-72287357 on Build GRCh38
rs1003574784 Chr.15: 72287465-72287465 on Build GRCh38
rs1003665538 Chr.15: 72287401-72287401 on Build GRCh38
rs536248281 Chr.15: 72287461-72287461 on Build GRCh38
rs202206831 Chr.15: 72287230-72287230 on Build GRCh38
rs761476973 Chr.15: 72287283-72287283 on Build GRCh38
rs772722189 Chr.15: 72287433-72287433 on Build GRCh38
rs370681265 Chr.15: 72287256-72287256 on Build GRCh38
rs201046812 Chr.15: 72287237-72287237 on Build GRCh38
rs890724677 Chr.15: 72287234-72287234 on Build GRCh38
rs199611770 Chr.15: 72287250-72287250 on Build GRCh38
rs763919037 Chr.15: 72287458-72287458 on Build GRCh38
rs758559041 Chr.15: 72287348-72287348 on Build GRCh38
rs201504443 Chr.15: 72287320-72287320 on Build GRCh38

View Full Product Details