Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 67209035-67209626 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AAGAB
Entrez Gene ID
79719
Gene Name
alpha- and gamma-adaptin binding protein
Gene Aliases
KPPP1, PPKP1, PPKP1A, p34
UniGene
Hs.254642
Gene Chromosome Location
Chr.15: 67200675-67255198 on Build GRCh38
Transcripts
NM_001271885.1 NM_001271886.1 NM_024666.4 XM_011522020.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5913639 c.536-8C>T p.? Chr.15: 67209552-67209552 on Build GRCh38
238612 c.540G>T p.R180S Chr.15: 67209540-67209540 on Build GRCh38
278484 c.549C>A p.G183G Chr.15: 67209531-67209531 on Build GRCh38
3503368 c.540G>A p.R180R Chr.15: 67209540-67209540 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1026814015 Chr.15: 67209587-67209587 on Build GRCh38
rs952150125 Chr.15: 67209256-67209256 on Build GRCh38
rs914657144 Chr.15: 67209163-67209163 on Build GRCh38
rs532057896 Chr.15: 67209364-67209364 on Build GRCh38
rs144607325 Chr.15: 67209236-67209236 on Build GRCh38
rs184821611 Chr.15: 67209445-67209445 on Build GRCh38
rs778154330 Chr.15: 67209374-67209374 on Build GRCh38
rs781169916 Chr.15: 67209463-67209463 on Build GRCh38
rs745359969 Chr.15: 67209510-67209510 on Build GRCh38
rs912806711 Chr.15: 67209386-67209386 on Build GRCh38
rs890805128 Chr.15: 67209457-67209457 on Build GRCh38
rs377196286 Chr.15: 67209574-67209574 on Build GRCh38
rs930237052 Chr.15: 67209105-67209105 on Build GRCh38
rs1023355379 Chr.15: 67209267-67209267 on Build GRCh38
rs748600139 Chr.15: 67209098-67209098 on Build GRCh38
rs28693872 Chr.15: 67209444-67209444 on Build GRCh38
rs778740230 Chr.15: 67209506-67209506 on Build GRCh38
rs372570873 Chr.15: 67209459-67209459 on Build GRCh38
rs755937432 Chr.15: 67209468-67209468 on Build GRCh38
rs147319532 Chr.15: 67209080-67209080 on Build GRCh38
rs148522618 Chr.15: 67209436-67209436 on Build GRCh38
rs1043568619 Chr.15: 67209203-67209203 on Build GRCh38
rs35202044 Chr.15: 67209476-67209477 on Build GRCh38
rs1018169546 Chr.15: 67209156-67209156 on Build GRCh38
rs769167919 Chr.15: 67209414-67209414 on Build GRCh38
rs774489003 Chr.15: 67209566-67209566 on Build GRCh38
rs763336419 Chr.15: 67209550-67209550 on Build GRCh38
rs1025859017 Chr.15: 67209340-67209340 on Build GRCh38
rs995658576 Chr.15: 67209303-67209303 on Build GRCh38
rs907869983 Chr.15: 67209302-67209302 on Build GRCh38
rs140745485 Chr.15: 67209130-67209130 on Build GRCh38
rs777052173 Chr.15: 67209429-67209429 on Build GRCh38
rs777190296 Chr.15: 67209514-67209514 on Build GRCh38
rs759650985 Chr.15: 67209582-67209582 on Build GRCh38
rs189242691 Chr.15: 67209502-67209502 on Build GRCh38
rs796134149 Chr.15: 67209308-67209310 on Build GRCh38
rs1054537703 Chr.15: 67209399-67209399 on Build GRCh38
rs946524087 Chr.15: 67209171-67209171 on Build GRCh38
rs963939805 Chr.15: 67209551-67209551 on Build GRCh38
rs912596896 Chr.15: 67209113-67209113 on Build GRCh38
rs777645464 Chr.15: 67209484-67209484 on Build GRCh38
rs752636792 Chr.15: 67209466-67209466 on Build GRCh38
rs111424098 Chr.15: 67209271-67209271 on Build GRCh38
rs16953610 Chr.15: 67209254-67209254 on Build GRCh38
rs1045973895 Chr.15: 67209331-67209331 on Build GRCh38
rs201808670 Chr.15: 67209494-67209494 on Build GRCh38
rs751615611 Chr.15: 67209454-67209454 on Build GRCh38
rs963887014 Chr.15: 67209218-67209218 on Build GRCh38
rs574338868 Chr.15: 67209099-67209099 on Build GRCh38
rs145787268 Chr.15: 67209265-67209265 on Build GRCh38
rs562936101 Chr.15: 67209359-67209359 on Build GRCh38
rs765657234 Chr.15: 67209433-67209433 on Build GRCh38
rs748536642 Chr.15: 67209517-67209517 on Build GRCh38
rs986798319 Chr.15: 67209111-67209111 on Build GRCh38
rs1019940826 Chr.15: 67209096-67209096 on Build GRCh38
rs112294512 Chr.15: 67209377-67209377 on Build GRCh38
rs942701398 Chr.15: 67209117-67209117 on Build GRCh38
rs1032739551 Chr.15: 67209059-67209059 on Build GRCh38
rs1010344128 Chr.15: 67209084-67209084 on Build GRCh38
rs771537657 Chr.15: 67209511-67209511 on Build GRCh38
rs766554130 Chr.15: 67209561-67209561 on Build GRCh38
rs773512116 Chr.15: 67209540-67209540 on Build GRCh38
rs767563608 Chr.15: 67209590-67209590 on Build GRCh38
rs950055580 Chr.15: 67209326-67209326 on Build GRCh38
rs770294425 Chr.15: 67209531-67209531 on Build GRCh38
rs1056319038 Chr.15: 67209251-67209251 on Build GRCh38
rs892866805 Chr.15: 67209079-67209079 on Build GRCh38

View Full Product Details