Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 83256846-83257442 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BNC1
Entrez Gene ID
646
Gene Name
basonuclin 1
Gene Aliases
BNC, BSN1, HsT19447
UniGene
Hs.459153
Gene Chromosome Location
Chr.15: 83255884-83284716 on Build GRCh38
Transcripts
NM_001301206.1 NM_001717.3 XM_011521893.1 XM_011521894.1

SNP Detail

SNP ID Chromosome Location
rs769192834 Chr.15: 83257284-83257284 on Build GRCh38
rs191641929 Chr.15: 83257386-83257386 on Build GRCh38
rs1006063522 Chr.15: 83256910-83256910 on Build GRCh38
rs977703355 Chr.15: 83257314-83257314 on Build GRCh38
rs1015259237 Chr.15: 83257329-83257329 on Build GRCh38
rs867775441 Chr.15: 83257162-83257162 on Build GRCh38
rs1006116017 Chr.15: 83257192-83257192 on Build GRCh38
rs569920758 Chr.15: 83257373-83257373 on Build GRCh38
rs996786584 Chr.15: 83257275-83257275 on Build GRCh38
rs111633086 Chr.15: 83257374-83257374 on Build GRCh38
rs533617983 Chr.15: 83257051-83257051 on Build GRCh38
rs755236554 Chr.15: 83257398-83257398 on Build GRCh38
rs759082211 Chr.15: 83257228-83257228 on Build GRCh38
rs977652789 Chr.15: 83257075-83257075 on Build GRCh38
rs186799026 Chr.15: 83256967-83256967 on Build GRCh38
rs73438937 Chr.15: 83257068-83257068 on Build GRCh38
rs150026149 Chr.15: 83257357-83257357 on Build GRCh38
rs1021827130 Chr.15: 83257289-83257289 on Build GRCh38
rs914716132 Chr.15: 83257062-83257062 on Build GRCh38
rs948832981 Chr.15: 83257360-83257360 on Build GRCh38
rs968872055 Chr.15: 83257303-83257303 on Build GRCh38
rs868594764 Chr.15: 83257393-83257393 on Build GRCh38
rs953210239 Chr.15: 83257045-83257045 on Build GRCh38
rs1057433037 Chr.15: 83257109-83257109 on Build GRCh38
rs1044887325 Chr.15: 83257144-83257144 on Build GRCh38
rs948780577 Chr.15: 83257135-83257135 on Build GRCh38
rs1014837678 Chr.15: 83257198-83257198 on Build GRCh38
rs894562624 Chr.15: 83257134-83257134 on Build GRCh38
rs1021286664 Chr.15: 83257191-83257191 on Build GRCh38
rs976273436 Chr.15: 83257024-83257024 on Build GRCh38
rs1054322127 Chr.15: 83257225-83257225 on Build GRCh38
rs551692814 Chr.15: 83257336-83257336 on Build GRCh38
rs757201026 Chr.15: 83257055-83257055 on Build GRCh38
rs924644795 Chr.15: 83257085-83257085 on Build GRCh38
rs1014571169 Chr.15: 83256912-83256912 on Build GRCh38
rs924896122 Chr.15: 83257325-83257325 on Build GRCh38
rs954993852 Chr.15: 83256878-83256878 on Build GRCh38
rs961414398 Chr.15: 83256958-83256958 on Build GRCh38
rs370946316 Chr.15: 83257278-83257278 on Build GRCh38
rs1030091413 Chr.15: 83257027-83257027 on Build GRCh38
rs563123197 Chr.15: 83256950-83256950 on Build GRCh38
rs1049638005 Chr.15: 83257298-83257298 on Build GRCh38
rs66891986 Chr.15: 83257043-83257044 on Build GRCh38
rs567941770 Chr.15: 83257288-83257288 on Build GRCh38
rs528231407 Chr.15: 83257119-83257119 on Build GRCh38
rs1025220322 Chr.15: 83257404-83257404 on Build GRCh38
rs892934889 Chr.15: 83257256-83257256 on Build GRCh38
rs985864714 Chr.15: 83257286-83257286 on Build GRCh38
rs992861305 Chr.15: 83257346-83257346 on Build GRCh38
rs553542242 Chr.15: 83256915-83256915 on Build GRCh38
rs938687094 Chr.15: 83257098-83257098 on Build GRCh38
rs140650192 Chr.15: 83257042-83257043 on Build GRCh38
rs959672048 Chr.15: 83257034-83257034 on Build GRCh38
rs536331449 Chr.15: 83257290-83257290 on Build GRCh38
rs960529944 Chr.15: 83257331-83257331 on Build GRCh38
rs974300729 Chr.15: 83257016-83257017 on Build GRCh38
rs897704218 Chr.15: 83257214-83257214 on Build GRCh38
rs762617437 Chr.15: 83257418-83257418 on Build GRCh38
rs909088320 Chr.15: 83257415-83257415 on Build GRCh38

View Full Product Details