Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 123630160-123630582 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF2B1
Entrez Gene ID
1967
Gene Name
eukaryotic translation initiation factor 2B subunit alpha
Gene Aliases
EIF2B, EIF2BA
UniGene
Hs.741273
Gene Chromosome Location
Chr.12: 123621023-123633776 on Build GRCh38
Transcripts
NM_001414.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3376158 c.252+9C>A p.? Chr.12: 123630388-123630388 on Build GRCh38
6021188 c.200C>T p.S67F Chr.12: 123630449-123630449 on Build GRCh38
3376159 c.252+1G>A p.? Chr.12: 123630396-123630396 on Build GRCh38
299867 c.251C>T p.S84F Chr.12: 123630398-123630398 on Build GRCh38
5940258 c.145C>T p.L49F Chr.12: 123630504-123630504 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200972398 Chr.12: 123630551-123630551 on Build GRCh38
rs757985338 Chr.12: 123630246-123630246 on Build GRCh38
rs374302327 Chr.12: 123630307-123630307 on Build GRCh38
rs863225050 Chr.12: 123630210-123630210 on Build GRCh38
rs962257544 Chr.12: 123630509-123630509 on Build GRCh38
rs759728057 Chr.12: 123630193-123630193 on Build GRCh38
rs781471132 Chr.12: 123630253-123630254 on Build GRCh38
rs184545759 Chr.12: 123630471-123630471 on Build GRCh38
rs765287085 Chr.12: 123630205-123630205 on Build GRCh38
rs749254137 Chr.12: 123630414-123630414 on Build GRCh38
rs199544322 Chr.12: 123630473-123630473 on Build GRCh38
rs775739205 Chr.12: 123630217-123630217 on Build GRCh38
rs202096209 Chr.12: 123630537-123630537 on Build GRCh38
rs768881324 Chr.12: 123630379-123630379 on Build GRCh38
rs906502322 Chr.12: 123630422-123630422 on Build GRCh38
rs779910329 Chr.12: 123630508-123630508 on Build GRCh38
rs769901842 Chr.12: 123630352-123630352 on Build GRCh38
rs768076568 Chr.12: 123630249-123630249 on Build GRCh38
rs1003540837 Chr.12: 123630424-123630424 on Build GRCh38
rs774371735 Chr.12: 123630380-123630380 on Build GRCh38
rs748375449 Chr.12: 123630255-123630256 on Build GRCh38
rs781247492 Chr.12: 123630317-123630317 on Build GRCh38
rs777547006 Chr.12: 123630457-123630457 on Build GRCh38
rs371060103 Chr.12: 123630381-123630381 on Build GRCh38
rs764262014 Chr.12: 123630218-123630218 on Build GRCh38
rs930417910 Chr.12: 123630332-123630332 on Build GRCh38
rs769746532 Chr.12: 123630499-123630499 on Build GRCh38
rs746822849 Chr.12: 123630308-123630308 on Build GRCh38
rs528467752 Chr.12: 123630386-123630386 on Build GRCh38
rs142755878 Chr.12: 123630257-123630257 on Build GRCh38
rs761922213 Chr.12: 123630552-123630552 on Build GRCh38
rs200139223 Chr.12: 123630384-123630384 on Build GRCh38
rs113994006 Chr.12: 123630396-123630396 on Build GRCh38
rs141016684 Chr.12: 123630403-123630403 on Build GRCh38
rs774696782 Chr.12: 123630549-123630549 on Build GRCh38
rs768721677 Chr.12: 123630514-123630514 on Build GRCh38
rs762973483 Chr.12: 123630377-123630377 on Build GRCh38
rs368227950 Chr.12: 123630347-123630347 on Build GRCh38
rs751315607 Chr.12: 123630448-123630448 on Build GRCh38
rs752266121 Chr.12: 123630445-123630445 on Build GRCh38
rs747617594 Chr.12: 123630265-123630265 on Build GRCh38
rs1050397 Chr.12: 123630252-123630252 on Build GRCh38
rs1012516401 Chr.12: 123630408-123630408 on Build GRCh38
rs757509025 Chr.12: 123630248-123630248 on Build GRCh38
rs776365178 Chr.12: 123630190-123630190 on Build GRCh38
rs770625902 Chr.12: 123630312-123630312 on Build GRCh38
rs745623947 Chr.12: 123630486-123630486 on Build GRCh38
rs749376947 Chr.12: 123630513-123630513 on Build GRCh38
rs775578205 Chr.12: 123630357-123630357 on Build GRCh38
rs758142806 Chr.12: 123630269-123630269 on Build GRCh38
rs371367101 Chr.12: 123630481-123630481 on Build GRCh38
rs138110703 Chr.12: 123630397-123630397 on Build GRCh38
rs189474095 Chr.12: 123630561-123630561 on Build GRCh38

View Full Product Details