Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 56973050-56973648 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CETP
Entrez Gene ID
1071
Gene Name
cholesteryl ester transfer protein
Gene Aliases
BPIFF, HDLCQ10
UniGene
Hs.89538
Gene Chromosome Location
Chr.16: 56961923-56983844 on Build GRCh38
Transcripts
NM_000078.2 NM_001286085.1 XM_006721124.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5567936 c.824C>A p.S275Y Chr.16: 56973404-56973404 on Build GRCh38
971753 c.774C>A p.V258V Chr.16: 56973354-56973354 on Build GRCh38
3402380 c.751G>A p.G251S Chr.16: 56973331-56973331 on Build GRCh38
4671837 c.846C>A p.F282L Chr.16: 56973426-56973426 on Build GRCh38
971754 c.859T>G p.F287V Chr.16: 56973439-56973439 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766679880 Chr.16: 56973482-56973482 on Build GRCh38
rs750662640 Chr.16: 56973407-56973407 on Build GRCh38
rs768951962 Chr.16: 56973306-56973306 on Build GRCh38
rs138281147 Chr.16: 56973599-56973599 on Build GRCh38
rs202228107 Chr.16: 56973314-56973314 on Build GRCh38
rs12691052 Chr.16: 56973600-56973600 on Build GRCh38
rs11076176 Chr.16: 56973534-56973534 on Build GRCh38
rs187468344 Chr.16: 56973128-56973128 on Build GRCh38
rs752622552 Chr.16: 56973327-56973327 on Build GRCh38
rs765955727 Chr.16: 56973557-56973558 on Build GRCh38
rs544043137 Chr.16: 56973155-56973155 on Build GRCh38
rs375666118 Chr.16: 56973447-56973447 on Build GRCh38
rs763347312 Chr.16: 56973296-56973296 on Build GRCh38
rs1002599666 Chr.16: 56973293-56973293 on Build GRCh38
rs766234439 Chr.16: 56973558-56973558 on Build GRCh38
rs765185843 Chr.16: 56973321-56973321 on Build GRCh38
rs753227095 Chr.16: 56973500-56973501 on Build GRCh38
rs779536353 Chr.16: 56973502-56973502 on Build GRCh38
rs9930761 Chr.16: 56973280-56973280 on Build GRCh38
rs773875617 Chr.16: 56973556-56973556 on Build GRCh38
rs986880151 Chr.16: 56973172-56973172 on Build GRCh38
rs760335739 Chr.16: 56973309-56973309 on Build GRCh38
rs142459781 Chr.16: 56973475-56973475 on Build GRCh38
rs756383548 Chr.16: 56973408-56973408 on Build GRCh38
rs566144069 Chr.16: 56973528-56973528 on Build GRCh38
rs375348327 Chr.16: 56973335-56973335 on Build GRCh38
rs373324495 Chr.16: 56973189-56973189 on Build GRCh38
rs775928502 Chr.16: 56973285-56973285 on Build GRCh38
rs749397888 Chr.16: 56973412-56973412 on Build GRCh38
rs562343117 Chr.16: 56973215-56973215 on Build GRCh38
rs748767072 Chr.16: 56973547-56973547 on Build GRCh38
rs5883 Chr.16: 56973441-56973441 on Build GRCh38
rs968095821 Chr.16: 56973107-56973107 on Build GRCh38
rs779285308 Chr.16: 56973573-56973573 on Build GRCh38
rs750341621 Chr.16: 56973558-56973559 on Build GRCh38
rs776797827 Chr.16: 56973409-56973409 on Build GRCh38
rs776459790 Chr.16: 56973381-56973381 on Build GRCh38
rs769805074 Chr.16: 56973368-56973368 on Build GRCh38
rs752437798 Chr.16: 56973371-56973371 on Build GRCh38
rs289714 Chr.16: 56973539-56973539 on Build GRCh38
rs780590693 Chr.16: 56973370-56973370 on Build GRCh38
rs267604581 Chr.16: 56973458-56973458 on Build GRCh38
rs761062555 Chr.16: 56973385-56973385 on Build GRCh38
rs758374809 Chr.16: 56973331-56973331 on Build GRCh38
rs772609946 Chr.16: 56973434-56973434 on Build GRCh38
rs753383942 Chr.16: 56973312-56973312 on Build GRCh38
rs956673991 Chr.16: 56973369-56973369 on Build GRCh38
rs577324229 Chr.16: 56973255-56973255 on Build GRCh38
rs184684961 Chr.16: 56973592-56973592 on Build GRCh38
rs768204247 Chr.16: 56973551-56973551 on Build GRCh38
rs766142881 Chr.16: 56973310-56973310 on Build GRCh38
rs767253260 Chr.16: 56973470-56973470 on Build GRCh38
rs781455323 Chr.16: 56973357-56973357 on Build GRCh38
rs28381707 Chr.16: 56973384-56973384 on Build GRCh38
rs773646267 Chr.16: 56973438-56973438 on Build GRCh38
rs756231195 Chr.16: 56973476-56973476 on Build GRCh38
rs369941536 Chr.16: 56973449-56973449 on Build GRCh38
rs772059097 Chr.16: 56973559-56973559 on Build GRCh38
rs745956017 Chr.16: 56973358-56973358 on Build GRCh38
rs561462414 Chr.16: 56973377-56973377 on Build GRCh38
rs757221175 Chr.16: 56973539-56973540 on Build GRCh38
rs762587120 Chr.16: 56973444-56973444 on Build GRCh38
rs148510735 Chr.16: 56973559-56973560 on Build GRCh38
rs755528414 Chr.16: 56973413-56973413 on Build GRCh38
rs774807909 Chr.16: 56973443-56973443 on Build GRCh38
rs200375648 Chr.16: 56973452-56973452 on Build GRCh38
rs779930161 Chr.16: 56973411-56973411 on Build GRCh38
rs367582606 Chr.16: 56973294-56973294 on Build GRCh38
rs767971569 Chr.16: 56973363-56973366 on Build GRCh38
rs745468187 Chr.16: 56973439-56973439 on Build GRCh38
rs775689281 Chr.16: 56973316-56973316 on Build GRCh38
rs751288556 Chr.16: 56973575-56973575 on Build GRCh38
rs749711042 Chr.16: 56973372-56973372 on Build GRCh38
rs1024277141 Chr.16: 56973096-56973096 on Build GRCh38
rs935439147 Chr.16: 56973154-56973154 on Build GRCh38
rs201503514 Chr.16: 56973398-56973398 on Build GRCh38
rs144460063 Chr.16: 56973332-56973332 on Build GRCh38
rs747682595 Chr.16: 56973122-56973122 on Build GRCh38
rs898366112 Chr.16: 56973218-56973218 on Build GRCh38
rs750039511 Chr.16: 56973162-56973165 on Build GRCh38
rs779630141 Chr.16: 56973414-56973414 on Build GRCh38
rs912422055 Chr.16: 56973195-56973195 on Build GRCh38
rs1056899781 Chr.16: 56973489-56973489 on Build GRCh38
rs139594305 Chr.16: 56973374-56973374 on Build GRCh38
rs28381708 Chr.16: 56973501-56973501 on Build GRCh38
rs757111919 Chr.16: 56973400-56973400 on Build GRCh38
rs147566657 Chr.16: 56973406-56973406 on Build GRCh38
rs764036569 Chr.16: 56973392-56973392 on Build GRCh38
rs748488549 Chr.16: 56973433-56973433 on Build GRCh38
rs760236205 Chr.16: 56973394-56973395 on Build GRCh38
rs919778015 Chr.16: 56973610-56973610 on Build GRCh38
rs930987455 Chr.16: 56973229-56973229 on Build GRCh38
rs150475423 Chr.16: 56973364-56973364 on Build GRCh38
rs1012804012 Chr.16: 56973313-56973313 on Build GRCh38
rs942516270 Chr.16: 56973203-56973203 on Build GRCh38
rs774886096 Chr.16: 56973307-56973307 on Build GRCh38
rs573864685 Chr.16: 56973141-56973141 on Build GRCh38
rs747307952 Chr.16: 56973383-56973383 on Build GRCh38
rs779564582 Chr.16: 56973171-56973171 on Build GRCh38
rs748614926 Chr.16: 56973518-56973518 on Build GRCh38
rs763714949 Chr.16: 56973428-56973428 on Build GRCh38
rs35878799 Chr.16: 56973336-56973336 on Build GRCh38
rs762734977 Chr.16: 56973387-56973387 on Build GRCh38
rs777935460 Chr.16: 56973533-56973533 on Build GRCh38
rs924032449 Chr.16: 56973132-56973132 on Build GRCh38

View Full Product Details