Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 6678883-6679435 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C3
Entrez Gene ID
718
Gene Name
complement component 3
Gene Aliases
AHUS5, ARMD9, ASP, C3a, C3b, CPAMD1, HEL-S-62p
UniGene
Hs.529053
Gene Chromosome Location
Chr.19: 6677835-6720682 on Build GRCh38
Transcripts
NM_000064.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
309717 c.4628A>G p.Y1543C Chr.19: 6679127-6679127 on Build GRCh38
243366 c.4538G>T p.C1513F Chr.19: 6679415-6679415 on Build GRCh38
3540982 c.4533C>T p.C1511C Chr.19: 6679420-6679420 on Build GRCh38
5584645 c.4528C>T p.L1510L Chr.19: 6679425-6679425 on Build GRCh38
4749353 c.4558A>T p.I1520L Chr.19: 6679197-6679197 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs879060382 Chr.19: 6679298-6679298 on Build GRCh38
rs762786430 Chr.19: 6679376-6679376 on Build GRCh38
rs763214910 Chr.19: 6679134-6679134 on Build GRCh38
rs748579533 Chr.19: 6679113-6679113 on Build GRCh38
rs773558774 Chr.19: 6679119-6679119 on Build GRCh38
rs1054800933 Chr.19: 6679072-6679072 on Build GRCh38
rs771542274 Chr.19: 6679099-6679099 on Build GRCh38
rs112746915 Chr.19: 6679124-6679124 on Build GRCh38
rs998970929 Chr.19: 6679290-6679290 on Build GRCh38
rs770231810 Chr.19: 6679114-6679114 on Build GRCh38
rs917002976 Chr.19: 6678991-6678991 on Build GRCh38
rs368187091 Chr.19: 6679019-6679019 on Build GRCh38
rs765181421 Chr.19: 6679253-6679253 on Build GRCh38
rs377076572 Chr.19: 6679389-6679389 on Build GRCh38
rs979929863 Chr.19: 6679032-6679032 on Build GRCh38
rs765198711 Chr.19: 6679359-6679359 on Build GRCh38
rs377581390 Chr.19: 6679353-6679353 on Build GRCh38
rs1036489609 Chr.19: 6678977-6678977 on Build GRCh38
rs957097059 Chr.19: 6679309-6679309 on Build GRCh38
rs344555 Chr.19: 6679349-6679349 on Build GRCh38
rs1042405176 Chr.19: 6679106-6679106 on Build GRCh38
rs565817722 Chr.19: 6679331-6679331 on Build GRCh38
rs372710787 Chr.19: 6679242-6679242 on Build GRCh38
rs1022556048 Chr.19: 6679333-6679333 on Build GRCh38
rs748754304 Chr.19: 6679220-6679220 on Build GRCh38
rs750272939 Chr.19: 6679370-6679370 on Build GRCh38
rs778120984 Chr.19: 6679235-6679235 on Build GRCh38
rs141010917 Chr.19: 6679182-6679182 on Build GRCh38
rs779812568 Chr.19: 6679195-6679195 on Build GRCh38
rs368008466 Chr.19: 6679075-6679075 on Build GRCh38
rs754564033 Chr.19: 6679218-6679218 on Build GRCh38
rs754579424 Chr.19: 6679390-6679390 on Build GRCh38
rs370699397 Chr.19: 6679082-6679082 on Build GRCh38
rs897907408 Chr.19: 6679061-6679061 on Build GRCh38
rs758128289 Chr.19: 6679194-6679194 on Build GRCh38
rs375949866 Chr.19: 6679244-6679244 on Build GRCh38
rs530827970 Chr.19: 6678909-6678909 on Build GRCh38
rs112249284 Chr.19: 6679123-6679123 on Build GRCh38
rs745556347 Chr.19: 6679078-6679078 on Build GRCh38
rs971149882 Chr.19: 6679303-6679303 on Build GRCh38
rs1017157400 Chr.19: 6678933-6678933 on Build GRCh38
rs979834917 Chr.19: 6679305-6679305 on Build GRCh38
rs774422907 Chr.19: 6679154-6679154 on Build GRCh38
rs761625759 Chr.19: 6679186-6679186 on Build GRCh38
rs45511693 Chr.19: 6679310-6679310 on Build GRCh38
rs771480437 Chr.19: 6678951-6678952 on Build GRCh38
rs267605750 Chr.19: 6679170-6679170 on Build GRCh38
rs751135751 Chr.19: 6679207-6679207 on Build GRCh38
rs766252333 Chr.19: 6679380-6679380 on Build GRCh38
rs758120022 Chr.19: 6679080-6679080 on Build GRCh38
rs759624343 Chr.19: 6679160-6679160 on Build GRCh38
rs148377730 Chr.19: 6679259-6679259 on Build GRCh38
rs775429724 Chr.19: 6679258-6679258 on Build GRCh38
rs746452478 Chr.19: 6679098-6679098 on Build GRCh38
rs771087404 Chr.19: 6679136-6679136 on Build GRCh38
rs112584257 Chr.19: 6679405-6679405 on Build GRCh38
rs1019532370 Chr.19: 6679208-6679208 on Build GRCh38
rs930659280 Chr.19: 6679090-6679090 on Build GRCh38
rs939435600 Chr.19: 6678941-6678941 on Build GRCh38
rs935822386 Chr.19: 6679052-6679052 on Build GRCh38
rs772236798 Chr.19: 6679246-6679246 on Build GRCh38
rs770184497 Chr.19: 6679232-6679232 on Build GRCh38
rs765147079 Chr.19: 6679187-6679187 on Build GRCh38
rs769072644 Chr.19: 6679101-6679101 on Build GRCh38
rs949794362 Chr.19: 6678992-6678992 on Build GRCh38
rs750229972 Chr.19: 6679188-6679188 on Build GRCh38
rs111252441 Chr.19: 6679210-6679210 on Build GRCh38
rs181596974 Chr.19: 6679347-6679347 on Build GRCh38
rs768482009 Chr.19: 6679395-6679395 on Build GRCh38
rs369121729 Chr.19: 6679255-6679255 on Build GRCh38
rs927118067 Chr.19: 6679042-6679042 on Build GRCh38
rs779724493 Chr.19: 6679084-6679084 on Build GRCh38
rs972330952 Chr.19: 6678946-6678946 on Build GRCh38
rs377101999 Chr.19: 6679185-6679185 on Build GRCh38
rs749591746 Chr.19: 6679238-6679238 on Build GRCh38
rs563547775 Chr.19: 6678944-6678944 on Build GRCh38
rs7256789 Chr.19: 6679193-6679193 on Build GRCh38
rs780027654 Chr.19: 6679252-6679252 on Build GRCh38
rs544453915 Chr.19: 6678912-6678912 on Build GRCh38
rs199911426 Chr.19: 6679161-6679161 on Build GRCh38
rs781687401 Chr.19: 6679103-6679103 on Build GRCh38

View Full Product Details