Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 14411382-14411740 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX39A
Entrez Gene ID
10212
Gene Name
DEAD-box helicase 39A
Gene Aliases
BAT1, BAT1L, DDX39, DDXL, URH49
UniGene
Hs.311609
Gene Chromosome Location
Chr.19: 14408798-14419383 on Build GRCh38
Transcripts
NM_005804.3 NR_046366.2 XM_006722606.3 XM_011527620.1 XM_011527621.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6314448 c.400C>A p.R134S Chr.19: 14411535-14411535 on Build GRCh38
6316036 c.418C>A p.P140T Chr.19: 14411517-14411517 on Build GRCh38
4859222 c.350T>C p.V117A Chr.19: 14411585-14411585 on Build GRCh38
403734 c.397G>A p.E133K Chr.19: 14411538-14411538 on Build GRCh38
4074803 c.407C>T p.S136F Chr.19: 14411528-14411528 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs142457753 Chr.19: 14411688-14411688 on Build GRCh38
rs1052341673 Chr.19: 14411442-14411442 on Build GRCh38
rs778568165 Chr.19: 14411582-14411582 on Build GRCh38
rs772967062 Chr.19: 14411603-14411603 on Build GRCh38
rs944104534 Chr.19: 14411551-14411551 on Build GRCh38
rs1022201091 Chr.19: 14411683-14411683 on Build GRCh38
rs369868129 Chr.19: 14411495-14411495 on Build GRCh38
rs996046034 Chr.19: 14411683-14411684 on Build GRCh38
rs397787946 Chr.19: 14411628-14411632 on Build GRCh38
rs781689801 Chr.19: 14411563-14411563 on Build GRCh38
rs372373750 Chr.19: 14411445-14411445 on Build GRCh38
rs542691704 Chr.19: 14411655-14411655 on Build GRCh38
rs776395563 Chr.19: 14411618-14411618 on Build GRCh38
rs545574879 Chr.19: 14411581-14411581 on Build GRCh38
rs1018045258 Chr.19: 14411462-14411462 on Build GRCh38
rs750573544 Chr.19: 14411624-14411628 on Build GRCh38
rs758580560 Chr.19: 14411514-14411514 on Build GRCh38
rs981849543 Chr.19: 14411721-14411721 on Build GRCh38
rs991680058 Chr.19: 14411659-14411659 on Build GRCh38
rs371633217 Chr.19: 14411475-14411475 on Build GRCh38
rs770365031 Chr.19: 14411573-14411573 on Build GRCh38
rs766065577 Chr.19: 14411494-14411494 on Build GRCh38
rs746438041 Chr.19: 14411572-14411572 on Build GRCh38
rs149562590 Chr.19: 14411542-14411542 on Build GRCh38
rs752709024 Chr.19: 14411513-14411513 on Build GRCh38
rs748357665 Chr.19: 14411444-14411444 on Build GRCh38
rs962940911 Chr.19: 14411608-14411608 on Build GRCh38
rs759214392 Chr.19: 14411622-14411626 on Build GRCh38
rs777996900 Chr.19: 14411628-14411628 on Build GRCh38
rs752797000 Chr.19: 14411643-14411643 on Build GRCh38
rs55968966 Chr.19: 14411623-14411627 on Build GRCh38
rs751712404 Chr.19: 14411520-14411520 on Build GRCh38
rs968112779 Chr.19: 14411430-14411430 on Build GRCh38
rs899935581 Chr.19: 14411472-14411472 on Build GRCh38
rs977807065 Chr.19: 14411431-14411431 on Build GRCh38
rs572670295 Chr.19: 14411609-14411609 on Build GRCh38
rs947807714 Chr.19: 14411702-14411702 on Build GRCh38
rs375885508 Chr.19: 14411538-14411538 on Build GRCh38
rs999860505 Chr.19: 14411705-14411705 on Build GRCh38
rs948261776 Chr.19: 14411466-14411466 on Build GRCh38
rs778003530 Chr.19: 14411515-14411515 on Build GRCh38
rs758591645 Chr.19: 14411626-14411627 on Build GRCh38
rs1010723320 Chr.19: 14411461-14411461 on Build GRCh38
rs759476302 Chr.19: 14411619-14411619 on Build GRCh38
rs900335815 Chr.19: 14411679-14411679 on Build GRCh38
rs144413770 Chr.19: 14411587-14411587 on Build GRCh38
rs554181705 Chr.19: 14411593-14411593 on Build GRCh38
rs752558943 Chr.19: 14411623-14411626 on Build GRCh38
rs765121123 Chr.19: 14411629-14411629 on Build GRCh38
rs36127505 Chr.19: 14411511-14411511 on Build GRCh38
rs1022261772 Chr.19: 14411403-14411403 on Build GRCh38
rs765315803 Chr.19: 14411512-14411512 on Build GRCh38
rs201367742 Chr.19: 14411548-14411548 on Build GRCh38
rs772908071 Chr.19: 14411488-14411488 on Build GRCh38
rs761568680 Chr.19: 14411460-14411460 on Build GRCh38

View Full Product Details