Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 14632824-14633419 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADGRE3
Entrez Gene ID
84658
Gene Name
adhesion G protein-coupled receptor E3
Gene Aliases
EMR3
UniGene
Hs.658712
Gene Chromosome Location
Chr.19: 14600175-14674918 on Build GRCh38
Transcripts
NM_001289158.1 NM_001289159.1 NM_032571.4 XM_011528374.2 XM_017027383.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5897995 c.1531C>T p.P511S Chr.19: 14633256-14633256 on Build GRCh38
4467515 c.1494C>T p.L498L Chr.19: 14633293-14633293 on Build GRCh38
6465590 c.1602T>C p.L534L Chr.19: 14632962-14632962 on Build GRCh38
6465591 c.1601T>C p.L534P Chr.19: 14632963-14632963 on Build GRCh38
4528044 c.1507G>A p.G503R Chr.19: 14633280-14633280 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs148098184 Chr.19: 14633118-14633118 on Build GRCh38
rs951555556 Chr.19: 14633162-14633162 on Build GRCh38
rs996985199 Chr.19: 14633316-14633316 on Build GRCh38
rs763943954 Chr.19: 14633063-14633063 on Build GRCh38
rs762678750 Chr.19: 14633061-14633061 on Build GRCh38
rs1013692765 Chr.19: 14633059-14633059 on Build GRCh38
rs138384322 Chr.19: 14632979-14632979 on Build GRCh38
rs543225515 Chr.19: 14633034-14633034 on Build GRCh38
rs892172518 Chr.19: 14632868-14632868 on Build GRCh38
rs899217 Chr.19: 14633369-14633369 on Build GRCh38
rs935169469 Chr.19: 14633346-14633346 on Build GRCh38
rs752513062 Chr.19: 14633037-14633037 on Build GRCh38
rs1022080015 Chr.19: 14632909-14632909 on Build GRCh38
rs753823725 Chr.19: 14633290-14633290 on Build GRCh38
rs760169835 Chr.19: 14632951-14632951 on Build GRCh38
rs926528031 Chr.19: 14633113-14633113 on Build GRCh38
rs781326971 Chr.19: 14632998-14632998 on Build GRCh38
rs781299547 Chr.19: 14633196-14633196 on Build GRCh38
rs969091173 Chr.19: 14632846-14632846 on Build GRCh38
rs538668516 Chr.19: 14632850-14632850 on Build GRCh38
rs776152429 Chr.19: 14633312-14633312 on Build GRCh38
rs763926645 Chr.19: 14633338-14633338 on Build GRCh38
rs763522270 Chr.19: 14632897-14632897 on Build GRCh38
rs150182348 Chr.19: 14633350-14633350 on Build GRCh38
rs113130166 Chr.19: 14633234-14633234 on Build GRCh38
rs769451728 Chr.19: 14633325-14633325 on Build GRCh38
rs762724694 Chr.19: 14633336-14633336 on Build GRCh38
rs975422775 Chr.19: 14632907-14632907 on Build GRCh38
rs371566047 Chr.19: 14633327-14633327 on Build GRCh38
rs769242670 Chr.19: 14632915-14632915 on Build GRCh38
rs968229331 Chr.19: 14632918-14632918 on Build GRCh38
rs982995988 Chr.19: 14632884-14632884 on Build GRCh38
rs377132894 Chr.19: 14633058-14633058 on Build GRCh38
rs554746691 Chr.19: 14633015-14633015 on Build GRCh38
rs770464933 Chr.19: 14633019-14633019 on Build GRCh38
rs773045534 Chr.19: 14633067-14633067 on Build GRCh38
rs995212597 Chr.19: 14633121-14633121 on Build GRCh38
rs769054109 Chr.19: 14633011-14633011 on Build GRCh38
rs775550790 Chr.19: 14633220-14633238 on Build GRCh38
rs764996847 Chr.19: 14632970-14632970 on Build GRCh38
rs373442511 Chr.19: 14633232-14633232 on Build GRCh38
rs764830342 Chr.19: 14633035-14633035 on Build GRCh38
rs553905155 Chr.19: 14632953-14632953 on Build GRCh38
rs529453654 Chr.19: 14633192-14633192 on Build GRCh38
rs7249008 Chr.19: 14633130-14633130 on Build GRCh38
rs759388525 Chr.19: 14632964-14632964 on Build GRCh38
rs749669974 Chr.19: 14633226-14633226 on Build GRCh38
rs968667510 Chr.19: 14633323-14633323 on Build GRCh38
rs373143666 Chr.19: 14632988-14632988 on Build GRCh38
rs564990076 Chr.19: 14633219-14633219 on Build GRCh38
rs924661994 Chr.19: 14633342-14633342 on Build GRCh38
rs370790081 Chr.19: 14633349-14633349 on Build GRCh38
rs2045912 Chr.19: 14633090-14633090 on Build GRCh38
rs997434813 Chr.19: 14632933-14632933 on Build GRCh38
rs142461083 Chr.19: 14632996-14632996 on Build GRCh38
rs770320312 Chr.19: 14633004-14633004 on Build GRCh38
rs751604025 Chr.19: 14632990-14632990 on Build GRCh38
rs776318881 Chr.19: 14633030-14633030 on Build GRCh38
rs748670693 Chr.19: 14633285-14633285 on Build GRCh38
rs947445050 Chr.19: 14632963-14632963 on Build GRCh38
rs1056435653 Chr.19: 14633143-14633143 on Build GRCh38
rs1027101142 Chr.19: 14632851-14632851 on Build GRCh38
rs745334587 Chr.19: 14633319-14633319 on Build GRCh38
rs776186924 Chr.19: 14632891-14632891 on Build GRCh38
rs749618477 Chr.19: 14633010-14633010 on Build GRCh38
rs560321303 Chr.19: 14632950-14632950 on Build GRCh38
rs921379037 Chr.19: 14632917-14632917 on Build GRCh38
rs751560347 Chr.19: 14633383-14633383 on Build GRCh38
rs376096757 Chr.19: 14633243-14633243 on Build GRCh38
rs1004362285 Chr.19: 14633203-14633203 on Build GRCh38
rs147058520 Chr.19: 14633062-14633062 on Build GRCh38
rs772617742 Chr.19: 14632937-14632937 on Build GRCh38
rs113412752 Chr.19: 14633330-14633330 on Build GRCh38
rs951669844 Chr.19: 14632866-14632866 on Build GRCh38
rs750489395 Chr.19: 14633217-14633217 on Build GRCh38
rs865838192 Chr.19: 14633263-14633263 on Build GRCh38
rs763168696 Chr.19: 14633230-14633233 on Build GRCh38
rs533852689 Chr.19: 14632957-14632957 on Build GRCh38
rs770390654 Chr.19: 14633309-14633309 on Build GRCh38
rs751500719 Chr.19: 14633064-14633064 on Build GRCh38
rs732722 Chr.19: 14633220-14633220 on Build GRCh38
rs779350546 Chr.19: 14633261-14633261 on Build GRCh38

View Full Product Details