Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 35498579-35499171 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DMKN
Entrez Gene ID
93099
Gene Name
dermokine
Gene Aliases
UNQ729, ZD52F10
UniGene
Hs.417795
Gene Chromosome Location
Chr.19: 35497217-35513678 on Build GRCh38
Transcripts
NM_001035516.3 NM_001126056.2 NM_001126059.2 NM_001190347.1 NM_001308380.1 NM_001308383.1 NM_033317.4 NR_033746.1 XM_006723477.1 XM_006723484.1 XM_006723489.1 XM_006723493.2 XM_006723494.2 XM_006723503.2 XM_011527494.2 XM_011527495.2 XM_011527496.2 XM_011527497.2 XM_011527498.2 XM_011527499.2 XM_011527500.2 XM_011527501.2 XM_011527502.2 XM_011527503.2 XM_011527504.2 XM_011527505.2 XM_011527506.2 XM_011527507.2 XM_011527508.2 XM_011527509.2 XM_011527510.2 XM_011527513.1 XM_011527514.1 XM_017027475.1 XM_017027476.1 XM_017027477.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4336511 c.1283delG p.G428fs*19 Chr.19: 35498893-35498893 on Build GRCh38
6885786 c.1380G>A p.Q460Q Chr.19: 35498734-35498734 on Build GRCh38
5056540 c.233G>A p.R78Q Chr.19: 35498756-35498756 on Build GRCh38
6003589 c.1340A>T p.K447M Chr.19: 35498726-35498726 on Build GRCh38
6003590 c.560A>T p.K187M Chr.19: 35498726-35498726 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs578028672 Chr.19: 35498657-35498657 on Build GRCh38
rs781635477 Chr.19: 35498814-35498814 on Build GRCh38
rs752915737 Chr.19: 35498793-35498793 on Build GRCh38
rs778215902 Chr.19: 35498854-35498854 on Build GRCh38
rs748931721 Chr.19: 35498840-35498840 on Build GRCh38
rs368794417 Chr.19: 35498932-35498932 on Build GRCh38
rs900394351 Chr.19: 35499020-35499020 on Build GRCh38
rs764267042 Chr.19: 35498896-35498896 on Build GRCh38
rs749735815 Chr.19: 35498861-35498861 on Build GRCh38
rs568720181 Chr.19: 35498977-35498977 on Build GRCh38
rs140257981 Chr.19: 35499042-35499042 on Build GRCh38
rs768276508 Chr.19: 35498751-35498751 on Build GRCh38
rs184563318 Chr.19: 35498898-35498898 on Build GRCh38
rs1036016487 Chr.19: 35498787-35498787 on Build GRCh38
rs1048167617 Chr.19: 35499013-35499013 on Build GRCh38
rs543664793 Chr.19: 35498682-35498682 on Build GRCh38
rs142000561 Chr.19: 35498897-35498897 on Build GRCh38
rs564520372 Chr.19: 35498638-35498638 on Build GRCh38
rs2293693 Chr.19: 35499070-35499070 on Build GRCh38
rs369692635 Chr.19: 35498756-35498756 on Build GRCh38
rs997424261 Chr.19: 35499023-35499023 on Build GRCh38
rs34509229 Chr.19: 35498802-35498803 on Build GRCh38
rs1039686923 Chr.19: 35498602-35498602 on Build GRCh38
rs746266009 Chr.19: 35498815-35498815 on Build GRCh38
rs753544971 Chr.19: 35498935-35498935 on Build GRCh38
rs1006170571 Chr.19: 35498633-35498633 on Build GRCh38
rs758445247 Chr.19: 35498681-35498681 on Build GRCh38
rs754635250 Chr.19: 35498937-35498937 on Build GRCh38
rs777029827 Chr.19: 35498947-35498947 on Build GRCh38
rs923483447 Chr.19: 35498979-35498979 on Build GRCh38
rs537352785 Chr.19: 35499133-35499133 on Build GRCh38
rs942119957 Chr.19: 35499053-35499053 on Build GRCh38
rs780222714 Chr.19: 35498816-35498816 on Build GRCh38
rs778506275 Chr.19: 35498944-35498944 on Build GRCh38
rs752827490 Chr.19: 35498671-35498671 on Build GRCh38
rs369467672 Chr.19: 35498875-35498875 on Build GRCh38
rs1029697201 Chr.19: 35499054-35499054 on Build GRCh38
rs202112325 Chr.19: 35498923-35498923 on Build GRCh38
rs774726109 Chr.19: 35499068-35499068 on Build GRCh38
rs201569702 Chr.19: 35498719-35498719 on Build GRCh38
rs756515969 Chr.19: 35498830-35498830 on Build GRCh38
rs895695041 Chr.19: 35499140-35499140 on Build GRCh38
rs779918983 Chr.19: 35498832-35498832 on Build GRCh38
rs1006434408 Chr.19: 35498978-35498978 on Build GRCh38
rs775702761 Chr.19: 35498876-35498876 on Build GRCh38
rs559409693 Chr.19: 35498603-35498603 on Build GRCh38
rs765487589 Chr.19: 35498792-35498792 on Build GRCh38
rs887616566 Chr.19: 35498952-35498952 on Build GRCh38
rs761594834 Chr.19: 35498757-35498757 on Build GRCh38
rs765015666 Chr.19: 35498784-35498785 on Build GRCh38
rs916113019 Chr.19: 35498822-35498822 on Build GRCh38
rs111882428 Chr.19: 35499139-35499139 on Build GRCh38
rs768290760 Chr.19: 35498848-35498848 on Build GRCh38
rs375350843 Chr.19: 35498697-35498697 on Build GRCh38
rs375476987 Chr.19: 35498812-35498812 on Build GRCh38
rs545562159 Chr.19: 35498836-35498836 on Build GRCh38
rs760534903 Chr.19: 35498874-35498874 on Build GRCh38
rs771954574 Chr.19: 35498862-35498862 on Build GRCh38
rs374185829 Chr.19: 35498776-35498776 on Build GRCh38
rs1038965074 Chr.19: 35499001-35499001 on Build GRCh38
rs959737752 Chr.19: 35498791-35498791 on Build GRCh38
rs550847808 Chr.19: 35499138-35499138 on Build GRCh38
rs748846245 Chr.19: 35498741-35498741 on Build GRCh38
rs769520500 Chr.19: 35498731-35498731 on Build GRCh38
rs763096533 Chr.19: 35498883-35498883 on Build GRCh38
rs948425059 Chr.19: 35498866-35498866 on Build GRCh38
rs767346282 Chr.19: 35498761-35498761 on Build GRCh38
rs745567998 Chr.19: 35498728-35498728 on Build GRCh38
rs764025438 Chr.19: 35498893-35498893 on Build GRCh38
rs773160478 Chr.19: 35498878-35498880 on Build GRCh38
rs758253239 Chr.19: 35498892-35498893 on Build GRCh38
rs756604194 Chr.19: 35498927-35498927 on Build GRCh38
rs867609957 Chr.19: 35498658-35498658 on Build GRCh38
rs773173663 Chr.19: 35498870-35498870 on Build GRCh38
rs775260075 Chr.19: 35498733-35498733 on Build GRCh38
rs757655167 Chr.19: 35498714-35498714 on Build GRCh38
rs752112054 Chr.19: 35498807-35498807 on Build GRCh38
rs901881956 Chr.19: 35498631-35498632 on Build GRCh38
rs758531197 Chr.19: 35498801-35498801 on Build GRCh38
rs1047469017 Chr.19: 35498926-35498926 on Build GRCh38
rs199524471 Chr.19: 35498762-35498762 on Build GRCh38
rs759098592 Chr.19: 35498673-35498674 on Build GRCh38
rs998978430 Chr.19: 35499125-35499125 on Build GRCh38
rs765325835 Chr.19: 35498666-35498666 on Build GRCh38
rs192954904 Chr.19: 35498696-35498696 on Build GRCh38
rs1029765737 Chr.19: 35498685-35498685 on Build GRCh38
rs905736901 Chr.19: 35499069-35499069 on Build GRCh38
rs764103924 Chr.19: 35498805-35498805 on Build GRCh38
rs541540517 Chr.19: 35499048-35499048 on Build GRCh38

View Full Product Details