Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 55591542-55592051 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FIZ1
Entrez Gene ID
84922
Gene Name
FLT3 interacting zinc finger 1
Gene Aliases
ZNF798
UniGene
Hs.515617
Gene Chromosome Location
Chr.19: 55591371-55599703 on Build GRCh38
Transcripts
NM_032836.2 XM_005259352.4 XM_011527426.2

SNP Detail

SNP ID Chromosome Location
rs964040310 Chr.19: 55591715-55591716 on Build GRCh38
rs912223552 Chr.19: 55591590-55591590 on Build GRCh38
rs964086291 Chr.19: 55592013-55592013 on Build GRCh38
rs907236020 Chr.19: 55591869-55591869 on Build GRCh38
rs1019894574 Chr.19: 55592016-55592016 on Build GRCh38
rs866074938 Chr.19: 55591679-55591679 on Build GRCh38
rs545994201 Chr.19: 55592012-55592012 on Build GRCh38
rs572345419 Chr.19: 55591772-55591772 on Build GRCh38
rs995609871 Chr.19: 55591942-55591942 on Build GRCh38
rs559442864 Chr.19: 55591706-55591706 on Build GRCh38
rs1052865482 Chr.19: 55591980-55591980 on Build GRCh38
rs1052284549 Chr.19: 55591790-55591790 on Build GRCh38
rs569854100 Chr.19: 55591817-55591817 on Build GRCh38
rs952933515 Chr.19: 55591765-55591765 on Build GRCh38
rs1027082853 Chr.19: 55591757-55591757 on Build GRCh38
rs867986076 Chr.19: 55591607-55591607 on Build GRCh38
rs951599815 Chr.19: 55591987-55591987 on Build GRCh38
rs184739562 Chr.19: 55591627-55591627 on Build GRCh38
rs145689282 Chr.19: 55591813-55591813 on Build GRCh38
rs369884337 Chr.19: 55591740-55591740 on Build GRCh38
rs975385806 Chr.19: 55591733-55591733 on Build GRCh38
rs892632105 Chr.19: 55591660-55591660 on Build GRCh38
rs74622788 Chr.19: 55591934-55591934 on Build GRCh38
rs1008517966 Chr.19: 55591661-55591661 on Build GRCh38
rs1034876346 Chr.19: 55591924-55591924 on Build GRCh38
rs948405118 Chr.19: 55591848-55591848 on Build GRCh38
rs753815878 Chr.19: 55591624-55591624 on Build GRCh38
rs375787649 Chr.19: 55591971-55591971 on Build GRCh38
rs975179519 Chr.19: 55592022-55592022 on Build GRCh38
rs975609808 Chr.19: 55591644-55591644 on Build GRCh38
rs1001617875 Chr.19: 55591917-55591917 on Build GRCh38
rs1019841926 Chr.19: 55591678-55591678 on Build GRCh38
rs892357485 Chr.19: 55591797-55591797 on Build GRCh38
rs925135217 Chr.19: 55591693-55591693 on Build GRCh38
rs1025781912 Chr.19: 55591986-55591986 on Build GRCh38
rs1042733951 Chr.19: 55591852-55591852 on Build GRCh38
rs1031610011 Chr.19: 55591652-55591652 on Build GRCh38
rs115512498 Chr.19: 55592009-55592009 on Build GRCh38
rs992707456 Chr.19: 55591799-55591799 on Build GRCh38
rs938497190 Chr.19: 55591794-55591797 on Build GRCh38
rs1045722227 Chr.19: 55591839-55591839 on Build GRCh38
rs528175993 Chr.19: 55591806-55591806 on Build GRCh38

View Full Product Details