Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 74983703-74984299 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C9orf41-AS1
Entrez Gene ID
101927380
Gene Name
C9orf41 antisense RNA 1
Gene Aliases
-
UniGene
Hs.257556
Gene Chromosome Location
Chr.9: 74952965-74997382 on Build GRCh38
Transcripts
NR_121183.1 NM_001320497.1 NM_152420.2 NR_135282.1 XM_005251701.3 XM_017014269.1 XM_017014270.1 XM_017014271.1

Gene Symbol
CARNMT1
Entrez Gene ID
138199
Gene Name
carnosine N-methyltransferase 1
Gene Aliases
C9orf41, UPF0586
UniGene
Hs.567688
Gene Chromosome Location
Chr.9: 74980794-75028456 on Build GRCh38
Transcripts
NR_121183.1 NM_001320497.1 NM_152420.2 NR_135282.1 XM_005251701.3 XM_017014269.1 XM_017014270.1 XM_017014271.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5441452 c.1216C>T p.R406C Chr.9: 74983781-74983781 on Build GRCh38
4451355 c.1177A>C p.M393L Chr.9: 74983820-74983820 on Build GRCh38
5517142 c.1154C>A p.T385K Chr.9: 74983843-74983843 on Build GRCh38
6473230 c.1206G>T p.L402F Chr.9: 74983791-74983791 on Build GRCh38
6616958 c.1174T>G p.S392A Chr.9: 74983823-74983823 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs138458998 Chr.9: 74983985-74983985 on Build GRCh38
rs898240189 Chr.9: 74984013-74984013 on Build GRCh38
rs768644505 Chr.9: 74983849-74983849 on Build GRCh38
rs150086282 Chr.9: 74983893-74983893 on Build GRCh38
rs981748364 Chr.9: 74984119-74984119 on Build GRCh38
rs577616693 Chr.9: 74984024-74984024 on Build GRCh38
rs778727873 Chr.9: 74983823-74983823 on Build GRCh38
rs752072330 Chr.9: 74983787-74983787 on Build GRCh38
rs779606276 Chr.9: 74983838-74983838 on Build GRCh38
rs888277243 Chr.9: 74984143-74984143 on Build GRCh38
rs936972672 Chr.9: 74984105-74984105 on Build GRCh38
rs773375247 Chr.9: 74983771-74983771 on Build GRCh38
rs1003892230 Chr.9: 74983989-74983989 on Build GRCh38
rs779956834 Chr.9: 74984158-74984158 on Build GRCh38
rs900519452 Chr.9: 74984209-74984209 on Build GRCh38
rs375158217 Chr.9: 74983920-74983920 on Build GRCh38
rs774502792 Chr.9: 74983908-74983908 on Build GRCh38
rs746279507 Chr.9: 74984271-74984271 on Build GRCh38
rs762086955 Chr.9: 74983811-74983811 on Build GRCh38
rs996603476 Chr.9: 74984217-74984217 on Build GRCh38
rs370186141 Chr.9: 74983764-74983764 on Build GRCh38
rs556841337 Chr.9: 74984157-74984157 on Build GRCh38
rs760073875 Chr.9: 74983793-74983793 on Build GRCh38
rs781069455 Chr.9: 74983876-74983876 on Build GRCh38
rs768397946 Chr.9: 74983734-74983734 on Build GRCh38
rs954966182 Chr.9: 74983939-74983939 on Build GRCh38
rs759463559 Chr.9: 74983910-74983910 on Build GRCh38
rs1037761422 Chr.9: 74984159-74984159 on Build GRCh38
rs767308536 Chr.9: 74983733-74983733 on Build GRCh38
rs112830265 Chr.9: 74984118-74984118 on Build GRCh38
rs554853949 Chr.9: 74984125-74984125 on Build GRCh38
rs757916216 Chr.9: 74983836-74983836 on Build GRCh38
rs1012718975 Chr.9: 74984023-74984023 on Build GRCh38
rs752472108 Chr.9: 74983794-74983794 on Build GRCh38
rs767849997 Chr.9: 74983803-74983803 on Build GRCh38
rs747961734 Chr.9: 74983885-74983885 on Build GRCh38
rs113544239 Chr.9: 74983934-74983934 on Build GRCh38
rs770973375 Chr.9: 74983906-74983906 on Build GRCh38
rs1051369105 Chr.9: 74983965-74983965 on Build GRCh38
rs756503848 Chr.9: 74983821-74983821 on Build GRCh38
rs768166012 Chr.9: 74983918-74983918 on Build GRCh38
rs750072371 Chr.9: 74983824-74983824 on Build GRCh38
rs934234188 Chr.9: 74983960-74983960 on Build GRCh38
rs1030321989 Chr.9: 74984228-74984229 on Build GRCh38
rs117142304 Chr.9: 74984108-74984108 on Build GRCh38
rs746450155 Chr.9: 74983842-74983842 on Build GRCh38
rs975563116 Chr.9: 74984241-74984241 on Build GRCh38
rs769556901 Chr.9: 74983888-74983888 on Build GRCh38
rs752111772 Chr.9: 74984248-74984248 on Build GRCh38
rs935358185 Chr.9: 74983860-74983860 on Build GRCh38
rs946782331 Chr.9: 74984178-74984178 on Build GRCh38
rs962507189 Chr.9: 74983756-74983756 on Build GRCh38
rs921414733 Chr.9: 74984265-74984265 on Build GRCh38
rs1052458421 Chr.9: 74983875-74983875 on Build GRCh38
rs769848575 Chr.9: 74983795-74983796 on Build GRCh38
rs1057063962 Chr.9: 74984106-74984106 on Build GRCh38
rs770036060 Chr.9: 74983757-74983757 on Build GRCh38
rs762989606 Chr.9: 74983779-74983779 on Build GRCh38
rs1010443587 Chr.9: 74984267-74984267 on Build GRCh38
rs765411795 Chr.9: 74983943-74983943 on Build GRCh38
rs368315108 Chr.9: 74983745-74983745 on Build GRCh38
rs541086681 Chr.9: 74984107-74984107 on Build GRCh38
rs900031777 Chr.9: 74983951-74983951 on Build GRCh38
rs929896356 Chr.9: 74984109-74984109 on Build GRCh38
rs1016668567 Chr.9: 74983995-74983995 on Build GRCh38
rs753112350 Chr.9: 74983809-74983809 on Build GRCh38
rs988121202 Chr.9: 74984140-74984140 on Build GRCh38
rs576172618 Chr.9: 74984124-74984124 on Build GRCh38
rs1016719276 Chr.9: 74983744-74983744 on Build GRCh38
rs1040283104 Chr.9: 74983948-74983948 on Build GRCh38
rs757608908 Chr.9: 74984033-74984033 on Build GRCh38

View Full Product Details