Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 124878498-124879085 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GOLGA1
Entrez Gene ID
2800
Gene Name
golgin A1
Gene Aliases
golgin-97
UniGene
Hs.133469
Gene Chromosome Location
Chr.9: 124878294-124941114 on Build GRCh38
Transcripts
NM_002077.3 XM_005251929.3 XM_006717062.3 XM_006717063.3

SNP Detail

SNP ID Chromosome Location
rs1014158516 Chr.9: 124879042-124879042 on Build GRCh38
rs144348004 Chr.9: 124878713-124878713 on Build GRCh38
rs752059747 Chr.9: 124878561-124878561 on Build GRCh38
rs565654941 Chr.9: 124878928-124878928 on Build GRCh38
rs917103515 Chr.9: 124878804-124878804 on Build GRCh38
rs532548983 Chr.9: 124878664-124878664 on Build GRCh38
rs144077576 Chr.9: 124878699-124878700 on Build GRCh38
rs534742350 Chr.9: 124878970-124878970 on Build GRCh38
rs1054197642 Chr.9: 124879004-124879004 on Build GRCh38
rs867814092 Chr.9: 124878742-124878742 on Build GRCh38
rs938022974 Chr.9: 124878990-124878990 on Build GRCh38
rs972196307 Chr.9: 124878915-124878915 on Build GRCh38
rs760169240 Chr.9: 124878760-124878760 on Build GRCh38
rs187394708 Chr.9: 124879036-124879036 on Build GRCh38
rs770701880 Chr.9: 124878912-124878912 on Build GRCh38
rs919442499 Chr.9: 124878925-124878925 on Build GRCh38
rs992683537 Chr.9: 124878550-124878550 on Build GRCh38
rs1028002983 Chr.9: 124878590-124878590 on Build GRCh38
rs982204643 Chr.9: 124878967-124878967 on Build GRCh38
rs958778126 Chr.9: 124878525-124878525 on Build GRCh38
rs926733693 Chr.9: 124878988-124878988 on Build GRCh38
rs1016515858 Chr.9: 124878898-124878898 on Build GRCh38
rs749475915 Chr.9: 124878850-124878850 on Build GRCh38
rs993047330 Chr.9: 124878787-124878787 on Build GRCh38
rs1000159124 Chr.9: 124879024-124879024 on Build GRCh38
rs941054635 Chr.9: 124878674-124878674 on Build GRCh38
rs953727325 Chr.9: 124878824-124878824 on Build GRCh38
rs1051939115 Chr.9: 124878706-124878706 on Build GRCh38
rs574529057 Chr.9: 124878584-124878584 on Build GRCh38
rs558630214 Chr.9: 124878910-124878910 on Build GRCh38
rs528592560 Chr.9: 124878692-124878692 on Build GRCh38
rs1037914172 Chr.9: 124878681-124878681 on Build GRCh38
rs759176237 Chr.9: 124879028-124879028 on Build GRCh38
rs548263436 Chr.9: 124878709-124878709 on Build GRCh38
rs1042179644 Chr.9: 124879049-124879049 on Build GRCh38
rs2781055 Chr.9: 124878759-124878759 on Build GRCh38
rs571015597 Chr.9: 124878789-124878789 on Build GRCh38
rs1042211315 Chr.9: 124878756-124878756 on Build GRCh38
rs904618025 Chr.9: 124879020-124879020 on Build GRCh38
rs953735159 Chr.9: 124878621-124878621 on Build GRCh38
rs544447816 Chr.9: 124878908-124878908 on Build GRCh38
rs909674806 Chr.9: 124878876-124878876 on Build GRCh38
rs545836405 Chr.9: 124878665-124878665 on Build GRCh38
rs958653002 Chr.9: 124878778-124878778 on Build GRCh38
rs900795160 Chr.9: 124878699-124878699 on Build GRCh38
rs938038331 Chr.9: 124878746-124878746 on Build GRCh38
rs565039053 Chr.9: 124878673-124878673 on Build GRCh38
rs1005667141 Chr.9: 124878888-124878888 on Build GRCh38
rs916948079 Chr.9: 124878586-124878586 on Build GRCh38
rs540127802 Chr.9: 124878816-124878816 on Build GRCh38
rs574579832 Chr.9: 124879006-124879006 on Build GRCh38
rs778122312 Chr.9: 124878812-124878812 on Build GRCh38
rs1024664872 Chr.9: 124879045-124879045 on Build GRCh38
rs778711790 Chr.9: 124879030-124879030 on Build GRCh38
rs112061540 Chr.9: 124878654-124878654 on Build GRCh38
rs926623680 Chr.9: 124878740-124878740 on Build GRCh38
rs755569441 Chr.9: 124878725-124878725 on Build GRCh38
rs970906437 Chr.9: 124878939-124878939 on Build GRCh38
rs982476295 Chr.9: 124878836-124878841 on Build GRCh38
rs183460974 Chr.9: 124879005-124879005 on Build GRCh38
rs893763070 Chr.9: 124878754-124878754 on Build GRCh38
rs530983991 Chr.9: 124878730-124878730 on Build GRCh38
rs531110416 Chr.9: 124878979-124878981 on Build GRCh38
rs867358990 Chr.9: 124879016-124879016 on Build GRCh38
rs1028515446 Chr.9: 124878805-124878805 on Build GRCh38
rs543713749 Chr.9: 124878611-124878611 on Build GRCh38
rs530734854 Chr.9: 124878806-124878806 on Build GRCh38
rs1009315179 Chr.9: 124878755-124878755 on Build GRCh38
rs752871075 Chr.9: 124878747-124878747 on Build GRCh38
rs997963834 Chr.9: 124878779-124878779 on Build GRCh38
rs1046803481 Chr.9: 124878734-124878734 on Build GRCh38
rs776157544 Chr.9: 124878917-124878917 on Build GRCh38
rs748454386 Chr.9: 124878865-124878866 on Build GRCh38
rs900995923 Chr.9: 124879052-124879052 on Build GRCh38
rs559321972 Chr.9: 124878678-124878678 on Build GRCh38

View Full Product Details