Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 50309218-50309810 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCNB3
Entrez Gene ID
85417
Gene Name
cyclin B3
Gene Aliases
-
UniGene
Hs.130310
Gene Chromosome Location
Chr.X: 50202713-50351914 on Build GRCh38
Transcripts
NM_033031.2 NM_033670.2 XM_017029913.1 XM_017029914.1 XM_017029915.1 XM_017029916.1 XM_017029917.1 XM_017029918.1

SNP Detail

SNP ID Chromosome Location
rs782488987 Chr.X: 50309727-50309727 on Build GRCh38
rs782552876 Chr.X: 50309255-50309255 on Build GRCh38
rs531559311 Chr.X: 50309770-50309770 on Build GRCh38
rs782756885 Chr.X: 50309446-50309446 on Build GRCh38
rs782163522 Chr.X: 50309696-50309696 on Build GRCh38
rs781817587 Chr.X: 50309447-50309447 on Build GRCh38
rs782744756 Chr.X: 50309728-50309728 on Build GRCh38
rs782207060 Chr.X: 50309614-50309614 on Build GRCh38
rs902569645 Chr.X: 50309665-50309665 on Build GRCh38
rs781790506 Chr.X: 50309397-50309397 on Build GRCh38
rs143042560 Chr.X: 50309737-50309737 on Build GRCh38
rs781966723 Chr.X: 50309341-50309341 on Build GRCh38
rs369716053 Chr.X: 50309534-50309534 on Build GRCh38
rs376736828 Chr.X: 50309513-50309513 on Build GRCh38
rs781892305 Chr.X: 50309688-50309688 on Build GRCh38
rs782260477 Chr.X: 50309318-50309318 on Build GRCh38
rs200972903 Chr.X: 50309590-50309590 on Build GRCh38
rs782801587 Chr.X: 50309399-50309399 on Build GRCh38
rs781873338 Chr.X: 50309735-50309735 on Build GRCh38
rs927012458 Chr.X: 50309507-50309507 on Build GRCh38
rs782570615 Chr.X: 50309266-50309266 on Build GRCh38
rs372451442 Chr.X: 50309359-50309359 on Build GRCh38
rs782796186 Chr.X: 50309705-50309705 on Build GRCh38
rs782371565 Chr.X: 50309320-50309320 on Build GRCh38
rs781960096 Chr.X: 50309632-50309632 on Build GRCh38
rs782301242 Chr.X: 50309646-50309646 on Build GRCh38
rs782027977 Chr.X: 50309604-50309604 on Build GRCh38
rs782039469 Chr.X: 50309441-50309441 on Build GRCh38
rs782518239 Chr.X: 50309495-50309495 on Build GRCh38
rs781906156 Chr.X: 50309253-50309253 on Build GRCh38
rs782172227 Chr.X: 50309398-50309398 on Build GRCh38
rs782246616 Chr.X: 50309563-50309563 on Build GRCh38
rs782662713 Chr.X: 50309562-50309562 on Build GRCh38
rs375406438 Chr.X: 50309471-50309471 on Build GRCh38
rs782764236 Chr.X: 50309658-50309658 on Build GRCh38
rs782534876 Chr.X: 50309515-50309515 on Build GRCh38
rs782396014 Chr.X: 50309358-50309358 on Build GRCh38
rs201480633 Chr.X: 50309267-50309267 on Build GRCh38
rs782761363 Chr.X: 50309500-50309500 on Build GRCh38
rs781946058 Chr.X: 50309404-50309404 on Build GRCh38
rs1007536169 Chr.X: 50309760-50309760 on Build GRCh38
rs200489365 Chr.X: 50309333-50309333 on Build GRCh38
rs782597059 Chr.X: 50309282-50309282 on Build GRCh38
rs782667233 Chr.X: 50309703-50309703 on Build GRCh38
rs782214588 Chr.X: 50309350-50309350 on Build GRCh38
rs782561925 Chr.X: 50309524-50309524 on Build GRCh38
rs782482934 Chr.X: 50309268-50309268 on Build GRCh38
rs782076860 Chr.X: 50309656-50309656 on Build GRCh38
rs375109537 Chr.X: 50309618-50309618 on Build GRCh38
rs782186514 Chr.X: 50309405-50309405 on Build GRCh38
rs782227314 Chr.X: 50309533-50309533 on Build GRCh38
rs781804199 Chr.X: 50309712-50309712 on Build GRCh38
rs972916400 Chr.X: 50309296-50309296 on Build GRCh38
rs199624438 Chr.X: 50309345-50309345 on Build GRCh38
rs893059963 Chr.X: 50309576-50309576 on Build GRCh38

View Full Product Details