Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 30559252-30559848 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CXorf21
Entrez Gene ID
80231
Gene Name
chromosome X open reading frame 21
Gene Aliases
-
UniGene
Hs.665009
Gene Chromosome Location
Chr.X: 30558824-30577916 on Build GRCh38
Transcripts
NM_025159.2

SNP Detail

SNP ID Chromosome Location
rs867518231 Chr.X: 30559415-30559415 on Build GRCh38
rs892502392 Chr.X: 30559795-30559795 on Build GRCh38
rs963341018 Chr.X: 30559686-30559686 on Build GRCh38
rs745999065 Chr.X: 30559743-30559743 on Build GRCh38
rs781054658 Chr.X: 30559716-30559716 on Build GRCh38
rs764811432 Chr.X: 30559402-30559402 on Build GRCh38
rs139726582 Chr.X: 30559669-30559669 on Build GRCh38
rs772162280 Chr.X: 30559751-30559751 on Build GRCh38
rs776798262 Chr.X: 30559611-30559611 on Build GRCh38
rs188451154 Chr.X: 30559425-30559425 on Build GRCh38
rs747880237 Chr.X: 30559728-30559728 on Build GRCh38
rs980974253 Chr.X: 30559368-30559368 on Build GRCh38
rs375928267 Chr.X: 30559540-30559540 on Build GRCh38
rs929032324 Chr.X: 30559464-30559464 on Build GRCh38
rs1051534138 Chr.X: 30559486-30559486 on Build GRCh38
rs1045431408 Chr.X: 30559367-30559367 on Build GRCh38
rs749987055 Chr.X: 30559632-30559632 on Build GRCh38
rs762083306 Chr.X: 30559814-30559814 on Build GRCh38
rs777114441 Chr.X: 30559808-30559808 on Build GRCh38
rs751575628 Chr.X: 30559668-30559668 on Build GRCh38
rs144775134 Chr.X: 30559630-30559630 on Build GRCh38
rs749847301 Chr.X: 30559405-30559405 on Build GRCh38
rs1026213052 Chr.X: 30559750-30559750 on Build GRCh38
rs778242950 Chr.X: 30559429-30559429 on Build GRCh38
rs950582378 Chr.X: 30559771-30559771 on Build GRCh38
rs745933194 Chr.X: 30559538-30559538 on Build GRCh38
rs868775173 Chr.X: 30559792-30559792 on Build GRCh38
rs761123616 Chr.X: 30559597-30559597 on Build GRCh38
rs149817919 Chr.X: 30559535-30559535 on Build GRCh38
rs918053470 Chr.X: 30559409-30559409 on Build GRCh38
rs1051544175 Chr.X: 30559515-30559515 on Build GRCh38
rs764476691 Chr.X: 30559605-30559605 on Build GRCh38
rs936888670 Chr.X: 30559393-30559393 on Build GRCh38
rs780069492 Chr.X: 30559752-30559752 on Build GRCh38
rs887369 Chr.X: 30559729-30559729 on Build GRCh38
rs747522617 Chr.X: 30559796-30559796 on Build GRCh38
rs770999789 Chr.X: 30559469-30559469 on Build GRCh38
rs111847753 Chr.X: 30559658-30559658 on Build GRCh38
rs765935213 Chr.X: 30559638-30559638 on Build GRCh38
rs371704109 Chr.X: 30559426-30559426 on Build GRCh38
rs370176469 Chr.X: 30559573-30559573 on Build GRCh38
rs374413778 Chr.X: 30559623-30559623 on Build GRCh38
rs747370788 Chr.X: 30559499-30559499 on Build GRCh38
rs749530592 Chr.X: 30559457-30559457 on Build GRCh38
rs1009586625 Chr.X: 30559807-30559807 on Build GRCh38
rs1017655976 Chr.X: 30559660-30559660 on Build GRCh38
rs377147619 Chr.X: 30559633-30559633 on Build GRCh38
rs926906463 Chr.X: 30559372-30559372 on Build GRCh38
rs949426771 Chr.X: 30559295-30559295 on Build GRCh38
rs780809127 Chr.X: 30559451-30559451 on Build GRCh38

View Full Product Details