Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 120537995-120538534 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CUL4B
Entrez Gene ID
8450
Gene Name
cullin 4B
Gene Aliases
CUL-4B, MRXHF2, MRXS15, MRXSC, SFM2
UniGene
Hs.102914
Gene Chromosome Location
Chr.X: 120524589-120575829 on Build GRCh38
Transcripts
NM_001079872.1 NM_003588.3 XM_005262481.1 XM_006724784.1 XM_006724785.2 XM_011531399.2 XM_011531400.2 XM_011531401.1

SNP Detail

SNP ID Chromosome Location
rs764182117 Chr.X: 120538408-120538408 on Build GRCh38
rs1031320073 Chr.X: 120538406-120538406 on Build GRCh38
rs773389833 Chr.X: 120538493-120538493 on Build GRCh38
rs896412935 Chr.X: 120538222-120538222 on Build GRCh38
rs762432016 Chr.X: 120538417-120538417 on Build GRCh38
rs749631148 Chr.X: 120538163-120538163 on Build GRCh38
rs577291875 Chr.X: 120538260-120538260 on Build GRCh38
rs780749145 Chr.X: 120538142-120538142 on Build GRCh38
rs1016011236 Chr.X: 120538275-120538275 on Build GRCh38
rs900208886 Chr.X: 120538300-120538300 on Build GRCh38
rs1049879238 Chr.X: 120538077-120538077 on Build GRCh38
rs1030382124 Chr.X: 120538360-120538360 on Build GRCh38
rs977067200 Chr.X: 120538428-120538428 on Build GRCh38
rs186860428 Chr.X: 120538246-120538246 on Build GRCh38
rs756655536 Chr.X: 120538073-120538073 on Build GRCh38
rs368304478 Chr.X: 120538145-120538145 on Build GRCh38
rs752972392 Chr.X: 120538027-120538027 on Build GRCh38
rs749311084 Chr.X: 120538126-120538126 on Build GRCh38
rs762674517 Chr.X: 120538248-120538248 on Build GRCh38
rs755952239 Chr.X: 120538033-120538033 on Build GRCh38
rs111897905 Chr.X: 120538410-120538410 on Build GRCh38
rs986994155 Chr.X: 120538413-120538413 on Build GRCh38
rs765203891 Chr.X: 120538477-120538478 on Build GRCh38
rs1022719103 Chr.X: 120538268-120538268 on Build GRCh38
rs756665325 Chr.X: 120538136-120538136 on Build GRCh38
rs763881116 Chr.X: 120538255-120538255 on Build GRCh38
rs374891698 Chr.X: 120538082-120538082 on Build GRCh38
rs866016207 Chr.X: 120538125-120538125 on Build GRCh38
rs183560146 Chr.X: 120538238-120538238 on Build GRCh38
rs1043803405 Chr.X: 120538032-120538032 on Build GRCh38
rs192247022 Chr.X: 120538235-120538235 on Build GRCh38
rs748410967 Chr.X: 120538228-120538228 on Build GRCh38
rs187407491 Chr.X: 120538234-120538234 on Build GRCh38
rs1030326774 Chr.X: 120538443-120538443 on Build GRCh38
rs922826294 Chr.X: 120538052-120538052 on Build GRCh38
rs370421802 Chr.X: 120538072-120538072 on Build GRCh38
rs999876399 Chr.X: 120538376-120538376 on Build GRCh38
rs966842604 Chr.X: 120538474-120538474 on Build GRCh38
rs181887874 Chr.X: 120538226-120538226 on Build GRCh38
rs776991033 Chr.X: 120538416-120538416 on Build GRCh38
rs769133141 Chr.X: 120538350-120538350 on Build GRCh38
rs769166770 Chr.X: 120538191-120538191 on Build GRCh38
rs745426768 Chr.X: 120538100-120538100 on Build GRCh38
rs772252891 Chr.X: 120538230-120538230 on Build GRCh38
rs11796939 Chr.X: 120538437-120538437 on Build GRCh38
rs148700620 Chr.X: 120538205-120538205 on Build GRCh38
rs866657955 Chr.X: 120538188-120538188 on Build GRCh38

View Full Product Details