Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 120625525-120626098 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1GALT1C1
Entrez Gene ID
29071
Gene Name
C1GALT1 specific chaperone 1
Gene Aliases
C1GALT2, C38H2-L1, COSMC, HSPC067, MST143, TNPS
UniGene
Hs.643920
Gene Chromosome Location
Chr.X: 120625674-120630150 on Build GRCh38
Transcripts
NM_001011551.2 NM_152692.4

SNP Detail

SNP ID Chromosome Location
rs1031845064 Chr.X: 120625618-120625618 on Build GRCh38
rs1037328717 Chr.X: 120625739-120625739 on Build GRCh38
rs137972928 Chr.X: 120625751-120625751 on Build GRCh38
rs749718983 Chr.X: 120625877-120625877 on Build GRCh38
rs760133577 Chr.X: 120626033-120626034 on Build GRCh38
rs774933003 Chr.X: 120625947-120625949 on Build GRCh38
rs762494937 Chr.X: 120625643-120625644 on Build GRCh38
rs890271686 Chr.X: 120625821-120625821 on Build GRCh38
rs992063492 Chr.X: 120626041-120626041 on Build GRCh38
rs751222455 Chr.X: 120625908-120625909 on Build GRCh38
rs180816799 Chr.X: 120625641-120625641 on Build GRCh38
rs1044847908 Chr.X: 120625556-120625556 on Build GRCh38
rs904626938 Chr.X: 120625580-120625580 on Build GRCh38
rs779922291 Chr.X: 120625567-120625570 on Build GRCh38
rs1033003076 Chr.X: 120625950-120625950 on Build GRCh38
rs931700624 Chr.X: 120625767-120625767 on Build GRCh38
rs916516650 Chr.X: 120626046-120626046 on Build GRCh38
rs908798040 Chr.X: 120625711-120625711 on Build GRCh38
rs983028402 Chr.X: 120625670-120625670 on Build GRCh38
rs754414726 Chr.X: 120625665-120625665 on Build GRCh38
rs962493984 Chr.X: 120625801-120625801 on Build GRCh38
rs769554529 Chr.X: 120625785-120625785 on Build GRCh38
rs1009160460 Chr.X: 120625843-120625843 on Build GRCh38
rs771444348 Chr.X: 120625909-120625909 on Build GRCh38
rs998388623 Chr.X: 120625658-120625658 on Build GRCh38
rs909599212 Chr.X: 120625743-120625743 on Build GRCh38
rs1000733907 Chr.X: 120625593-120625593 on Build GRCh38
rs56332141 Chr.X: 120626072-120626072 on Build GRCh38
rs200236264 Chr.X: 120625951-120625954 on Build GRCh38
rs953700558 Chr.X: 120625692-120625692 on Build GRCh38
rs1041580285 Chr.X: 120625845-120625845 on Build GRCh38
rs941642719 Chr.X: 120625727-120625727 on Build GRCh38
rs918421561 Chr.X: 120625894-120625894 on Build GRCh38
rs956240010 Chr.X: 120625625-120625625 on Build GRCh38
rs1050601768 Chr.X: 120625776-120625776 on Build GRCh38
rs780764659 Chr.X: 120625747-120625747 on Build GRCh38
rs958701111 Chr.X: 120626015-120626015 on Build GRCh38
rs186413154 Chr.X: 120625848-120625848 on Build GRCh38

View Full Product Details