Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.Y: 20592378-20592901 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF1AY
Entrez Gene ID
9086
Gene Name
eukaryotic translation initiation factor 1A, Y-linked
Gene Aliases
eIF-4C
UniGene
Hs.461178
Gene Chromosome Location
Chr.Y: 20575711-20593154 on Build GRCh38
Transcripts
NM_001278612.1 NM_004681.3

SNP Detail

SNP ID Chromosome Location
rs1064253 Chr.Y: 20592684-20592684 on Build GRCh38
rs1031430965 Chr.Y: 20592810-20592810 on Build GRCh38
rs890819867 Chr.Y: 20592628-20592628 on Build GRCh38
rs747272238 Chr.Y: 20592590-20592590 on Build GRCh38
rs368647544 Chr.Y: 20592784-20592785 on Build GRCh38
rs1039141753 Chr.Y: 20592448-20592448 on Build GRCh38
rs998911674 Chr.Y: 20592728-20592728 on Build GRCh38
rs768956288 Chr.Y: 20592730-20592730 on Build GRCh38
rs796933583 Chr.Y: 20592785-20592786 on Build GRCh38
rs201962034 Chr.Y: 20592785-20592785 on Build GRCh38
rs758018269 Chr.Y: 20592462-20592462 on Build GRCh38
rs997759500 Chr.Y: 20592553-20592553 on Build GRCh38
rs201040082 Chr.Y: 20592785-20592786 on Build GRCh38

View Full Product Details