Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 17804844-17805425 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCAF16
Entrez Gene ID
54876
Gene Name
DDB1 and CUL4 associated factor 16
Gene Aliases
C4orf30
UniGene
Hs.614787
Gene Chromosome Location
Chr.4: 17798236-17810761 on Build GRCh38
Transcripts
NM_017741.3 XM_005248169.3 XM_005248171.3 XM_006713966.3 XM_017008327.1 XM_017008328.1

SNP Detail

SNP ID Chromosome Location
rs73800401 Chr.4: 17805190-17805190 on Build GRCh38
rs576497298 Chr.4: 17805085-17805085 on Build GRCh38
rs1006510916 Chr.4: 17804893-17804893 on Build GRCh38
rs889325788 Chr.4: 17805168-17805168 on Build GRCh38
rs1025451074 Chr.4: 17804927-17804927 on Build GRCh38
rs1135724 Chr.4: 17805382-17805382 on Build GRCh38
rs913800058 Chr.4: 17805394-17805394 on Build GRCh38
rs114648388 Chr.4: 17805153-17805153 on Build GRCh38
rs956597296 Chr.4: 17805069-17805069 on Build GRCh38
rs1032676788 Chr.4: 17805188-17805188 on Build GRCh38
rs993581028 Chr.4: 17804925-17804925 on Build GRCh38
rs772568622 Chr.4: 17805071-17805071 on Build GRCh38
rs999896938 Chr.4: 17805178-17805178 on Build GRCh38
rs548701567 Chr.4: 17805008-17805008 on Build GRCh38
rs1048371644 Chr.4: 17805005-17805005 on Build GRCh38
rs1040737615 Chr.4: 17805094-17805094 on Build GRCh38
rs778647304 Chr.4: 17805171-17805171 on Build GRCh38
rs1007730699 Chr.4: 17805210-17805210 on Build GRCh38
rs770159725 Chr.4: 17805084-17805085 on Build GRCh38
rs189350225 Chr.4: 17805377-17805377 on Build GRCh38
rs550681423 Chr.4: 17804928-17804928 on Build GRCh38
rs937423083 Chr.4: 17805043-17805043 on Build GRCh38
rs780990874 Chr.4: 17805086-17805087 on Build GRCh38
rs1002332785 Chr.4: 17805264-17805264 on Build GRCh38
rs1055847143 Chr.4: 17805091-17805091 on Build GRCh38
rs11543278 Chr.4: 17805198-17805198 on Build GRCh38
rs567015802 Chr.4: 17805038-17805038 on Build GRCh38
rs542695114 Chr.4: 17804981-17804981 on Build GRCh38
rs911383098 Chr.4: 17804903-17804903 on Build GRCh38
rs1016268862 Chr.4: 17804901-17804902 on Build GRCh38
rs1041678239 Chr.4: 17805155-17805155 on Build GRCh38
rs930005054 Chr.4: 17805004-17805004 on Build GRCh38
rs976871328 Chr.4: 17805402-17805402 on Build GRCh38
rs965541713 Chr.4: 17804907-17804907 on Build GRCh38
rs993882204 Chr.4: 17805203-17805203 on Build GRCh38
rs1018668198 Chr.4: 17805258-17805258 on Build GRCh38
rs753397932 Chr.4: 17804917-17804917 on Build GRCh38
rs1024976656 Chr.4: 17805217-17805217 on Build GRCh38
rs989286539 Chr.4: 17805335-17805335 on Build GRCh38
rs753463325 Chr.4: 17805085-17805087 on Build GRCh38
rs989380908 Chr.4: 17805067-17805067 on Build GRCh38
rs578216259 Chr.4: 17805308-17805308 on Build GRCh38
rs569257754 Chr.4: 17804957-17804957 on Build GRCh38
rs970854883 Chr.4: 17804943-17804943 on Build GRCh38
rs971185185 Chr.4: 17805250-17805250 on Build GRCh38
rs771329386 Chr.4: 17805272-17805272 on Build GRCh38
rs74667233 Chr.4: 17805101-17805104 on Build GRCh38
rs962059864 Chr.4: 17804910-17804910 on Build GRCh38
rs922308023 Chr.4: 17805104-17805105 on Build GRCh38
rs182377100 Chr.4: 17805093-17805093 on Build GRCh38
rs556649074 Chr.4: 17805336-17805336 on Build GRCh38

View Full Product Details