Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 105552367-105552915 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGEF38
Entrez Gene ID
54848
Gene Name
Rho guanine nucleotide exchange factor 38
Gene Aliases
-
UniGene
Hs.585224
Gene Chromosome Location
Chr.4: 105552603-105682539 on Build GRCh38
Transcripts
NM_001242729.1 NM_017700.1 XM_011532050.2 XM_011532051.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1642376 c.32A>C p.N11T Chr.4: 105552797-105552797 on Build GRCh38
6604311 c.39C>T p.V13V Chr.4: 105552804-105552804 on Build GRCh38
77724 c.12A>C p.K4N Chr.4: 105552777-105552777 on Build GRCh38
7356353 c.61T>C p.F21L Chr.4: 105552826-105552826 on Build GRCh38
1642375 c.32A>C p.N11T Chr.4: 105552797-105552797 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758535583 Chr.4: 105552846-105552846 on Build GRCh38
rs757174787 Chr.4: 105552765-105552765 on Build GRCh38
rs965565840 Chr.4: 105552780-105552780 on Build GRCh38
rs752417042 Chr.4: 105552715-105552715 on Build GRCh38
rs1044198890 Chr.4: 105552572-105552572 on Build GRCh38
rs762338044 Chr.4: 105552816-105552816 on Build GRCh38
rs75086757 Chr.4: 105552705-105552705 on Build GRCh38
rs1019776161 Chr.4: 105552730-105552730 on Build GRCh38
rs1054756246 Chr.4: 105552492-105552492 on Build GRCh38
rs1027618260 Chr.4: 105552699-105552699 on Build GRCh38
rs948661192 Chr.4: 105552851-105552851 on Build GRCh38
rs375204038 Chr.4: 105552818-105552818 on Build GRCh38
rs912771333 Chr.4: 105552416-105552416 on Build GRCh38
rs777443919 Chr.4: 105552720-105552720 on Build GRCh38
rs138889321 Chr.4: 105552841-105552841 on Build GRCh38
rs112195119 Chr.4: 105552642-105552642 on Build GRCh38
rs893054566 Chr.4: 105552504-105552504 on Build GRCh38
rs780289579 Chr.4: 105552878-105552878 on Build GRCh38
rs1010130237 Chr.4: 105552441-105552441 on Build GRCh38
rs775849401 Chr.4: 105552808-105552808 on Build GRCh38
rs147662568 Chr.4: 105552824-105552824 on Build GRCh38
rs146139851 Chr.4: 105552834-105552834 on Build GRCh38
rs983609319 Chr.4: 105552833-105552833 on Build GRCh38
rs61413380 Chr.4: 105552823-105552823 on Build GRCh38
rs747288600 Chr.4: 105552811-105552811 on Build GRCh38
rs754455990 Chr.4: 105552714-105552714 on Build GRCh38
rs562263895 Chr.4: 105552767-105552767 on Build GRCh38
rs199819596 Chr.4: 105552886-105552886 on Build GRCh38
rs557343221 Chr.4: 105552533-105552533 on Build GRCh38
rs149171462 Chr.4: 105552581-105552581 on Build GRCh38
rs61753342 Chr.4: 105552826-105552826 on Build GRCh38
rs572294831 Chr.4: 105552679-105552679 on Build GRCh38
rs1002942686 Chr.4: 105552606-105552606 on Build GRCh38
rs749184437 Chr.4: 105552763-105552763 on Build GRCh38
rs745919785 Chr.4: 105552786-105552786 on Build GRCh38
rs755727014 Chr.4: 105552829-105552829 on Build GRCh38
rs769002524 Chr.4: 105552812-105552812 on Build GRCh38
rs907083398 Chr.4: 105552583-105552583 on Build GRCh38
rs371638919 Chr.4: 105552860-105552860 on Build GRCh38
rs897141518 Chr.4: 105552650-105552650 on Build GRCh38
rs772317417 Chr.4: 105552791-105552791 on Build GRCh38
rs934357097 Chr.4: 105552477-105552477 on Build GRCh38
rs944437099 Chr.4: 105552421-105552421 on Build GRCh38
rs759047037 Chr.4: 105552821-105552821 on Build GRCh38
rs1034056168 Chr.4: 105552627-105552627 on Build GRCh38
rs776862032 Chr.4: 105552815-105552815 on Build GRCh38
rs754240515 Chr.4: 105552873-105552874 on Build GRCh38
rs755925399 Chr.4: 105552718-105552718 on Build GRCh38
rs778960894 Chr.4: 105552843-105552843 on Build GRCh38
rs779085578 Chr.4: 105552778-105552778 on Build GRCh38
rs975854629 Chr.4: 105552454-105552454 on Build GRCh38
rs954317005 Chr.4: 105552398-105552398 on Build GRCh38
rs758224060 Chr.4: 105552612-105552612 on Build GRCh38
rs924261348 Chr.4: 105552472-105552472 on Build GRCh38
rs745866842 Chr.4: 105552844-105552844 on Build GRCh38
rs559868186 Chr.4: 105552637-105552637 on Build GRCh38
rs763912179 Chr.4: 105552831-105552831 on Build GRCh38
rs948523971 Chr.4: 105552532-105552532 on Build GRCh38
rs989839055 Chr.4: 105552835-105552835 on Build GRCh38
rs537461597 Chr.4: 105552407-105552407 on Build GRCh38
rs147684533 Chr.4: 105552579-105552579 on Build GRCh38

View Full Product Details