Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 101724242-101724823 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP97
Entrez Gene ID
79598
Gene Name
centrosomal protein 97
Gene Aliases
2810403B08Rik, LRRIQ2
UniGene
Hs.444135
Gene Chromosome Location
Chr.3: 101724098-101770562 on Build GRCh38
Transcripts
NM_001303401.1 NM_024548.3 XM_006713743.3 XM_011513127.1 XM_017007178.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3584666 c.38G>A p.G13E Chr.3: 101724714-101724714 on Build GRCh38
3584667 c.38G>A p.G13E Chr.3: 101724714-101724714 on Build GRCh38
6095443 c.36C>T p.P12P Chr.3: 101724712-101724712 on Build GRCh38
5863116 c.31C>T p.P11S Chr.3: 101724707-101724707 on Build GRCh38
1035828 c.40G>A p.E14K Chr.3: 101724716-101724716 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs141029319 Chr.3: 101724560-101724560 on Build GRCh38
rs1040201547 Chr.3: 101724624-101724624 on Build GRCh38
rs35159419 Chr.3: 101724302-101724302 on Build GRCh38
rs1004691549 Chr.3: 101724549-101724549 on Build GRCh38
rs748809597 Chr.3: 101724714-101724714 on Build GRCh38
rs145352421 Chr.3: 101724463-101724463 on Build GRCh38
rs899760226 Chr.3: 101724491-101724491 on Build GRCh38
rs769300227 Chr.3: 101724645-101724645 on Build GRCh38
rs886971375 Chr.3: 101724541-101724541 on Build GRCh38
rs532883279 Chr.3: 101724503-101724503 on Build GRCh38
rs566730951 Chr.3: 101724542-101724542 on Build GRCh38
rs924754267 Chr.3: 101724608-101724608 on Build GRCh38
rs937597199 Chr.3: 101724546-101724546 on Build GRCh38
rs6790109 Chr.3: 101724656-101724656 on Build GRCh38
rs1023138076 Chr.3: 101724564-101724564 on Build GRCh38
rs766700963 Chr.3: 101724468-101724468 on Build GRCh38
rs137996409 Chr.3: 101724511-101724511 on Build GRCh38
rs947519172 Chr.3: 101724327-101724327 on Build GRCh38
rs757072345 Chr.3: 101724694-101724694 on Build GRCh38
rs115499192 Chr.3: 101724613-101724613 on Build GRCh38
rs377040183 Chr.3: 101724730-101724730 on Build GRCh38
rs1032873536 Chr.3: 101724300-101724300 on Build GRCh38
rs572567778 Chr.3: 101724777-101724777 on Build GRCh38
rs113730099 Chr.3: 101724651-101724651 on Build GRCh38
rs578050989 Chr.3: 101724649-101724649 on Build GRCh38
rs377554540 Chr.3: 101724761-101724761 on Build GRCh38
rs958892813 Chr.3: 101724791-101724791 on Build GRCh38
rs934871801 Chr.3: 101724609-101724609 on Build GRCh38
rs916182380 Chr.3: 101724325-101724325 on Build GRCh38
rs773569838 Chr.3: 101724667-101724667 on Build GRCh38
rs968769796 Chr.3: 101724785-101724785 on Build GRCh38
rs753924919 Chr.3: 101724687-101724687 on Build GRCh38
rs764007295 Chr.3: 101724768-101724768 on Build GRCh38
rs201771736 Chr.3: 101724690-101724690 on Build GRCh38
rs969367946 Chr.3: 101724582-101724582 on Build GRCh38
rs780812135 Chr.3: 101724639-101724639 on Build GRCh38
rs928622308 Chr.3: 101724272-101724272 on Build GRCh38
rs534165252 Chr.3: 101724543-101724543 on Build GRCh38
rs796765680 Chr.3: 101724275-101724275 on Build GRCh38
rs745451146 Chr.3: 101724641-101724641 on Build GRCh38
rs191271728 Chr.3: 101724759-101724759 on Build GRCh38
rs761345831 Chr.3: 101724639-101724641 on Build GRCh38
rs72935871 Chr.3: 101724495-101724495 on Build GRCh38
rs1035750473 Chr.3: 101724550-101724550 on Build GRCh38
rs537723235 Chr.3: 101724287-101724287 on Build GRCh38
rs772575367 Chr.3: 101724752-101724752 on Build GRCh38
rs769175929 Chr.3: 101724650-101724650 on Build GRCh38
rs758311009 Chr.3: 101724689-101724689 on Build GRCh38
rs373337025 Chr.3: 101724704-101724704 on Build GRCh38
rs1012988556 Chr.3: 101724776-101724776 on Build GRCh38
rs748534690 Chr.3: 101724744-101724744 on Build GRCh38
rs368851013 Chr.3: 101724736-101724736 on Build GRCh38
rs989211499 Chr.3: 101724368-101724368 on Build GRCh38
rs77649996 Chr.3: 101724430-101724430 on Build GRCh38
rs375142336 Chr.3: 101724635-101724635 on Build GRCh38
rs979073820 Chr.3: 101724583-101724583 on Build GRCh38
rs562060557 Chr.3: 101724365-101724365 on Build GRCh38
rs1034418154 Chr.3: 101724781-101724781 on Build GRCh38
rs377037537 Chr.3: 101724680-101724680 on Build GRCh38
rs769100893 Chr.3: 101724726-101724726 on Build GRCh38
rs1052213120 Chr.3: 101724417-101724417 on Build GRCh38
rs397875404 Chr.3: 101724303-101724303 on Build GRCh38
rs1043623618 Chr.3: 101724361-101724361 on Build GRCh38
rs755715767 Chr.3: 101724706-101724706 on Build GRCh38
rs768028749 Chr.3: 101724746-101724746 on Build GRCh38
rs186323336 Chr.3: 101724421-101724421 on Build GRCh38
rs991672643 Chr.3: 101724296-101724296 on Build GRCh38
rs779572200 Chr.3: 101724710-101724710 on Build GRCh38
rs886913005 Chr.3: 101724719-101724719 on Build GRCh38
rs774925234 Chr.3: 101724648-101724648 on Build GRCh38
rs111828531 Chr.3: 101724653-101724653 on Build GRCh38
rs369522245 Chr.3: 101724612-101724612 on Build GRCh38
rs1022918710 Chr.3: 101724782-101724782 on Build GRCh38
rs773659559 Chr.3: 101724755-101724755 on Build GRCh38
rs201090433 Chr.3: 101724684-101724684 on Build GRCh38
rs763332096 Chr.3: 101724390-101724390 on Build GRCh38
rs370058080 Chr.3: 101724697-101724697 on Build GRCh38
rs995953225 Chr.3: 101724637-101724638 on Build GRCh38
rs759651144 Chr.3: 101724766-101724766 on Build GRCh38
rs777464678 Chr.3: 101724626-101724626 on Build GRCh38
rs71315254 Chr.3: 101724362-101724362 on Build GRCh38
rs935071661 Chr.3: 101724393-101724393 on Build GRCh38
rs549310443 Chr.3: 101724554-101724554 on Build GRCh38
rs994573 Chr.3: 101724617-101724617 on Build GRCh38
rs746792187 Chr.3: 101724633-101724633 on Build GRCh38
rs778731545 Chr.3: 101724267-101724267 on Build GRCh38
rs931206640 Chr.3: 101724664-101724664 on Build GRCh38

View Full Product Details