Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 122120100-122120626 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD86
Entrez Gene ID
942
Gene Name
CD86 molecule
Gene Aliases
B7-2, B7.2, B70, CD28LG2, LAB72
UniGene
Hs.171182
Gene Chromosome Location
Chr.3: 122055362-122121143 on Build GRCh38
Transcripts
NM_001206924.1 NM_001206925.1 NM_006889.4 NM_175862.4 NM_176892.1

SNP Detail

SNP ID Chromosome Location
rs113741816 Chr.3: 122120353-122120353 on Build GRCh38
rs1048775484 Chr.3: 122120296-122120296 on Build GRCh38
rs1002795066 Chr.3: 122120255-122120255 on Build GRCh38
rs946106802 Chr.3: 122120264-122120264 on Build GRCh38
rs751366701 Chr.3: 122120326-122120326 on Build GRCh38
rs945655821 Chr.3: 122120201-122120201 on Build GRCh38
rs892119361 Chr.3: 122120146-122120146 on Build GRCh38
rs966395490 Chr.3: 122120267-122120267 on Build GRCh38
rs368722938 Chr.3: 122120396-122120397 on Build GRCh38
rs913631417 Chr.3: 122120502-122120502 on Build GRCh38
rs1055738525 Chr.3: 122120574-122120574 on Build GRCh38
rs910295649 Chr.3: 122120128-122120128 on Build GRCh38
rs919050590 Chr.3: 122120280-122120280 on Build GRCh38
rs756086127 Chr.3: 122120457-122120457 on Build GRCh38
rs112875445 Chr.3: 122120162-122120162 on Build GRCh38
rs945058192 Chr.3: 122120503-122120503 on Build GRCh38
rs569419464 Chr.3: 122120422-122120422 on Build GRCh38
rs896614284 Chr.3: 122120343-122120343 on Build GRCh38
rs111445779 Chr.3: 122120409-122120409 on Build GRCh38
rs1055234044 Chr.3: 122120379-122120379 on Build GRCh38
rs976027402 Chr.3: 122120270-122120270 on Build GRCh38
rs985486590 Chr.3: 122120225-122120225 on Build GRCh38
rs966908410 Chr.3: 122120135-122120135 on Build GRCh38
rs925530977 Chr.3: 122120541-122120541 on Build GRCh38
rs944456488 Chr.3: 122120134-122120134 on Build GRCh38
rs375030172 Chr.3: 122120392-122120392 on Build GRCh38
rs961385356 Chr.3: 122120381-122120381 on Build GRCh38
rs571805253 Chr.3: 122120223-122120223 on Build GRCh38
rs531230600 Chr.3: 122120477-122120477 on Build GRCh38
rs1019993630 Chr.3: 122120534-122120534 on Build GRCh38
rs1043364889 Chr.3: 122120519-122120519 on Build GRCh38
rs1032444341 Chr.3: 122120151-122120151 on Build GRCh38
rs976376514 Chr.3: 122120592-122120592 on Build GRCh38
rs938276984 Chr.3: 122120549-122120549 on Build GRCh38
rs1001055459 Chr.3: 122120434-122120434 on Build GRCh38
rs774798899 Chr.3: 122120291-122120291 on Build GRCh38
rs541158845 Chr.3: 122120260-122120260 on Build GRCh38
rs567953312 Chr.3: 122120588-122120588 on Build GRCh38
rs546153682 Chr.3: 122120447-122120447 on Build GRCh38
rs1006892343 Chr.3: 122120474-122120474 on Build GRCh38
rs986408055 Chr.3: 122120431-122120431 on Build GRCh38
rs902654828 Chr.3: 122120420-122120420 on Build GRCh38
rs951956766 Chr.3: 122120411-122120411 on Build GRCh38
rs775772991 Chr.3: 122120426-122120426 on Build GRCh38
rs114171166 Chr.3: 122120478-122120478 on Build GRCh38
rs549664582 Chr.3: 122120400-122120400 on Build GRCh38
rs975786766 Chr.3: 122120461-122120461 on Build GRCh38
rs140955956 Chr.3: 122120448-122120448 on Build GRCh38
rs904076956 Chr.3: 122120247-122120247 on Build GRCh38
rs1044389598 Chr.3: 122120222-122120222 on Build GRCh38
rs1015624024 Chr.3: 122120365-122120365 on Build GRCh38
rs182733603 Chr.3: 122120138-122120138 on Build GRCh38

View Full Product Details