Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 105158147-105158746 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BVES-AS1
Entrez Gene ID
154442
Gene Name
BVES antisense RNA 1
Gene Aliases
C6orf112, bA99L11.2
UniGene
Hs.586271
Gene Chromosome Location
Chr.6: 105137687-105169945 on Build GRCh38
Transcripts
NR_037157.1 NM_022361.4 NR_024539.1 XM_011536067.2 XM_017011194.1 XM_017011195.1

Gene Symbol
POPDC3
Entrez Gene ID
64208
Gene Name
popeye domain containing 3
Gene Aliases
POP3, bA355M14.1
UniGene
Hs.458336
Gene Chromosome Location
Chr.6: 105157900-105179983 on Build GRCh38
Transcripts
NR_037157.1 NM_022361.4 NR_024539.1 XM_011536067.2 XM_017011194.1 XM_017011195.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6752728 c.622C>T p.R208* Chr.6: 105158724-105158724 on Build GRCh38
211579 c.804G>T p.M268I Chr.6: 105158542-105158542 on Build GRCh38
4558193 c.758G>C p.G253A Chr.6: 105158588-105158588 on Build GRCh38
200431 c.764G>T p.R255I Chr.6: 105158582-105158582 on Build GRCh38
6752727 c.684C>T p.S228S Chr.6: 105158662-105158662 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781713331 Chr.6: 105158513-105158514 on Build GRCh38
rs765304783 Chr.6: 105158670-105158670 on Build GRCh38
rs555748197 Chr.6: 105158243-105158243 on Build GRCh38
rs747606562 Chr.6: 105158637-105158637 on Build GRCh38
rs750799364 Chr.6: 105158513-105158513 on Build GRCh38
rs770763097 Chr.6: 105158539-105158539 on Build GRCh38
rs1758 Chr.6: 105158185-105158185 on Build GRCh38
rs764297745 Chr.6: 105158448-105158448 on Build GRCh38
rs145514874 Chr.6: 105158445-105158445 on Build GRCh38
rs568369449 Chr.6: 105158652-105158652 on Build GRCh38
rs142611329 Chr.6: 105158535-105158535 on Build GRCh38
rs773031019 Chr.6: 105158430-105158430 on Build GRCh38
rs781686581 Chr.6: 105158635-105158635 on Build GRCh38
rs376904506 Chr.6: 105158431-105158431 on Build GRCh38
rs766174156 Chr.6: 105158433-105158433 on Build GRCh38
rs903217056 Chr.6: 105158504-105158506 on Build GRCh38
rs762010270 Chr.6: 105158581-105158581 on Build GRCh38
rs774148644 Chr.6: 105158544-105158544 on Build GRCh38
rs371162422 Chr.6: 105158664-105158664 on Build GRCh38
rs139964706 Chr.6: 105158598-105158598 on Build GRCh38
rs752998090 Chr.6: 105158425-105158427 on Build GRCh38
rs1018579467 Chr.6: 105158630-105158630 on Build GRCh38
rs999292554 Chr.6: 105158252-105158252 on Build GRCh38
rs763282966 Chr.6: 105158714-105158714 on Build GRCh38
rs763216966 Chr.6: 105158427-105158428 on Build GRCh38
rs750662233 Chr.6: 105158600-105158600 on Build GRCh38
rs751620708 Chr.6: 105158434-105158435 on Build GRCh38
rs936036418 Chr.6: 105158679-105158679 on Build GRCh38
rs758687042 Chr.6: 105158601-105158601 on Build GRCh38
rs773294339 Chr.6: 105158676-105158676 on Build GRCh38
rs1053653663 Chr.6: 105158372-105158372 on Build GRCh38
rs371952749 Chr.6: 105158425-105158425 on Build GRCh38
rs1014656575 Chr.6: 105158458-105158458 on Build GRCh38
rs779312528 Chr.6: 105158469-105158469 on Build GRCh38
rs770615770 Chr.6: 105158653-105158653 on Build GRCh38
rs941493640 Chr.6: 105158691-105158691 on Build GRCh38
rs138128513 Chr.6: 105158486-105158486 on Build GRCh38
rs200555248 Chr.6: 105158390-105158390 on Build GRCh38
rs751524942 Chr.6: 105158439-105158439 on Build GRCh38
rs964015622 Chr.6: 105158308-105158308 on Build GRCh38
rs1041793055 Chr.6: 105158293-105158293 on Build GRCh38
rs757769023 Chr.6: 105158437-105158437 on Build GRCh38
rs146013025 Chr.6: 105158575-105158575 on Build GRCh38
rs922320572 Chr.6: 105158359-105158359 on Build GRCh38
rs776338492 Chr.6: 105158648-105158648 on Build GRCh38
rs989800841 Chr.6: 105158266-105158271 on Build GRCh38
rs750769346 Chr.6: 105158473-105158473 on Build GRCh38
rs757597113 Chr.6: 105158459-105158459 on Build GRCh38
rs894737754 Chr.6: 105158453-105158453 on Build GRCh38
rs765481307 Chr.6: 105158673-105158673 on Build GRCh38
rs541277995 Chr.6: 105158608-105158608 on Build GRCh38
rs1054547392 Chr.6: 105158695-105158695 on Build GRCh38
rs780236689 Chr.6: 105158668-105158672 on Build GRCh38
rs111547826 Chr.6: 105158672-105158672 on Build GRCh38
rs768036293 Chr.6: 105158508-105158508 on Build GRCh38
rs976426092 Chr.6: 105158310-105158310 on Build GRCh38
rs780728201 Chr.6: 105158516-105158516 on Build GRCh38
rs374388891 Chr.6: 105158565-105158565 on Build GRCh38
rs909637766 Chr.6: 105158683-105158683 on Build GRCh38
rs747767403 Chr.6: 105158519-105158519 on Build GRCh38
rs893652983 Chr.6: 105158382-105158382 on Build GRCh38
rs751986613 Chr.6: 105158616-105158616 on Build GRCh38
rs143503012 Chr.6: 105158638-105158638 on Build GRCh38
rs1030775562 Chr.6: 105158256-105158256 on Build GRCh38
rs769396346 Chr.6: 105158525-105158525 on Build GRCh38
rs924713953 Chr.6: 105158328-105158328 on Build GRCh38
rs761727705 Chr.6: 105158407-105158407 on Build GRCh38
rs764280860 Chr.6: 105158427-105158430 on Build GRCh38
rs911875875 Chr.6: 105158240-105158240 on Build GRCh38
rs911143272 Chr.6: 105158282-105158282 on Build GRCh38
rs986373190 Chr.6: 105158596-105158596 on Build GRCh38
rs761510109 Chr.6: 105158421-105158421 on Build GRCh38
rs368997977 Chr.6: 105158660-105158660 on Build GRCh38
rs749034445 Chr.6: 105158645-105158645 on Build GRCh38
rs756465603 Chr.6: 105158654-105158665 on Build GRCh38
rs755547219 Chr.6: 105158619-105158619 on Build GRCh38
rs957930030 Chr.6: 105158258-105158258 on Build GRCh38
rs140524770 Chr.6: 105158273-105158273 on Build GRCh38
rs772870574 Chr.6: 105158526-105158526 on Build GRCh38
rs376176931 Chr.6: 105158642-105158642 on Build GRCh38
rs200001271 Chr.6: 105158723-105158723 on Build GRCh38
rs986794027 Chr.6: 105158221-105158221 on Build GRCh38
rs944719264 Chr.6: 105158288-105158288 on Build GRCh38
rs998884326 Chr.6: 105158521-105158521 on Build GRCh38
rs935150428 Chr.6: 105158337-105158337 on Build GRCh38
rs533264918 Chr.6: 105158451-105158451 on Build GRCh38
rs397887227 Chr.6: 105158659-105158659 on Build GRCh38
rs202208117 Chr.6: 105158582-105158582 on Build GRCh38
rs977865090 Chr.6: 105158668-105158668 on Build GRCh38
rs370790250 Chr.6: 105158487-105158487 on Build GRCh38
rs758804753 Chr.6: 105158483-105158483 on Build GRCh38
rs759571147 Chr.6: 105158560-105158560 on Build GRCh38
rs745683947 Chr.6: 105158661-105158661 on Build GRCh38
rs35286940 Chr.6: 105158564-105158565 on Build GRCh38

View Full Product Details