Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 138785023-138785558 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC28A
Entrez Gene ID
25901
Gene Name
coiled-coil domain containing 28A
Gene Aliases
C6orf80, CCRL1AP
UniGene
Hs.412019
Gene Chromosome Location
Chr.6: 138773517-138793319 on Build GRCh38
Transcripts
NM_015439.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6105524 c.629G>A p.R210Q Chr.6: 138785263-138785263 on Build GRCh38
3829169 c.709A>C p.T237P Chr.6: 138785343-138785343 on Build GRCh38
1440812 c.642G>T p.E214D Chr.6: 138785276-138785276 on Build GRCh38
6622032 c.665A>T p.E222V Chr.6: 138785299-138785299 on Build GRCh38
5650991 c.693T>C p.P231P Chr.6: 138785327-138785327 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs147173471 Chr.6: 138785382-138785382 on Build GRCh38
rs766119341 Chr.6: 138785388-138785388 on Build GRCh38
rs779370032 Chr.6: 138785308-138785308 on Build GRCh38
rs771285553 Chr.6: 138785210-138785210 on Build GRCh38
rs1051278389 Chr.6: 138785399-138785399 on Build GRCh38
rs757518652 Chr.6: 138785306-138785306 on Build GRCh38
rs757825011 Chr.6: 138785415-138785415 on Build GRCh38
rs191513067 Chr.6: 138785464-138785464 on Build GRCh38
rs751125896 Chr.6: 138785287-138785287 on Build GRCh38
rs117540720 Chr.6: 138785183-138785183 on Build GRCh38
rs113233430 Chr.6: 138785455-138785455 on Build GRCh38
rs754351642 Chr.6: 138785303-138785303 on Build GRCh38
rs761634317 Chr.6: 138785262-138785262 on Build GRCh38
rs768774318 Chr.6: 138785246-138785246 on Build GRCh38
rs78283939 Chr.6: 138785172-138785172 on Build GRCh38
rs747015801 Chr.6: 138785392-138785392 on Build GRCh38
rs891923897 Chr.6: 138785392-138785392 on Build GRCh38
rs576099091 Chr.6: 138785168-138785168 on Build GRCh38
rs576241297 Chr.6: 138785242-138785242 on Build GRCh38
rs746049394 Chr.6: 138785310-138785310 on Build GRCh38
rs3763185 Chr.6: 138785462-138785462 on Build GRCh38
rs770794674 Chr.6: 138785453-138785453 on Build GRCh38
rs1046710292 Chr.6: 138785499-138785499 on Build GRCh38
rs780777022 Chr.6: 138785419-138785420 on Build GRCh38
rs779127383 Chr.6: 138785419-138785419 on Build GRCh38
rs369287588 Chr.6: 138785430-138785430 on Build GRCh38
rs946218947 Chr.6: 138785520-138785520 on Build GRCh38
rs74551613 Chr.6: 138785466-138785466 on Build GRCh38
rs747153743 Chr.6: 138785333-138785333 on Build GRCh38
rs144679333 Chr.6: 138785516-138785516 on Build GRCh38
rs372791693 Chr.6: 138785255-138785255 on Build GRCh38
rs147991567 Chr.6: 138785281-138785281 on Build GRCh38
rs777556563 Chr.6: 138785335-138785336 on Build GRCh38
rs143442772 Chr.6: 138785263-138785263 on Build GRCh38
rs750002558 Chr.6: 138785279-138785279 on Build GRCh38
rs768862392 Chr.6: 138785335-138785335 on Build GRCh38
rs927243719 Chr.6: 138785484-138785484 on Build GRCh38
rs200099976 Chr.6: 138785189-138785189 on Build GRCh38
rs927377712 Chr.6: 138785364-138785364 on Build GRCh38
rs778140975 Chr.6: 138785285-138785285 on Build GRCh38
rs778182507 Chr.6: 138785302-138785302 on Build GRCh38
rs779645423 Chr.6: 138785188-138785188 on Build GRCh38
rs541939937 Chr.6: 138785334-138785334 on Build GRCh38
rs962786050 Chr.6: 138785429-138785429 on Build GRCh38
rs1009435707 Chr.6: 138785203-138785203 on Build GRCh38
rs141328613 Chr.6: 138785400-138785400 on Build GRCh38
rs548279361 Chr.6: 138785507-138785507 on Build GRCh38
rs1034817624 Chr.6: 138785322-138785322 on Build GRCh38
rs754306673 Chr.6: 138785410-138785410 on Build GRCh38
rs568176588 Chr.6: 138785509-138785509 on Build GRCh38
rs772387526 Chr.6: 138785312-138785312 on Build GRCh38
rs547448968 Chr.6: 138785531-138785531 on Build GRCh38
rs1025940002 Chr.6: 138785449-138785449 on Build GRCh38
rs202062726 Chr.6: 138785286-138785286 on Build GRCh38
rs779059884 Chr.6: 138785394-138785394 on Build GRCh38
rs531738683 Chr.6: 138785493-138785493 on Build GRCh38
rs763303493 Chr.6: 138785057-138785057 on Build GRCh38
rs749716228 Chr.6: 138785208-138785208 on Build GRCh38
rs577030036 Chr.6: 138785192-138785192 on Build GRCh38
rs147073588 Chr.6: 138785343-138785344 on Build GRCh38
rs556373419 Chr.6: 138785241-138785241 on Build GRCh38
rs962413356 Chr.6: 138785186-138785186 on Build GRCh38
rs756605621 Chr.6: 138785300-138785300 on Build GRCh38
rs771872912 Chr.6: 138785519-138785519 on Build GRCh38
rs745780448 Chr.6: 138785472-138785472 on Build GRCh38
rs201703653 Chr.6: 138785338-138785338 on Build GRCh38
rs143504509 Chr.6: 138785336-138785336 on Build GRCh38
rs920949062 Chr.6: 138785149-138785149 on Build GRCh38
rs865942596 Chr.6: 138785071-138785071 on Build GRCh38
rs780356967 Chr.6: 138785425-138785425 on Build GRCh38
rs749416695 Chr.6: 138785146-138785146 on Build GRCh38
rs773106448 Chr.6: 138785378-138785378 on Build GRCh38
rs77141562 Chr.6: 138785343-138785343 on Build GRCh38
rs779987154 Chr.6: 138785328-138785328 on Build GRCh38
rs778949414 Chr.6: 138785218-138785218 on Build GRCh38
rs756450578 Chr.6: 138785269-138785269 on Build GRCh38
rs548217301 Chr.6: 138785154-138785154 on Build GRCh38
rs562913988 Chr.6: 138785420-138785420 on Build GRCh38
rs760669204 Chr.6: 138785244-138785244 on Build GRCh38
rs138428828 Chr.6: 138785240-138785240 on Build GRCh38
rs750095345 Chr.6: 138785181-138785181 on Build GRCh38
rs955038075 Chr.6: 138785086-138785086 on Build GRCh38
rs764865873 Chr.6: 138785177-138785177 on Build GRCh38
rs770315275 Chr.6: 138785411-138785411 on Build GRCh38
rs747247688 Chr.6: 138785426-138785426 on Build GRCh38

View Full Product Details