Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 158636991-158637574 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DYNLT1
Entrez Gene ID
6993
Gene Name
dynein light chain Tctex-type 1
Gene Aliases
CW-1, TCTEL1, tctex-1
UniGene
Hs.445999
Gene Chromosome Location
Chr.6: 158636474-158644786 on Build GRCh38
Transcripts
NM_001291602.1 NM_001291603.1 NM_006519.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6223129 c.142G>T p.A48S Chr.6: 158637050-158637050 on Build GRCh38
4352639 c.53G>C p.S18T Chr.6: 158637139-158637139 on Build GRCh38
3374127 c.245C>T p.S82F Chr.6: 158637154-158637154 on Build GRCh38
7198885 c.22T>C p.L8L Chr.6: 158637170-158637170 on Build GRCh38
3374125 c.263C>T p.S88F Chr.6: 158637136-158637136 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754853453 Chr.6: 158637078-158637078 on Build GRCh38
rs115281201 Chr.6: 158637301-158637301 on Build GRCh38
rs761351502 Chr.6: 158637067-158637067 on Build GRCh38
rs552230520 Chr.6: 158637481-158637481 on Build GRCh38
rs568229136 Chr.6: 158637515-158637515 on Build GRCh38
rs1040290838 Chr.6: 158637384-158637384 on Build GRCh38
rs777699477 Chr.6: 158637032-158637032 on Build GRCh38
rs1031415920 Chr.6: 158637064-158637064 on Build GRCh38
rs180883614 Chr.6: 158637328-158637328 on Build GRCh38
rs770242853 Chr.6: 158637061-158637061 on Build GRCh38
rs758542251 Chr.6: 158637129-158637129 on Build GRCh38
rs919325174 Chr.6: 158637309-158637309 on Build GRCh38
rs928225646 Chr.6: 158637514-158637514 on Build GRCh38
rs746346206 Chr.6: 158637154-158637154 on Build GRCh38
rs878875450 Chr.6: 158637316-158637316 on Build GRCh38
rs746399279 Chr.6: 158637054-158637054 on Build GRCh38
rs775290987 Chr.6: 158637219-158637219 on Build GRCh38
rs538093945 Chr.6: 158637507-158637507 on Build GRCh38
rs567681596 Chr.6: 158637351-158637351 on Build GRCh38
rs574590014 Chr.6: 158637048-158637048 on Build GRCh38
rs1036350575 Chr.6: 158637306-158637306 on Build GRCh38
rs751764752 Chr.6: 158637239-158637239 on Build GRCh38
rs761035906 Chr.6: 158637039-158637039 on Build GRCh38
rs780553228 Chr.6: 158637189-158637189 on Build GRCh38
rs1044459304 Chr.6: 158637017-158637017 on Build GRCh38
rs1028066288 Chr.6: 158637192-158637192 on Build GRCh38
rs778919480 Chr.6: 158637089-158637090 on Build GRCh38
rs764990508 Chr.6: 158637102-158637102 on Build GRCh38
rs556416245 Chr.6: 158637513-158637513 on Build GRCh38
rs998562831 Chr.6: 158637045-158637045 on Build GRCh38
rs770392224 Chr.6: 158637204-158637204 on Build GRCh38
rs772595262 Chr.6: 158637075-158637075 on Build GRCh38
rs762748996 Chr.6: 158637221-158637221 on Build GRCh38
rs764307943 Chr.6: 158637133-158637133 on Build GRCh38
rs905991189 Chr.6: 158637037-158637037 on Build GRCh38
rs548241289 Chr.6: 158637420-158637420 on Build GRCh38
rs757561354 Chr.6: 158637296-158637296 on Build GRCh38
rs778705807 Chr.6: 158637019-158637019 on Build GRCh38
rs781740798 Chr.6: 158637053-158637053 on Build GRCh38
rs756685903 Chr.6: 158637244-158637244 on Build GRCh38
rs1000572952 Chr.6: 158637138-158637138 on Build GRCh38
rs750564461 Chr.6: 158637346-158637346 on Build GRCh38
rs1023143851 Chr.6: 158637483-158637483 on Build GRCh38
rs530103756 Chr.6: 158637404-158637404 on Build GRCh38
rs1057303139 Chr.6: 158637056-158637056 on Build GRCh38
rs757310929 Chr.6: 158637147-158637147 on Build GRCh38
rs763815494 Chr.6: 158637223-158637223 on Build GRCh38
rs901790681 Chr.6: 158637400-158637400 on Build GRCh38
rs1043310834 Chr.6: 158637378-158637378 on Build GRCh38
rs946228691 Chr.6: 158637367-158637367 on Build GRCh38
rs969973139 Chr.6: 158637229-158637229 on Build GRCh38
rs796960014 Chr.6: 158637513-158637515 on Build GRCh38
rs747177314 Chr.6: 158637038-158637038 on Build GRCh38
rs556355151 Chr.6: 158637018-158637018 on Build GRCh38
rs913386956 Chr.6: 158637340-158637340 on Build GRCh38
rs750589506 Chr.6: 158637242-158637242 on Build GRCh38
rs758692460 Chr.6: 158637028-158637028 on Build GRCh38
rs931306601 Chr.6: 158637280-158637280 on Build GRCh38
rs752078518 Chr.6: 158637117-158637118 on Build GRCh38
rs769842892 Chr.6: 158637497-158637497 on Build GRCh38
rs779180781 Chr.6: 158637254-158637254 on Build GRCh38
rs751742683 Chr.6: 158637135-158637135 on Build GRCh38
rs530155809 Chr.6: 158637368-158637368 on Build GRCh38
rs768908895 Chr.6: 158637195-158637195 on Build GRCh38
rs919877970 Chr.6: 158637276-158637276 on Build GRCh38
rs376253495 Chr.6: 158637251-158637251 on Build GRCh38
rs952591108 Chr.6: 158637122-158637129 on Build GRCh38
rs965703230 Chr.6: 158637026-158637026 on Build GRCh38
rs767683046 Chr.6: 158637240-158637240 on Build GRCh38
rs930705463 Chr.6: 158637312-158637312 on Build GRCh38
rs749729960 Chr.6: 158637190-158637190 on Build GRCh38
rs781307360 Chr.6: 158637454-158637454 on Build GRCh38
rs777358326 Chr.6: 158637207-158637208 on Build GRCh38
rs541921345 Chr.6: 158637065-158637065 on Build GRCh38
rs904809280 Chr.6: 158637379-158637380 on Build GRCh38
rs1000497962 Chr.6: 158637409-158637409 on Build GRCh38
rs759504987 Chr.6: 158637094-158637094 on Build GRCh38
rs147885568 Chr.6: 158637237-158637237 on Build GRCh38
rs1019459035 Chr.6: 158637222-158637222 on Build GRCh38
rs752592189 Chr.6: 158637111-158637111 on Build GRCh38
rs757955371 Chr.6: 158637207-158637211 on Build GRCh38
rs756734827 Chr.6: 158637167-158637167 on Build GRCh38
rs760167769 Chr.6: 158637078-158637078 on Build GRCh38
rs566782048 Chr.6: 158637432-158637432 on Build GRCh38
rs752880803 Chr.6: 158637020-158637020 on Build GRCh38
rs546969859 Chr.6: 158637230-158637234 on Build GRCh38
rs776162631 Chr.6: 158637215-158637215 on Build GRCh38
rs144685549 Chr.6: 158637218-158637218 on Build GRCh38
rs562695198 Chr.6: 158637331-158637331 on Build GRCh38
rs938819081 Chr.6: 158637042-158637042 on Build GRCh38
rs113541579 Chr.6: 158637503-158637503 on Build GRCh38
rs749774745 Chr.6: 158637250-158637250 on Build GRCh38
rs776506875 Chr.6: 158637085-158637085 on Build GRCh38
rs892019689 Chr.6: 158637084-158637084 on Build GRCh38
rs532635061 Chr.6: 158637317-158637317 on Build GRCh38
rs764380659 Chr.6: 158637202-158637202 on Build GRCh38
rs748030404 Chr.6: 158637476-158637476 on Build GRCh38
rs780643006 Chr.6: 158637246-158637246 on Build GRCh38
rs746645634 Chr.6: 158637214-158637218 on Build GRCh38
rs781452396 Chr.6: 158637153-158637153 on Build GRCh38
rs527682149 Chr.6: 158637451-158637451 on Build GRCh38
rs1007809309 Chr.6: 158637358-158637358 on Build GRCh38

View Full Product Details