Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 30012105-30012703 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FKBP14
Entrez Gene ID
55033
Gene Name
FK506 binding protein 14
Gene Aliases
EDSKMH, FKBP22, IPBP12
UniGene
Hs.390838
Gene Chromosome Location
Chr.7: 30010583-30026801 on Build GRCh38
Transcripts
NM_017946.3 NR_046478.1 NR_046479.1

SNP Detail

SNP ID Chromosome Location
rs930343405 Chr.7: 30012143-30012143 on Build GRCh38
rs944849875 Chr.7: 30012228-30012228 on Build GRCh38
rs1003757715 Chr.7: 30012365-30012365 on Build GRCh38
rs188998675 Chr.7: 30012565-30012565 on Build GRCh38
rs948705826 Chr.7: 30012590-30012590 on Build GRCh38
rs1034877645 Chr.7: 30012372-30012372 on Build GRCh38
rs761927739 Chr.7: 30012580-30012580 on Build GRCh38
rs181137295 Chr.7: 30012652-30012652 on Build GRCh38
rs758323701 Chr.7: 30012567-30012567 on Build GRCh38
rs1026088677 Chr.7: 30012129-30012131 on Build GRCh38
rs769198507 Chr.7: 30012483-30012483 on Build GRCh38
rs56094929 Chr.7: 30012536-30012536 on Build GRCh38
rs781728051 Chr.7: 30012259-30012259 on Build GRCh38
rs1044387362 Chr.7: 30012600-30012600 on Build GRCh38
rs552729005 Chr.7: 30012629-30012629 on Build GRCh38
rs184238567 Chr.7: 30012249-30012249 on Build GRCh38
rs923163503 Chr.7: 30012398-30012398 on Build GRCh38
rs146908974 Chr.7: 30012412-30012412 on Build GRCh38
rs976635550 Chr.7: 30012512-30012512 on Build GRCh38
rs985815752 Chr.7: 30012448-30012448 on Build GRCh38
rs959621870 Chr.7: 30012217-30012217 on Build GRCh38
rs934875682 Chr.7: 30012547-30012547 on Build GRCh38
rs540214225 Chr.7: 30012666-30012666 on Build GRCh38
rs555974302 Chr.7: 30012413-30012413 on Build GRCh38
rs1040578183 Chr.7: 30012232-30012232 on Build GRCh38
rs907257331 Chr.7: 30012610-30012610 on Build GRCh38
rs775939136 Chr.7: 30012308-30012308 on Build GRCh38
rs746347649 Chr.7: 30012393-30012393 on Build GRCh38
rs769982543 Chr.7: 30012510-30012510 on Build GRCh38
rs139320690 Chr.7: 30012682-30012682 on Build GRCh38
rs962231258 Chr.7: 30012385-30012385 on Build GRCh38
rs538965979 Chr.7: 30012360-30012360 on Build GRCh38
rs893429552 Chr.7: 30012579-30012579 on Build GRCh38
rs924083514 Chr.7: 30012323-30012323 on Build GRCh38
rs534147666 Chr.7: 30012141-30012141 on Build GRCh38
rs972115893 Chr.7: 30012130-30012130 on Build GRCh38
rs893146592 Chr.7: 30012297-30012297 on Build GRCh38
rs1023004041 Chr.7: 30012306-30012306 on Build GRCh38
rs1011848323 Chr.7: 30012534-30012534 on Build GRCh38
rs972013833 Chr.7: 30012397-30012397 on Build GRCh38
rs71528593 Chr.7: 30012624-30012624 on Build GRCh38
rs886231466 Chr.7: 30012452-30012452 on Build GRCh38
rs747474125 Chr.7: 30012349-30012349 on Build GRCh38
rs985867822 Chr.7: 30012162-30012162 on Build GRCh38
rs879313380 Chr.7: 30012233-30012233 on Build GRCh38
rs999738325 Chr.7: 30012582-30012582 on Build GRCh38
rs1005879060 Chr.7: 30012151-30012151 on Build GRCh38
rs575477692 Chr.7: 30012447-30012447 on Build GRCh38
rs910288865 Chr.7: 30012463-30012463 on Build GRCh38
rs897648159 Chr.7: 30012672-30012672 on Build GRCh38
rs182397562 Chr.7: 30012179-30012179 on Build GRCh38
rs1012905173 Chr.7: 30012304-30012304 on Build GRCh38

View Full Product Details